Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150966032-150966534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS2
Entrez Gene ID
29956
Gene Name
ceramide synthase 2
Gene Aliases
L3, LASS2, SP260, TMSG1
UniGene
Hs.744017
Gene Chromosome Location
Chr.1: 150965173-150975003 on Build GRCh38
Transcripts
NM_022075.4 NM_181746.3 XM_011509451.2 XM_011509452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1333904 c.1110delC p.I371fs*>10 Chr.1: 150966181-150966181 on Build GRCh38
895644 c.1059G>A p.E353E Chr.1: 150966232-150966232 on Build GRCh38
3473541 c.1080A>G p.A360A Chr.1: 150966211-150966211 on Build GRCh38
5511565 c.1073delG p.G358fs*8 Chr.1: 150966218-150966218 on Build GRCh38
4647331 c.1093C>A p.L365I Chr.1: 150966198-150966198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78204240 Chr.1: 150966384-150966384 on Build GRCh38
rs748858848 Chr.1: 150966252-150966252 on Build GRCh38
rs374080024 Chr.1: 150966315-150966315 on Build GRCh38
rs774113710 Chr.1: 150966203-150966203 on Build GRCh38
rs759357107 Chr.1: 150966298-150966298 on Build GRCh38
rs886809193 Chr.1: 150966081-150966081 on Build GRCh38
rs753008002 Chr.1: 150966206-150966206 on Build GRCh38
rs955569375 Chr.1: 150966389-150966389 on Build GRCh38
rs1050845471 Chr.1: 150966060-150966060 on Build GRCh38
rs758837601 Chr.1: 150966238-150966238 on Build GRCh38
rs754659366 Chr.1: 150966103-150966103 on Build GRCh38
rs910895297 Chr.1: 150966349-150966349 on Build GRCh38
rs780873258 Chr.1: 150966167-150966167 on Build GRCh38
rs779228104 Chr.1: 150966161-150966161 on Build GRCh38
rs587657587 Chr.1: 150966162-150966162 on Build GRCh38
rs376324977 Chr.1: 150966240-150966240 on Build GRCh38
rs754066846 Chr.1: 150966318-150966318 on Build GRCh38
rs41308401 Chr.1: 150966401-150966401 on Build GRCh38
rs777422206 Chr.1: 150966446-150966446 on Build GRCh38
rs745874646 Chr.1: 150966456-150966456 on Build GRCh38
rs905215139 Chr.1: 150966476-150966476 on Build GRCh38
rs373644107 Chr.1: 150966364-150966364 on Build GRCh38
rs138395828 Chr.1: 150966204-150966204 on Build GRCh38
rs969429316 Chr.1: 150966212-150966212 on Build GRCh38
rs935184835 Chr.1: 150966406-150966406 on Build GRCh38
rs757246256 Chr.1: 150966222-150966222 on Build GRCh38
rs753978441 Chr.1: 150966221-150966221 on Build GRCh38
rs770904128 Chr.1: 150966202-150966202 on Build GRCh38
rs747771528 Chr.1: 150966172-150966172 on Build GRCh38
rs772698981 Chr.1: 150966175-150966175 on Build GRCh38
rs1016420014 Chr.1: 150966263-150966263 on Build GRCh38
rs774392793 Chr.1: 150966264-150966264 on Build GRCh38
rs267739 Chr.1: 150966140-150966140 on Build GRCh38
rs778013552 Chr.1: 150966304-150966306 on Build GRCh38
rs1028920087 Chr.1: 150966387-150966387 on Build GRCh38
rs1021184559 Chr.1: 150966399-150966400 on Build GRCh38
rs181377166 Chr.1: 150966419-150966419 on Build GRCh38
rs778281337 Chr.1: 150966455-150966455 on Build GRCh38
rs587715315 Chr.1: 150966394-150966394 on Build GRCh38
rs979853656 Chr.1: 150966421-150966421 on Build GRCh38
rs756828166 Chr.1: 150966453-150966453 on Build GRCh38
rs765824453 Chr.1: 150966291-150966292 on Build GRCh38
rs142260553 Chr.1: 150966487-150966487 on Build GRCh38
rs767189622 Chr.1: 150966205-150966205 on Build GRCh38
rs746824537 Chr.1: 150966486-150966486 on Build GRCh38
rs769453538 Chr.1: 150966241-150966241 on Build GRCh38
rs765201542 Chr.1: 150966326-150966326 on Build GRCh38
rs772878867 Chr.1: 150966102-150966105 on Build GRCh38
rs1003962586 Chr.1: 150966086-150966086 on Build GRCh38
rs757767614 Chr.1: 150966158-150966158 on Build GRCh38
rs991304680 Chr.1: 150966484-150966484 on Build GRCh38
rs762003346 Chr.1: 150966321-150966321 on Build GRCh38
rs942413824 Chr.1: 150966360-150966360 on Build GRCh38
rs772302151 Chr.1: 150966356-150966356 on Build GRCh38
rs775411798 Chr.1: 150966306-150966306 on Build GRCh38
rs199730233 Chr.1: 150966433-150966433 on Build GRCh38
rs925368333 Chr.1: 150966431-150966431 on Build GRCh38
rs8444 Chr.1: 150966095-150966095 on Build GRCh38
rs188189438 Chr.1: 150966336-150966336 on Build GRCh38
rs769422073 Chr.1: 150966407-150966407 on Build GRCh38
rs144130462 Chr.1: 150966216-150966216 on Build GRCh38
rs370703954 Chr.1: 150966262-150966262 on Build GRCh38
rs184134274 Chr.1: 150966505-150966505 on Build GRCh38
rs889570933 Chr.1: 150966071-150966071 on Build GRCh38
rs779497928 Chr.1: 150966227-150966227 on Build GRCh38
rs999420969 Chr.1: 150966133-150966133 on Build GRCh38
rs986905134 Chr.1: 150966343-150966343 on Build GRCh38
rs751386100 Chr.1: 150966100-150966100 on Build GRCh38
rs750873215 Chr.1: 150966229-150966229 on Build GRCh38
rs766147664 Chr.1: 150966098-150966098 on Build GRCh38
rs945416946 Chr.1: 150966199-150966199 on Build GRCh38
rs587736236 Chr.1: 150966373-150966373 on Build GRCh38
rs760487191 Chr.1: 150966307-150966307 on Build GRCh38
rs113620024 Chr.1: 150966475-150966475 on Build GRCh38
rs777343723 Chr.1: 150966247-150966247 on Build GRCh38
rs768378257 Chr.1: 150966146-150966146 on Build GRCh38
rs996988586 Chr.1: 150966385-150966385 on Build GRCh38
rs896429789 Chr.1: 150966417-150966417 on Build GRCh38
rs748866062 Chr.1: 150966447-150966447 on Build GRCh38
rs767808328 Chr.1: 150966109-150966109 on Build GRCh38
rs772147627 Chr.1: 150966462-150966462 on Build GRCh38
rs141621672 Chr.1: 150966173-150966173 on Build GRCh38
rs374432724 Chr.1: 150966136-150966136 on Build GRCh38
rs775499785 Chr.1: 150966482-150966482 on Build GRCh38
rs755293263 Chr.1: 150966442-150966442 on Build GRCh38
rs771969321 Chr.1: 150966305-150966305 on Build GRCh38
rs1043506238 Chr.1: 150966097-150966097 on Build GRCh38
rs147381709 Chr.1: 150966211-150966211 on Build GRCh38
rs955626155 Chr.1: 150966153-150966153 on Build GRCh38
rs756307512 Chr.1: 150966132-150966132 on Build GRCh38
rs762403970 Chr.1: 150966236-150966237 on Build GRCh38
rs762393823 Chr.1: 150966198-150966198 on Build GRCh38
rs1013741340 Chr.1: 150966196-150966197 on Build GRCh38
rs200766311 Chr.1: 150966335-150966335 on Build GRCh38
rs768549889 Chr.1: 150966497-150966497 on Build GRCh38

View Full Product Details