Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150967300-150967795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS2
Entrez Gene ID
29956
Gene Name
ceramide synthase 2
Gene Aliases
L3, LASS2, SP260, TMSG1
UniGene
Hs.744017
Gene Chromosome Location
Chr.1: 150965173-150975003 on Build GRCh38
Transcripts
NM_022075.4 NM_181746.3 XM_011509451.2 XM_011509452.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057884 c.519G>C p.Q173H Chr.1: 150967664-150967664 on Build GRCh38
895650 c.607C>T p.R203* Chr.1: 150967397-150967397 on Build GRCh38
5414760 c.511C>G p.P171A Chr.1: 150967672-150967672 on Build GRCh38
1333905 c.472C>A p.P158T Chr.1: 150967711-150967711 on Build GRCh38
675515 c.606G>T p.K202N Chr.1: 150967398-150967398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755153222 Chr.1: 150967502-150967502 on Build GRCh38
rs200270203 Chr.1: 150967651-150967651 on Build GRCh38
rs757091487 Chr.1: 150967354-150967354 on Build GRCh38
rs587756431 Chr.1: 150967555-150967555 on Build GRCh38
rs374297994 Chr.1: 150967710-150967710 on Build GRCh38
rs750803755 Chr.1: 150967345-150967345 on Build GRCh38
rs759815286 Chr.1: 150967703-150967703 on Build GRCh38
rs777933110 Chr.1: 150967518-150967518 on Build GRCh38
rs779805039 Chr.1: 150967640-150967640 on Build GRCh38
rs778365289 Chr.1: 150967728-150967728 on Build GRCh38
rs142698152 Chr.1: 150967696-150967696 on Build GRCh38
rs771756858 Chr.1: 150967464-150967464 on Build GRCh38
rs372956606 Chr.1: 150967610-150967610 on Build GRCh38
rs760073090 Chr.1: 150967717-150967717 on Build GRCh38
rs587636291 Chr.1: 150967395-150967395 on Build GRCh38
rs587661907 Chr.1: 150967636-150967636 on Build GRCh38
rs587691063 Chr.1: 150967498-150967498 on Build GRCh38
rs1053959967 Chr.1: 150967701-150967701 on Build GRCh38
rs769273032 Chr.1: 150967750-150967750 on Build GRCh38
rs1027295156 Chr.1: 150967576-150967576 on Build GRCh38
rs771824277 Chr.1: 150967429-150967429 on Build GRCh38
rs769756473 Chr.1: 150967691-150967691 on Build GRCh38
rs985058601 Chr.1: 150967605-150967605 on Build GRCh38
rs879040094 Chr.1: 150967606-150967606 on Build GRCh38
rs1000373232 Chr.1: 150967635-150967635 on Build GRCh38
rs750669002 Chr.1: 150967371-150967371 on Build GRCh38
rs762857759 Chr.1: 150967694-150967694 on Build GRCh38
rs932282351 Chr.1: 150967325-150967325 on Build GRCh38
rs747387121 Chr.1: 150967591-150967591 on Build GRCh38
rs760253427 Chr.1: 150967470-150967470 on Build GRCh38
rs1042153516 Chr.1: 150967403-150967403 on Build GRCh38
rs373275284 Chr.1: 150967529-150967529 on Build GRCh38
rs768132524 Chr.1: 150967478-150967478 on Build GRCh38
rs199945675 Chr.1: 150967418-150967418 on Build GRCh38
rs764049020 Chr.1: 150967349-150967349 on Build GRCh38
rs757679779 Chr.1: 150967735-150967735 on Build GRCh38
rs777352515 Chr.1: 150967733-150967737 on Build GRCh38
rs749555667 Chr.1: 150967454-150967454 on Build GRCh38
rs771149364 Chr.1: 150967638-150967638 on Build GRCh38
rs917700729 Chr.1: 150967385-150967385 on Build GRCh38
rs756219712 Chr.1: 150967727-150967727 on Build GRCh38
rs750238584 Chr.1: 150967494-150967494 on Build GRCh38
rs780710247 Chr.1: 150967413-150967413 on Build GRCh38
rs1052279259 Chr.1: 150967571-150967571 on Build GRCh38
rs370622852 Chr.1: 150967667-150967667 on Build GRCh38
rs587726816 Chr.1: 150967344-150967344 on Build GRCh38
rs948275539 Chr.1: 150967770-150967770 on Build GRCh38
rs772977125 Chr.1: 150967693-150967693 on Build GRCh38
rs762616596 Chr.1: 150967496-150967496 on Build GRCh38
rs751879336 Chr.1: 150967700-150967700 on Build GRCh38
rs904717025 Chr.1: 150967609-150967609 on Build GRCh38
rs747331682 Chr.1: 150967520-150967538 on Build GRCh38
rs148487744 Chr.1: 150967668-150967668 on Build GRCh38
rs746609596 Chr.1: 150967644-150967644 on Build GRCh38
rs776132025 Chr.1: 150967663-150967663 on Build GRCh38
rs1004966474 Chr.1: 150967519-150967519 on Build GRCh38
rs752826221 Chr.1: 150967726-150967726 on Build GRCh38
rs765013119 Chr.1: 150967361-150967361 on Build GRCh38
rs78875607 Chr.1: 150967407-150967407 on Build GRCh38
rs376323259 Chr.1: 150967720-150967720 on Build GRCh38
rs943746897 Chr.1: 150967620-150967620 on Build GRCh38
rs115425823 Chr.1: 150967336-150967336 on Build GRCh38
rs773694437 Chr.1: 150967440-150967440 on Build GRCh38
rs758661949 Chr.1: 150967382-150967382 on Build GRCh38
rs754275107 Chr.1: 150967732-150967732 on Build GRCh38
rs912274651 Chr.1: 150967355-150967355 on Build GRCh38
rs781421533 Chr.1: 150967509-150967509 on Build GRCh38
rs940585 Chr.1: 150967525-150967525 on Build GRCh38
rs753724848 Chr.1: 150967352-150967352 on Build GRCh38
rs771377355 Chr.1: 150967607-150967607 on Build GRCh38
rs587708607 Chr.1: 150967326-150967326 on Build GRCh38
rs1007606190 Chr.1: 150967577-150967577 on Build GRCh38
rs1012214039 Chr.1: 150967508-150967508 on Build GRCh38
rs778072497 Chr.1: 150967534-150967534 on Build GRCh38
rs919223742 Chr.1: 150967552-150967552 on Build GRCh38
rs753310629 Chr.1: 150967512-150967512 on Build GRCh38
rs765101077 Chr.1: 150967488-150967488 on Build GRCh38
rs774963430 Chr.1: 150967467-150967467 on Build GRCh38
rs769034701 Chr.1: 150967526-150967526 on Build GRCh38
rs779140092 Chr.1: 150967771-150967771 on Build GRCh38
rs761726667 Chr.1: 150967487-150967487 on Build GRCh38
rs770345018 Chr.1: 150967435-150967435 on Build GRCh38
rs587610841 Chr.1: 150967573-150967573 on Build GRCh38

View Full Product Details