Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151400358-151400869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMB4
Entrez Gene ID
5692
Gene Name
proteasome subunit beta 4
Gene Aliases
HN3, HsN3, PROS-26, PROS26
UniGene
Hs.89545
Gene Chromosome Location
Chr.1: 151399565-151401936 on Build GRCh38
Transcripts
NM_002796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
895799 c.375A>G p.G125G Chr.1: 151400469-151400469 on Build GRCh38
1626426 c.519T>C p.L173L Chr.1: 151400788-151400788 on Build GRCh38
895800 c.547G>A p.A183T Chr.1: 151400816-151400816 on Build GRCh38
246836 c.428G>A p.S143N Chr.1: 151400522-151400522 on Build GRCh38
4546104 c.392G>A p.R131K Chr.1: 151400486-151400486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746668281 Chr.1: 151400613-151400613 on Build GRCh38
rs763749279 Chr.1: 151400728-151400728 on Build GRCh38
rs778248319 Chr.1: 151400576-151400576 on Build GRCh38
rs768849990 Chr.1: 151400420-151400420 on Build GRCh38
rs985352638 Chr.1: 151400699-151400699 on Build GRCh38
rs1804239 Chr.1: 151400812-151400812 on Build GRCh38
rs770299573 Chr.1: 151400632-151400632 on Build GRCh38
rs201438006 Chr.1: 151400518-151400518 on Build GRCh38
rs371591363 Chr.1: 151400621-151400621 on Build GRCh38
rs3209715 Chr.1: 151400839-151400839 on Build GRCh38
rs753664008 Chr.1: 151400572-151400572 on Build GRCh38
rs763162526 Chr.1: 151400541-151400541 on Build GRCh38
rs750425737 Chr.1: 151400770-151400770 on Build GRCh38
rs754687002 Chr.1: 151400791-151400791 on Build GRCh38
rs866266207 Chr.1: 151400816-151400816 on Build GRCh38
rs763115700 Chr.1: 151400647-151400647 on Build GRCh38
rs894848845 Chr.1: 151400573-151400573 on Build GRCh38
rs765134636 Chr.1: 151400603-151400603 on Build GRCh38
rs777632466 Chr.1: 151400566-151400566 on Build GRCh38
rs923115397 Chr.1: 151400431-151400431 on Build GRCh38
rs775164354 Chr.1: 151400477-151400477 on Build GRCh38
rs757837361 Chr.1: 151400771-151400771 on Build GRCh38
rs368176363 Chr.1: 151400737-151400737 on Build GRCh38
rs534278528 Chr.1: 151400702-151400702 on Build GRCh38
rs533890009 Chr.1: 151400629-151400629 on Build GRCh38
rs770110723 Chr.1: 151400841-151400841 on Build GRCh38
rs2495391 Chr.1: 151400724-151400724 on Build GRCh38
rs17407993 Chr.1: 151400835-151400835 on Build GRCh38
rs11557385 Chr.1: 151400826-151400826 on Build GRCh38
rs745352663 Chr.1: 151400587-151400587 on Build GRCh38
rs750544134 Chr.1: 151400522-151400522 on Build GRCh38
rs761013630 Chr.1: 151400486-151400486 on Build GRCh38
rs766637481 Chr.1: 151400648-151400648 on Build GRCh38
rs778500444 Chr.1: 151400449-151400449 on Build GRCh38
rs1005931511 Chr.1: 151400715-151400715 on Build GRCh38
rs910660327 Chr.1: 151400519-151400519 on Build GRCh38
rs762393225 Chr.1: 151400515-151400515 on Build GRCh38
rs745464020 Chr.1: 151400472-151400472 on Build GRCh38
rs367717074 Chr.1: 151400524-151400524 on Build GRCh38
rs769075367 Chr.1: 151400490-151400490 on Build GRCh38
rs753700156 Chr.1: 151400730-151400730 on Build GRCh38
rs781090622 Chr.1: 151400397-151400397 on Build GRCh38
rs933171351 Chr.1: 151400445-151400445 on Build GRCh38
rs773855644 Chr.1: 151400845-151400845 on Build GRCh38
rs145190029 Chr.1: 151400442-151400442 on Build GRCh38
rs72996022 Chr.1: 151400687-151400687 on Build GRCh38
rs777075470 Chr.1: 151400625-151400625 on Build GRCh38
rs777562522 Chr.1: 151400422-151400422 on Build GRCh38
rs112085237 Chr.1: 151400589-151400589 on Build GRCh38
rs752505278 Chr.1: 151400400-151400400 on Build GRCh38
rs779677108 Chr.1: 151400607-151400607 on Build GRCh38
rs748559526 Chr.1: 151400631-151400631 on Build GRCh38
rs3209764 Chr.1: 151400539-151400539 on Build GRCh38
rs757042202 Chr.1: 151400757-151400757 on Build GRCh38
rs14276 Chr.1: 151400544-151400544 on Build GRCh38
rs3209765 Chr.1: 151400582-151400582 on Build GRCh38
rs921919023 Chr.1: 151400684-151400684 on Build GRCh38
rs756026557 Chr.1: 151400414-151400414 on Build GRCh38
rs973933466 Chr.1: 151400681-151400682 on Build GRCh38
rs902384505 Chr.1: 151400800-151400801 on Build GRCh38
rs1052721 Chr.1: 151400650-151400650 on Build GRCh38
rs757952274 Chr.1: 151400605-151400605 on Build GRCh38
rs765179156 Chr.1: 151400520-151400520 on Build GRCh38
rs746614860 Chr.1: 151400789-151400789 on Build GRCh38
rs776935448 Chr.1: 151400491-151400491 on Build GRCh38
rs371316814 Chr.1: 151400560-151400560 on Build GRCh38
rs114882311 Chr.1: 151400553-151400553 on Build GRCh38
rs748519387 Chr.1: 151400817-151400817 on Build GRCh38
rs1013543612 Chr.1: 151400658-151400658 on Build GRCh38
rs757243651 Chr.1: 151400575-151400575 on Build GRCh38
rs763305194 Chr.1: 151400802-151400803 on Build GRCh38
rs11557382 Chr.1: 151400811-151400811 on Build GRCh38
rs371862894 Chr.1: 151400755-151400755 on Build GRCh38
rs773637763 Chr.1: 151400642-151400642 on Build GRCh38
rs961713727 Chr.1: 151400740-151400740 on Build GRCh38
rs1012419260 Chr.1: 151400602-151400602 on Build GRCh38
rs771743580 Chr.1: 151400473-151400473 on Build GRCh38
rs140771260 Chr.1: 151400525-151400525 on Build GRCh38
rs1044814320 Chr.1: 151400667-151400667 on Build GRCh38
rs993355830 Chr.1: 151400799-151400799 on Build GRCh38
rs751892201 Chr.1: 151400565-151400565 on Build GRCh38
rs765199993 Chr.1: 151400759-151400759 on Build GRCh38
rs770282474 Chr.1: 151400448-151400448 on Build GRCh38
rs779551838 Chr.1: 151400776-151400776 on Build GRCh38
rs773282174 Chr.1: 151400638-151400639 on Build GRCh38

View Full Product Details