Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 1394950-1395454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXF2
Entrez Gene ID
2295
Gene Name
forkhead box F2
Gene Aliases
FKHL6, FREAC-2, FREAC2
UniGene
Hs.484423
Gene Chromosome Location
Chr.6: 1389834-1395597 on Build GRCh38
Transcripts
NM_001452.1

SNP Detail

SNP ID Chromosome Location
rs1047588801 Chr.6: 1394999-1394999 on Build GRCh38
rs1019279644 Chr.6: 1395073-1395073 on Build GRCh38
rs931228168 Chr.6: 1395153-1395153 on Build GRCh38
rs574899172 Chr.6: 1395052-1395052 on Build GRCh38
rs747986541 Chr.6: 1394995-1394995 on Build GRCh38
rs904638699 Chr.6: 1395076-1395076 on Build GRCh38
rs939223731 Chr.6: 1395011-1395011 on Build GRCh38
rs956419750 Chr.6: 1395065-1395065 on Build GRCh38
rs904744575 Chr.6: 1395280-1395280 on Build GRCh38
rs1013571607 Chr.6: 1395000-1395000 on Build GRCh38
rs1043090076 Chr.6: 1395270-1395270 on Build GRCh38
rs1031827644 Chr.6: 1395363-1395363 on Build GRCh38
rs939172897 Chr.6: 1395204-1395206 on Build GRCh38
rs34253622 Chr.6: 1395245-1395245 on Build GRCh38
rs562724614 Chr.6: 1395202-1395203 on Build GRCh38
rs749758264 Chr.6: 1395262-1395262 on Build GRCh38
rs969379671 Chr.6: 1395043-1395043 on Build GRCh38
rs948719869 Chr.6: 1395218-1395218 on Build GRCh38
rs1023250365 Chr.6: 1395015-1395015 on Build GRCh38
rs543498688 Chr.6: 1395055-1395055 on Build GRCh38
rs777435959 Chr.6: 1395124-1395124 on Build GRCh38
rs1031862759 Chr.6: 1395111-1395111 on Build GRCh38
rs1053621145 Chr.6: 1395343-1395343 on Build GRCh38
rs948711070 Chr.6: 1395209-1395209 on Build GRCh38
rs950522531 Chr.6: 1395202-1395202 on Build GRCh38
rs746451374 Chr.6: 1395129-1395129 on Build GRCh38
rs1019415889 Chr.6: 1395402-1395402 on Build GRCh38
rs1031857240 Chr.6: 1395058-1395058 on Build GRCh38
rs973128798 Chr.6: 1395164-1395164 on Build GRCh38
rs965502340 Chr.6: 1395423-1395423 on Build GRCh38
rs138247374 Chr.6: 1395131-1395131 on Build GRCh38
rs983887700 Chr.6: 1395162-1395162 on Build GRCh38
rs1057173024 Chr.6: 1395176-1395176 on Build GRCh38
rs1009060366 Chr.6: 1395400-1395400 on Build GRCh38
rs906427752 Chr.6: 1395001-1395001 on Build GRCh38
rs763037502 Chr.6: 1395241-1395241 on Build GRCh38
rs1000873883 Chr.6: 1395327-1395327 on Build GRCh38
rs765491423 Chr.6: 1395163-1395163 on Build GRCh38
rs544477727 Chr.6: 1395057-1395057 on Build GRCh38
rs1009313209 Chr.6: 1395386-1395386 on Build GRCh38
rs920327986 Chr.6: 1395192-1395194 on Build GRCh38
rs999415655 Chr.6: 1395081-1395081 on Build GRCh38
rs532641043 Chr.6: 1395204-1395204 on Build GRCh38
rs921150251 Chr.6: 1395143-1395143 on Build GRCh38
rs560783420 Chr.6: 1395194-1395194 on Build GRCh38
rs927751392 Chr.6: 1395203-1395204 on Build GRCh38
rs780815575 Chr.6: 1395217-1395217 on Build GRCh38
rs998927135 Chr.6: 1395284-1395284 on Build GRCh38
rs546327553 Chr.6: 1395383-1395383 on Build GRCh38
rs140059590 Chr.6: 1395203-1395204 on Build GRCh38
rs147119747 Chr.6: 1395112-1395112 on Build GRCh38
rs774830890 Chr.6: 1395397-1395397 on Build GRCh38
rs940090076 Chr.6: 1395173-1395173 on Build GRCh38
rs904586759 Chr.6: 1395282-1395282 on Build GRCh38
rs769249752 Chr.6: 1395289-1395289 on Build GRCh38
rs561435854 Chr.6: 1395066-1395066 on Build GRCh38

View Full Product Details