Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10411887-10412397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFAP2A
Entrez Gene ID
7020
Gene Name
transcription factor AP-2 alpha
Gene Aliases
AP-2, AP-2alpha, AP2TF, BOFS, TFAP2
UniGene
Hs.519880
Gene Chromosome Location
Chr.6: 10396677-10419897 on Build GRCh38
Transcripts
NM_001032280.2 NM_001042425.1 NM_003220.2 XM_006715175.2 XM_011514833.2 XM_017011232.1 NR_033910.1

Gene Symbol
TFAP2A-AS1
Entrez Gene ID
100130275
Gene Name
TFAP2A antisense RNA 1
Gene Aliases
-
UniGene
Hs.661431
Gene Chromosome Location
Chr.6: 10412318-10416169 on Build GRCh38
Transcripts
NM_001032280.2 NM_001042425.1 NM_003220.2 XM_006715175.2 XM_011514833.2 XM_017011232.1 NR_033910.1

SNP Detail

SNP ID Chromosome Location
rs1002859992 Chr.6: 10411914-10411914 on Build GRCh38
rs2763140 Chr.6: 10411922-10411922 on Build GRCh38
rs528943997 Chr.6: 10412179-10412179 on Build GRCh38
rs1040602202 Chr.6: 10412150-10412150 on Build GRCh38
rs576989721 Chr.6: 10412207-10412207 on Build GRCh38
rs1018041451 Chr.6: 10411949-10411949 on Build GRCh38
rs985806099 Chr.6: 10412058-10412058 on Build GRCh38
rs909689538 Chr.6: 10412003-10412003 on Build GRCh38
rs984463919 Chr.6: 10412002-10412002 on Build GRCh38
rs556250805 Chr.6: 10411974-10411974 on Build GRCh38
rs1016587055 Chr.6: 10412372-10412372 on Build GRCh38
rs917109274 Chr.6: 10412226-10412226 on Build GRCh38
rs1016278986 Chr.6: 10412131-10412131 on Build GRCh38
rs1675414 Chr.6: 10411955-10411955 on Build GRCh38
rs563290697 Chr.6: 10412242-10412242 on Build GRCh38
rs936242220 Chr.6: 10412113-10412113 on Build GRCh38
rs543565948 Chr.6: 10412137-10412137 on Build GRCh38
rs917329084 Chr.6: 10412048-10412048 on Build GRCh38
rs889304698 Chr.6: 10411931-10411931 on Build GRCh38
rs993667673 Chr.6: 10412146-10412146 on Build GRCh38
rs536291631 Chr.6: 10412009-10412009 on Build GRCh38
rs997857218 Chr.6: 10411973-10411973 on Build GRCh38
rs1053499858 Chr.6: 10412074-10412074 on Build GRCh38
rs1006612070 Chr.6: 10411933-10411933 on Build GRCh38
rs1006189465 Chr.6: 10412130-10412130 on Build GRCh38
rs900910078 Chr.6: 10412154-10412154 on Build GRCh38
rs769611036 Chr.6: 10412299-10412299 on Build GRCh38
rs958302076 Chr.6: 10412343-10412343 on Build GRCh38
rs2763141 Chr.6: 10411978-10411978 on Build GRCh38
rs542107736 Chr.6: 10412239-10412239 on Build GRCh38
rs991494022 Chr.6: 10412032-10412032 on Build GRCh38
rs1046223286 Chr.6: 10412205-10412205 on Build GRCh38
rs374147114 Chr.6: 10412134-10412134 on Build GRCh38
rs902032156 Chr.6: 10412326-10412326 on Build GRCh38
rs141438908 Chr.6: 10412198-10412198 on Build GRCh38
rs924605097 Chr.6: 10412235-10412235 on Build GRCh38
rs994027846 Chr.6: 10412237-10412244 on Build GRCh38
rs772960265 Chr.6: 10412322-10412322 on Build GRCh38
rs1055051744 Chr.6: 10412122-10412122 on Build GRCh38
rs894446205 Chr.6: 10412128-10412128 on Build GRCh38
rs559734334 Chr.6: 10412229-10412229 on Build GRCh38
rs771648656 Chr.6: 10412224-10412224 on Build GRCh38
rs998367112 Chr.6: 10412194-10412194 on Build GRCh38
rs1047259100 Chr.6: 10412339-10412339 on Build GRCh38
rs910815963 Chr.6: 10412170-10412170 on Build GRCh38
rs906439117 Chr.6: 10412103-10412103 on Build GRCh38
rs989166065 Chr.6: 10412192-10412193 on Build GRCh38
rs958777853 Chr.6: 10411952-10411952 on Build GRCh38
rs924628330 Chr.6: 10412099-10412099 on Build GRCh38
rs969993667 Chr.6: 10412323-10412323 on Build GRCh38
rs1016808027 Chr.6: 10412215-10412215 on Build GRCh38
rs1002567370 Chr.6: 10412104-10412104 on Build GRCh38
rs950210520 Chr.6: 10412063-10412063 on Build GRCh38
rs957664118 Chr.6: 10412187-10412187 on Build GRCh38
rs554546052 Chr.6: 10412097-10412097 on Build GRCh38
rs958538637 Chr.6: 10412007-10412007 on Build GRCh38
rs955009968 Chr.6: 10412245-10412245 on Build GRCh38
rs954981677 Chr.6: 10412156-10412156 on Build GRCh38
rs376944947 Chr.6: 10412220-10412220 on Build GRCh38
rs997918069 Chr.6: 10412329-10412329 on Build GRCh38
rs530497820 Chr.6: 10412295-10412295 on Build GRCh38
rs943630587 Chr.6: 10412286-10412286 on Build GRCh38
rs1005626191 Chr.6: 10412347-10412347 on Build GRCh38
rs1017848292 Chr.6: 10412332-10412332 on Build GRCh38
rs945170334 Chr.6: 10412087-10412087 on Build GRCh38
rs538409765 Chr.6: 10412200-10412200 on Build GRCh38
rs76414670 Chr.6: 10412234-10412234 on Build GRCh38
rs892234718 Chr.6: 10412075-10412075 on Build GRCh38
rs554079745 Chr.6: 10411929-10411929 on Build GRCh38
rs886736917 Chr.6: 10412345-10412345 on Build GRCh38
rs539202570 Chr.6: 10411927-10411927 on Build GRCh38
rs933596129 Chr.6: 10412057-10412057 on Build GRCh38
rs951312640 Chr.6: 10411980-10411980 on Build GRCh38
rs866536241 Chr.6: 10412237-10412237 on Build GRCh38
rs1055422439 Chr.6: 10412223-10412223 on Build GRCh38
rs547892972 Chr.6: 10412230-10412230 on Build GRCh38
rs923528214 Chr.6: 10412018-10412018 on Build GRCh38
rs910799689 Chr.6: 10412251-10412251 on Build GRCh38
rs1040548164 Chr.6: 10412305-10412305 on Build GRCh38
rs986749621 Chr.6: 10412160-10412160 on Build GRCh38
rs1007731514 Chr.6: 10412145-10412145 on Build GRCh38
rs541202974 Chr.6: 10412221-10412221 on Build GRCh38
rs767807606 Chr.6: 10412167-10412167 on Build GRCh38

View Full Product Details