Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13583836-13584338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT5
Entrez Gene ID
23408
Gene Name
sirtuin 5
Gene Aliases
SIR2L5
UniGene
Hs.567431
Gene Chromosome Location
Chr.6: 13574222-13615158 on Build GRCh38
Transcripts
NM_001193267.2 NM_001242827.1 NM_012241.4 NM_031244.3 XM_005248968.4 XM_011514444.2 XM_017010620.1 XM_017010621.1 XM_017010622.1 XM_017010623.1 XM_017010624.1 XM_017010625.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4882312 c.12C>A p.L4L Chr.6: 13584122-13584122 on Build GRCh38
3920768 c.38C>T p.S13F Chr.6: 13584148-13584148 on Build GRCh38
6778851 c.68C>T p.A23V Chr.6: 13584178-13584178 on Build GRCh38
4726204 c.77G>A p.R26Q Chr.6: 13584187-13584187 on Build GRCh38
7289407 c.78A>G p.R26R Chr.6: 13584188-13584188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201240980 Chr.6: 13584261-13584261 on Build GRCh38
rs200174442 Chr.6: 13584174-13584174 on Build GRCh38
rs537093573 Chr.6: 13583884-13583884 on Build GRCh38
rs939127615 Chr.6: 13584059-13584059 on Build GRCh38
rs144658395 Chr.6: 13584213-13584213 on Build GRCh38
rs758939301 Chr.6: 13584127-13584127 on Build GRCh38
rs759819026 Chr.6: 13584204-13584204 on Build GRCh38
rs962904261 Chr.6: 13583905-13583905 on Build GRCh38
rs745997920 Chr.6: 13584125-13584125 on Build GRCh38
rs11967464 Chr.6: 13583975-13583975 on Build GRCh38
rs751734951 Chr.6: 13584064-13584065 on Build GRCh38
rs530703942 Chr.6: 13584309-13584309 on Build GRCh38
rs1032215698 Chr.6: 13583934-13583934 on Build GRCh38
rs957849849 Chr.6: 13583937-13583937 on Build GRCh38
rs879598364 Chr.6: 13583974-13583974 on Build GRCh38
rs775076594 Chr.6: 13584071-13584071 on Build GRCh38
rs763811847 Chr.6: 13584079-13584079 on Build GRCh38
rs200888688 Chr.6: 13584114-13584114 on Build GRCh38
rs1016718272 Chr.6: 13583878-13583878 on Build GRCh38
rs76198501 Chr.6: 13584075-13584075 on Build GRCh38
rs895048268 Chr.6: 13583861-13583861 on Build GRCh38
rs755253508 Chr.6: 13584118-13584118 on Build GRCh38
rs756307658 Chr.6: 13584111-13584111 on Build GRCh38
rs774986916 Chr.6: 13584145-13584145 on Build GRCh38
rs955621994 Chr.6: 13584216-13584216 on Build GRCh38
rs375691582 Chr.6: 13583992-13583992 on Build GRCh38
rs147272755 Chr.6: 13584141-13584141 on Build GRCh38
rs867262607 Chr.6: 13584214-13584214 on Build GRCh38
rs200243694 Chr.6: 13584078-13584078 on Build GRCh38
rs571359010 Chr.6: 13583858-13583858 on Build GRCh38
rs964078585 Chr.6: 13584290-13584290 on Build GRCh38
rs767424598 Chr.6: 13584086-13584086 on Build GRCh38
rs199906141 Chr.6: 13584240-13584240 on Build GRCh38
rs202210550 Chr.6: 13584269-13584269 on Build GRCh38
rs139160010 Chr.6: 13584124-13584124 on Build GRCh38
rs570067378 Chr.6: 13584126-13584126 on Build GRCh38
rs200955506 Chr.6: 13584186-13584186 on Build GRCh38
rs1012149668 Chr.6: 13583877-13583877 on Build GRCh38
rs182075633 Chr.6: 13584041-13584041 on Build GRCh38
rs994226296 Chr.6: 13583918-13583918 on Build GRCh38
rs200688934 Chr.6: 13584154-13584154 on Build GRCh38
rs1036632643 Chr.6: 13584060-13584060 on Build GRCh38
rs779529411 Chr.6: 13584271-13584271 on Build GRCh38
rs914743154 Chr.6: 13584000-13584000 on Build GRCh38
rs1003461171 Chr.6: 13584282-13584282 on Build GRCh38
rs769414882 Chr.6: 13584068-13584068 on Build GRCh38
rs199570411 Chr.6: 13584098-13584098 on Build GRCh38
rs201550893 Chr.6: 13584033-13584033 on Build GRCh38
rs142111138 Chr.6: 13584083-13584083 on Build GRCh38
rs201478760 Chr.6: 13584226-13584226 on Build GRCh38
rs1020775105 Chr.6: 13584220-13584220 on Build GRCh38
rs753978673 Chr.6: 13584193-13584193 on Build GRCh38
rs140708883 Chr.6: 13584155-13584155 on Build GRCh38
rs367642009 Chr.6: 13584291-13584291 on Build GRCh38
rs774195427 Chr.6: 13584170-13584170 on Build GRCh38
rs751999295 Chr.6: 13584263-13584263 on Build GRCh38
rs544275137 Chr.6: 13583921-13583921 on Build GRCh38
rs774200473 Chr.6: 13584023-13584023 on Build GRCh38
rs528262167 Chr.6: 13584009-13584009 on Build GRCh38
rs753084607 Chr.6: 13584223-13584223 on Build GRCh38
rs35634029 Chr.6: 13584187-13584188 on Build GRCh38
rs1008344642 Chr.6: 13584254-13584254 on Build GRCh38
rs113600137 Chr.6: 13584172-13584172 on Build GRCh38
rs748867606 Chr.6: 13584273-13584273 on Build GRCh38
rs575619893 Chr.6: 13583895-13583895 on Build GRCh38
rs768327939 Chr.6: 13584276-13584276 on Build GRCh38
rs771018860 Chr.6: 13583945-13583945 on Build GRCh38
rs200135552 Chr.6: 13584179-13584179 on Build GRCh38
rs11965173 Chr.6: 13583962-13583962 on Build GRCh38
rs143890704 Chr.6: 13584138-13584138 on Build GRCh38
rs754070971 Chr.6: 13584116-13584116 on Build GRCh38
rs558327575 Chr.6: 13583933-13583933 on Build GRCh38
rs958822842 Chr.6: 13584011-13584011 on Build GRCh38
rs923705544 Chr.6: 13584055-13584055 on Build GRCh38
rs2804917 Chr.6: 13583969-13583969 on Build GRCh38
rs750480737 Chr.6: 13584097-13584097 on Build GRCh38
rs565882123 Chr.6: 13584106-13584106 on Build GRCh38
rs778193163 Chr.6: 13584128-13584128 on Build GRCh38
rs150779718 Chr.6: 13584115-13584115 on Build GRCh38
rs964275751 Chr.6: 13583979-13583979 on Build GRCh38
rs1035689693 Chr.6: 13584285-13584285 on Build GRCh38

View Full Product Details