Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 21597738-21598250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX4
Entrez Gene ID
6659
Gene Name
SRY-box 4
Gene Aliases
EVI16
UniGene
Hs.643910
Gene Chromosome Location
Chr.6: 21593741-21598619 on Build GRCh38
Transcripts
NM_003107.2

SNP Detail

SNP ID Chromosome Location
rs925228895 Chr.6: 21597903-21597903 on Build GRCh38
rs1015168588 Chr.6: 21597966-21597966 on Build GRCh38
rs942033488 Chr.6: 21598056-21598056 on Build GRCh38
rs774378839 Chr.6: 21597913-21597913 on Build GRCh38
rs778685534 Chr.6: 21598194-21598194 on Build GRCh38
rs879249672 Chr.6: 21598193-21598193 on Build GRCh38
rs1053331039 Chr.6: 21598174-21598174 on Build GRCh38
rs985834335 Chr.6: 21598072-21598072 on Build GRCh38
rs891577417 Chr.6: 21597836-21597836 on Build GRCh38
rs898053220 Chr.6: 21598061-21598062 on Build GRCh38
rs1050980175 Chr.6: 21598040-21598040 on Build GRCh38
rs562700047 Chr.6: 21598006-21598006 on Build GRCh38
rs765617011 Chr.6: 21598194-21598196 on Build GRCh38
rs184779509 Chr.6: 21597933-21597933 on Build GRCh38
rs1011329842 Chr.6: 21597864-21597864 on Build GRCh38
rs889119753 Chr.6: 21598049-21598049 on Build GRCh38
rs144734333 Chr.6: 21598068-21598068 on Build GRCh38
rs932619593 Chr.6: 21597768-21597768 on Build GRCh38
rs747212000 Chr.6: 21598212-21598212 on Build GRCh38
rs1001569510 Chr.6: 21597901-21597901 on Build GRCh38
rs961474714 Chr.6: 21597989-21597989 on Build GRCh38
rs749882049 Chr.6: 21597772-21597772 on Build GRCh38
rs868347616 Chr.6: 21597957-21597957 on Build GRCh38
rs905537503 Chr.6: 21597878-21597878 on Build GRCh38
rs755665020 Chr.6: 21597784-21597784 on Build GRCh38
rs555267051 Chr.6: 21597763-21597763 on Build GRCh38
rs544308760 Chr.6: 21597940-21597940 on Build GRCh38
rs200289141 Chr.6: 21597850-21597850 on Build GRCh38
rs974136530 Chr.6: 21598051-21598051 on Build GRCh38
rs944270672 Chr.6: 21598158-21598158 on Build GRCh38
rs776616562 Chr.6: 21598023-21598023 on Build GRCh38
rs367694993 Chr.6: 21598130-21598130 on Build GRCh38
rs1035825909 Chr.6: 21598089-21598089 on Build GRCh38
rs868020907 Chr.6: 21597781-21597781 on Build GRCh38
rs759689295 Chr.6: 21598076-21598076 on Build GRCh38
rs540233966 Chr.6: 21597787-21597787 on Build GRCh38
rs1023362818 Chr.6: 21598165-21598168 on Build GRCh38
rs1042831160 Chr.6: 21598218-21598218 on Build GRCh38
rs779963265 Chr.6: 21597871-21597871 on Build GRCh38
rs944232069 Chr.6: 21597813-21597813 on Build GRCh38
rs3192372 Chr.6: 21597994-21597994 on Build GRCh38
rs1035630370 Chr.6: 21597925-21597925 on Build GRCh38
rs772026753 Chr.6: 21597981-21597981 on Build GRCh38
rs913397592 Chr.6: 21598208-21598208 on Build GRCh38
rs564867245 Chr.6: 21597820-21597820 on Build GRCh38
rs529920193 Chr.6: 21598060-21598060 on Build GRCh38
rs527546877 Chr.6: 21598157-21598157 on Build GRCh38
rs566769836 Chr.6: 21598153-21598153 on Build GRCh38
rs905588228 Chr.6: 21598221-21598221 on Build GRCh38
rs546828315 Chr.6: 21597907-21597908 on Build GRCh38
rs180923473 Chr.6: 21597786-21597786 on Build GRCh38
rs969010916 Chr.6: 21598168-21598168 on Build GRCh38
rs969063549 Chr.6: 21597851-21597851 on Build GRCh38
rs895982070 Chr.6: 21598132-21598132 on Build GRCh38
rs960466258 Chr.6: 21597807-21597807 on Build GRCh38

View Full Product Details