Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28129352-28129852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN16
Entrez Gene ID
80345
Gene Name
zinc finger and SCAN domain containing 16
Gene Aliases
ZNF392, ZNF435, dJ265C24.3
UniGene
Hs.656641
Gene Chromosome Location
Chr.6: 28123753-28130086 on Build GRCh38
Transcripts
NM_001320555.1 NM_001320556.1 NM_001320557.1 NM_025231.2 XM_017011324.1 NR_103455.1 NR_103456.1

Gene Symbol
ZSCAN16-AS1
Entrez Gene ID
100129195
Gene Name
ZSCAN16 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659413
Gene Chromosome Location
Chr.6: 28121795-28137316 on Build GRCh38
Transcripts
NM_001320555.1 NM_001320556.1 NM_001320557.1 NM_025231.2 XM_017011324.1 NR_103455.1 NR_103456.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754716 c.841G>T p.G281* Chr.6: 28129744-28129744 on Build GRCh38
3861143 c.736A>G p.S246G Chr.6: 28129639-28129639 on Build GRCh38
1754717 c.842G>T p.G281V Chr.6: 28129745-28129745 on Build GRCh38
5869947 c.933G>C p.Q311H Chr.6: 28129836-28129836 on Build GRCh38
324589 c.679G>C p.E227Q Chr.6: 28129582-28129582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778816441 Chr.6: 28129684-28129684 on Build GRCh38
rs538390841 Chr.6: 28129697-28129697 on Build GRCh38
rs756394908 Chr.6: 28129670-28129670 on Build GRCh38
rs766113558 Chr.6: 28129760-28129760 on Build GRCh38
rs568217342 Chr.6: 28129585-28129585 on Build GRCh38
rs751395128 Chr.6: 28129446-28129446 on Build GRCh38
rs749020483 Chr.6: 28129790-28129790 on Build GRCh38
rs201154635 Chr.6: 28129686-28129686 on Build GRCh38
rs780811286 Chr.6: 28129470-28129470 on Build GRCh38
rs753548538 Chr.6: 28129400-28129400 on Build GRCh38
rs749450546 Chr.6: 28129679-28129679 on Build GRCh38
rs768177372 Chr.6: 28129802-28129802 on Build GRCh38
rs370893996 Chr.6: 28129475-28129475 on Build GRCh38
rs376016517 Chr.6: 28129825-28129825 on Build GRCh38
rs200359435 Chr.6: 28129780-28129780 on Build GRCh38
rs781087677 Chr.6: 28129628-28129628 on Build GRCh38
rs755916689 Chr.6: 28129752-28129752 on Build GRCh38
rs766200223 Chr.6: 28129824-28129824 on Build GRCh38
rs772887510 Chr.6: 28129718-28129721 on Build GRCh38
rs775305482 Chr.6: 28129731-28129731 on Build GRCh38
rs1051344606 Chr.6: 28129586-28129586 on Build GRCh38
rs371909530 Chr.6: 28129625-28129625 on Build GRCh38
rs777107874 Chr.6: 28129786-28129786 on Build GRCh38
rs771967409 Chr.6: 28129705-28129706 on Build GRCh38
rs769770985 Chr.6: 28129615-28129615 on Build GRCh38
rs151021669 Chr.6: 28129766-28129766 on Build GRCh38
rs200631257 Chr.6: 28129606-28129606 on Build GRCh38
rs367727118 Chr.6: 28129459-28129459 on Build GRCh38
rs762566422 Chr.6: 28129607-28129607 on Build GRCh38
rs773709352 Chr.6: 28129733-28129733 on Build GRCh38
rs4713140 Chr.6: 28129415-28129415 on Build GRCh38
rs767712705 Chr.6: 28129398-28129398 on Build GRCh38
rs199689547 Chr.6: 28129685-28129685 on Build GRCh38
rs201405736 Chr.6: 28129471-28129471 on Build GRCh38
rs142666616 Chr.6: 28129764-28129764 on Build GRCh38
rs777771536 Chr.6: 28129427-28129427 on Build GRCh38
rs532689014 Chr.6: 28129522-28129522 on Build GRCh38
rs768241579 Chr.6: 28129443-28129444 on Build GRCh38
rs935986660 Chr.6: 28129813-28129813 on Build GRCh38
rs113926481 Chr.6: 28129665-28129665 on Build GRCh38
rs139845128 Chr.6: 28129536-28129536 on Build GRCh38
rs374784537 Chr.6: 28129775-28129775 on Build GRCh38
rs368299349 Chr.6: 28129542-28129542 on Build GRCh38
rs921380597 Chr.6: 28129777-28129777 on Build GRCh38
rs998753094 Chr.6: 28129478-28129478 on Build GRCh38
rs750204745 Chr.6: 28129632-28129632 on Build GRCh38
rs370814227 Chr.6: 28129404-28129404 on Build GRCh38
rs763943760 Chr.6: 28129702-28129702 on Build GRCh38
rs774188742 Chr.6: 28129452-28129459 on Build GRCh38
rs148832646 Chr.6: 28129734-28129734 on Build GRCh38
rs767350945 Chr.6: 28129738-28129738 on Build GRCh38
rs750290493 Chr.6: 28129743-28129743 on Build GRCh38
rs568777140 Chr.6: 28129729-28129729 on Build GRCh38
rs200987807 Chr.6: 28129609-28129609 on Build GRCh38
rs189210919 Chr.6: 28129732-28129732 on Build GRCh38
rs759323511 Chr.6: 28129598-28129598 on Build GRCh38
rs994567434 Chr.6: 28129695-28129695 on Build GRCh38
rs766950970 Chr.6: 28129814-28129814 on Build GRCh38
rs773975435 Chr.6: 28129808-28129808 on Build GRCh38
rs748829860 Chr.6: 28129441-28129442 on Build GRCh38
rs758480529 Chr.6: 28129416-28129416 on Build GRCh38
rs527579792 Chr.6: 28129381-28129381 on Build GRCh38
rs35741057 Chr.6: 28129763-28129764 on Build GRCh38
rs940228904 Chr.6: 28129819-28129819 on Build GRCh38
rs779109473 Chr.6: 28129763-28129763 on Build GRCh38
rs778437676 Chr.6: 28129767-28129767 on Build GRCh38
rs202104399 Chr.6: 28129524-28129524 on Build GRCh38
rs145967433 Chr.6: 28129517-28129517 on Build GRCh38
rs1053761058 Chr.6: 28129429-28129429 on Build GRCh38
rs773782451 Chr.6: 28129718-28129718 on Build GRCh38
rs762668820 Chr.6: 28129680-28129680 on Build GRCh38
rs372143433 Chr.6: 28129761-28129761 on Build GRCh38
rs780260042 Chr.6: 28129673-28129673 on Build GRCh38
rs773429225 Chr.6: 28129562-28129562 on Build GRCh38
rs143274731 Chr.6: 28129699-28129699 on Build GRCh38
rs767467148 Chr.6: 28129817-28129817 on Build GRCh38
rs771364627 Chr.6: 28129730-28129730 on Build GRCh38
rs200539838 Chr.6: 28129630-28129630 on Build GRCh38
rs764981629 Chr.6: 28129600-28129600 on Build GRCh38
rs1383394 Chr.6: 28129717-28129717 on Build GRCh38
rs761536265 Chr.6: 28129627-28129628 on Build GRCh38

View Full Product Details