Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28148225-28148722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZKSCAN8
Entrez Gene ID
7745
Gene Name
zinc finger with KRAB and SCAN domains 8
Gene Aliases
LD5-1, ZNF192, ZSCAN40
UniGene
Hs.57679
Gene Chromosome Location
Chr.6: 28141644-28159472 on Build GRCh38
Transcripts
NM_001278119.1 NM_001278121.1 NM_001278122.1 NM_006298.3 XM_011514870.2 XM_011514873.2 XM_011514874.2 XM_017011266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994784 c.7G>A p.E3K Chr.6: 28148414-28148414 on Build GRCh38
3725213 c.145G>C p.V49L Chr.6: 28148552-28148552 on Build GRCh38
4353624 c.151C>T p.R51C Chr.6: 28148558-28148558 on Build GRCh38
4353628 c.252G>T p.L84F Chr.6: 28148659-28148659 on Build GRCh38
4751735 c.298T>G p.L100V Chr.6: 28148705-28148705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003762006 Chr.6: 28148448-28148448 on Build GRCh38
rs146729116 Chr.6: 28148479-28148479 on Build GRCh38
rs752977146 Chr.6: 28148550-28148550 on Build GRCh38
rs751761827 Chr.6: 28148358-28148358 on Build GRCh38
rs201156339 Chr.6: 28148673-28148673 on Build GRCh38
rs377289231 Chr.6: 28148403-28148403 on Build GRCh38
rs1003540435 Chr.6: 28148630-28148630 on Build GRCh38
rs745382215 Chr.6: 28148483-28148483 on Build GRCh38
rs373356722 Chr.6: 28148505-28148505 on Build GRCh38
rs112053323 Chr.6: 28148332-28148332 on Build GRCh38
rs766323123 Chr.6: 28148538-28148538 on Build GRCh38
rs755360556 Chr.6: 28148476-28148476 on Build GRCh38
rs756612582 Chr.6: 28148381-28148381 on Build GRCh38
rs767207132 Chr.6: 28148565-28148565 on Build GRCh38
rs374416230 Chr.6: 28148376-28148376 on Build GRCh38
rs749096573 Chr.6: 28148440-28148440 on Build GRCh38
rs914956249 Chr.6: 28148270-28148270 on Build GRCh38
rs900343526 Chr.6: 28148365-28148365 on Build GRCh38
rs138137752 Chr.6: 28148532-28148532 on Build GRCh38
rs765276486 Chr.6: 28148399-28148400 on Build GRCh38
rs772322982 Chr.6: 28148515-28148515 on Build GRCh38
rs748429265 Chr.6: 28148507-28148507 on Build GRCh38
rs760097552 Chr.6: 28148686-28148686 on Build GRCh38
rs376379490 Chr.6: 28148580-28148580 on Build GRCh38
rs754186096 Chr.6: 28148385-28148385 on Build GRCh38
rs1048913319 Chr.6: 28148691-28148691 on Build GRCh38
rs935522600 Chr.6: 28148310-28148310 on Build GRCh38
rs369163704 Chr.6: 28148501-28148501 on Build GRCh38
rs778909563 Chr.6: 28148602-28148602 on Build GRCh38
rs780627318 Chr.6: 28148480-28148480 on Build GRCh38
rs745845523 Chr.6: 28148429-28148429 on Build GRCh38
rs754942871 Chr.6: 28148597-28148597 on Build GRCh38
rs749982151 Chr.6: 28148576-28148576 on Build GRCh38
rs771696693 Chr.6: 28148627-28148627 on Build GRCh38
rs753765315 Chr.6: 28148364-28148367 on Build GRCh38
rs144390792 Chr.6: 28148564-28148564 on Build GRCh38
rs769759921 Chr.6: 28148430-28148430 on Build GRCh38
rs774732868 Chr.6: 28148451-28148451 on Build GRCh38
rs754955132 Chr.6: 28148257-28148257 on Build GRCh38
rs773230422 Chr.6: 28148526-28148526 on Build GRCh38
rs202039260 Chr.6: 28148604-28148604 on Build GRCh38
rs1040286585 Chr.6: 28148353-28148353 on Build GRCh38
rs139971461 Chr.6: 28148319-28148319 on Build GRCh38
rs750924319 Chr.6: 28148378-28148378 on Build GRCh38
rs562088137 Chr.6: 28148558-28148558 on Build GRCh38
rs369500730 Chr.6: 28148405-28148405 on Build GRCh38
rs755509173 Chr.6: 28148484-28148484 on Build GRCh38
rs778079466 Chr.6: 28148655-28148655 on Build GRCh38
rs149570086 Chr.6: 28148563-28148563 on Build GRCh38
rs80184076 Chr.6: 28148517-28148517 on Build GRCh38
rs988300602 Chr.6: 28148337-28148337 on Build GRCh38
rs768510722 Chr.6: 28148445-28148445 on Build GRCh38
rs755270774 Chr.6: 28148399-28148399 on Build GRCh38
rs1040979899 Chr.6: 28148497-28148497 on Build GRCh38
rs376159677 Chr.6: 28148545-28148545 on Build GRCh38
rs931885400 Chr.6: 28148593-28148593 on Build GRCh38
rs141619075 Chr.6: 28148477-28148477 on Build GRCh38
rs778635581 Chr.6: 28148318-28148318 on Build GRCh38
rs777870600 Chr.6: 28148681-28148681 on Build GRCh38
rs1035200430 Chr.6: 28148471-28148471 on Build GRCh38
rs773319293 Chr.6: 28148459-28148459 on Build GRCh38
rs767534634 Chr.6: 28148456-28148456 on Build GRCh38
rs967848988 Chr.6: 28148498-28148498 on Build GRCh38
rs145136963 Chr.6: 28148296-28148296 on Build GRCh38
rs749183992 Chr.6: 28148499-28148499 on Build GRCh38
rs759650086 Chr.6: 28148397-28148399 on Build GRCh38
rs200124524 Chr.6: 28148382-28148382 on Build GRCh38
rs147066723 Chr.6: 28148502-28148502 on Build GRCh38
rs377359410 Chr.6: 28148470-28148470 on Build GRCh38
rs761994134 Chr.6: 28148362-28148362 on Build GRCh38
rs755669536 Chr.6: 28148579-28148579 on Build GRCh38
rs201866786 Chr.6: 28148487-28148487 on Build GRCh38
rs565972276 Chr.6: 28148299-28148299 on Build GRCh38
rs754288086 Chr.6: 28148475-28148475 on Build GRCh38
rs762083747 Chr.6: 28148455-28148455 on Build GRCh38
rs765643034 Chr.6: 28148559-28148559 on Build GRCh38
rs766902901 Chr.6: 28148472-28148472 on Build GRCh38
rs780725960 Chr.6: 28148412-28148412 on Build GRCh38
rs990984788 Chr.6: 28148254-28148254 on Build GRCh38
rs944625078 Chr.6: 28148330-28148330 on Build GRCh38
rs756145802 Chr.6: 28148624-28148624 on Build GRCh38
rs747161257 Chr.6: 28148660-28148660 on Build GRCh38
rs143435480 Chr.6: 28148437-28148437 on Build GRCh38

View Full Product Details