Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38702814-38703318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLO1
Entrez Gene ID
2739
Gene Name
glyoxalase I
Gene Aliases
GLOD1, GLYI, HEL-S-74
UniGene
Hs.268849
Gene Chromosome Location
Chr.6: 38675925-38703176 on Build GRCh38
Transcripts
NM_006708.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586714 c.42G>T p.E14D Chr.6: 38703013-38703013 on Build GRCh38
3777630 c.51C>T p.L17L Chr.6: 38703004-38703004 on Build GRCh38
5701149 c.29_30insG p.L11fs*33 Chr.6: 38703025-38703026 on Build GRCh38
70911 c.59G>A p.C20Y Chr.6: 38702996-38702996 on Build GRCh38
116085 c.28G>T p.G10C Chr.6: 38703027-38703027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011629959 Chr.6: 38702937-38702937 on Build GRCh38
rs953632187 Chr.6: 38703083-38703083 on Build GRCh38
rs1024121517 Chr.6: 38703019-38703019 on Build GRCh38
rs932149895 Chr.6: 38702886-38702886 on Build GRCh38
rs998652747 Chr.6: 38703256-38703256 on Build GRCh38
rs771854203 Chr.6: 38702948-38702948 on Build GRCh38
rs75521110 Chr.6: 38703096-38703096 on Build GRCh38
rs1049346 Chr.6: 38703061-38703061 on Build GRCh38
rs771349920 Chr.6: 38703015-38703023 on Build GRCh38
rs533809608 Chr.6: 38702940-38702940 on Build GRCh38
rs776172656 Chr.6: 38702972-38702972 on Build GRCh38
rs1019925576 Chr.6: 38702945-38702945 on Build GRCh38
rs1052532079 Chr.6: 38703260-38703260 on Build GRCh38
rs911649720 Chr.6: 38702898-38702898 on Build GRCh38
rs527469839 Chr.6: 38703169-38703169 on Build GRCh38
rs915460454 Chr.6: 38702865-38702865 on Build GRCh38
rs553667476 Chr.6: 38702956-38702956 on Build GRCh38
rs944455329 Chr.6: 38702914-38702914 on Build GRCh38
rs561117954 Chr.6: 38703081-38703081 on Build GRCh38
rs370207834 Chr.6: 38703097-38703097 on Build GRCh38
rs9470920 Chr.6: 38702957-38702957 on Build GRCh38
rs953060079 Chr.6: 38703170-38703170 on Build GRCh38
rs541708855 Chr.6: 38703129-38703129 on Build GRCh38
rs147217818 Chr.6: 38702997-38702997 on Build GRCh38
rs78634088 Chr.6: 38703151-38703151 on Build GRCh38
rs564803716 Chr.6: 38702980-38702980 on Build GRCh38
rs558835278 Chr.6: 38703093-38703093 on Build GRCh38
rs377238075 Chr.6: 38702978-38702978 on Build GRCh38
rs112098082 Chr.6: 38702925-38702925 on Build GRCh38
rs1035810673 Chr.6: 38702977-38702977 on Build GRCh38
rs749384509 Chr.6: 38703031-38703031 on Build GRCh38
rs780352687 Chr.6: 38702933-38702933 on Build GRCh38
rs779302090 Chr.6: 38703040-38703040 on Build GRCh38
rs545096833 Chr.6: 38703091-38703091 on Build GRCh38
rs750549657 Chr.6: 38702884-38702884 on Build GRCh38
rs772479734 Chr.6: 38703196-38703196 on Build GRCh38
rs547218642 Chr.6: 38703224-38703224 on Build GRCh38
rs17855424 Chr.6: 38702999-38702999 on Build GRCh38
rs775181202 Chr.6: 38703082-38703082 on Build GRCh38
rs755284158 Chr.6: 38703116-38703116 on Build GRCh38
rs530466619 Chr.6: 38703138-38703138 on Build GRCh38
rs200613640 Chr.6: 38703066-38703066 on Build GRCh38
rs1041351504 Chr.6: 38703124-38703124 on Build GRCh38
rs767814917 Chr.6: 38703015-38703015 on Build GRCh38
rs561546223 Chr.6: 38703137-38703137 on Build GRCh38
rs113061497 Chr.6: 38703027-38703027 on Build GRCh38
rs1002573696 Chr.6: 38703132-38703132 on Build GRCh38
rs907035184 Chr.6: 38703198-38703198 on Build GRCh38
rs1040192528 Chr.6: 38702942-38702942 on Build GRCh38
rs748008510 Chr.6: 38702943-38702943 on Build GRCh38
rs759075699 Chr.6: 38703078-38703078 on Build GRCh38
rs374581240 Chr.6: 38703077-38703077 on Build GRCh38
rs551249207 Chr.6: 38702922-38702922 on Build GRCh38
rs1050280504 Chr.6: 38703110-38703110 on Build GRCh38
rs771867285 Chr.6: 38703060-38703060 on Build GRCh38
rs1003622368 Chr.6: 38703038-38703038 on Build GRCh38
rs116880431 Chr.6: 38703161-38703161 on Build GRCh38
rs748365336 Chr.6: 38703043-38703043 on Build GRCh38
rs567219973 Chr.6: 38703268-38703268 on Build GRCh38
rs756641310 Chr.6: 38703021-38703021 on Build GRCh38
rs986287990 Chr.6: 38702891-38702891 on Build GRCh38
rs921315089 Chr.6: 38703254-38703254 on Build GRCh38
rs2277109 Chr.6: 38702932-38702932 on Build GRCh38
rs11544282 Chr.6: 38703094-38703094 on Build GRCh38
rs944124621 Chr.6: 38702853-38702853 on Build GRCh38
rs757622327 Chr.6: 38703013-38703013 on Build GRCh38
rs539166002 Chr.6: 38702869-38702869 on Build GRCh38
rs1015597614 Chr.6: 38703199-38703199 on Build GRCh38
rs755212608 Chr.6: 38703033-38703033 on Build GRCh38
rs965411354 Chr.6: 38703204-38703204 on Build GRCh38
rs543831399 Chr.6: 38703139-38703139 on Build GRCh38
rs918972495 Chr.6: 38703126-38703126 on Build GRCh38
rs770864573 Chr.6: 38703074-38703074 on Build GRCh38
rs532456642 Chr.6: 38703153-38703153 on Build GRCh38
rs751878828 Chr.6: 38703005-38703005 on Build GRCh38
rs967082134 Chr.6: 38702949-38702949 on Build GRCh38
rs781735028 Chr.6: 38702926-38702926 on Build GRCh38
rs377328531 Chr.6: 38702996-38702996 on Build GRCh38
rs749509100 Chr.6: 38702934-38702934 on Build GRCh38
rs989730630 Chr.6: 38702857-38702857 on Build GRCh38
rs140507823 Chr.6: 38703020-38703020 on Build GRCh38
rs892525829 Chr.6: 38703284-38703284 on Build GRCh38
rs947348021 Chr.6: 38703117-38703117 on Build GRCh38
rs552662733 Chr.6: 38703160-38703160 on Build GRCh38
rs759126965 Chr.6: 38702981-38702981 on Build GRCh38
rs779216513 Chr.6: 38703159-38703159 on Build GRCh38
rs746414991 Chr.6: 38702928-38702928 on Build GRCh38

View Full Product Details