Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39079027-39079522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLP1R
Entrez Gene ID
2740
Gene Name
glucagon like peptide 1 receptor
Gene Aliases
GLP-1, GLP-1-R, GLP-1R
UniGene
Hs.351883
Gene Chromosome Location
Chr.6: 39048781-39091303 on Build GRCh38
Transcripts
NM_002062.4 NR_136562.1 NR_136563.1 XM_017010750.1 XM_017010751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7416152 c.956T>A p.V319E Chr.6: 39079113-39079113 on Build GRCh38
1697171 c.1013A>T p.N338I Chr.6: 39079170-39079170 on Build GRCh38
3873825 c.981C>A p.V327V Chr.6: 39079138-39079138 on Build GRCh38
6174068 c.991G>C p.V331L Chr.6: 39079148-39079148 on Build GRCh38
3830439 c.957G>A p.V319V Chr.6: 39079114-39079114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201157675 Chr.6: 39079101-39079101 on Build GRCh38
rs755882647 Chr.6: 39079340-39079340 on Build GRCh38
rs200281379 Chr.6: 39079102-39079102 on Build GRCh38
rs755228090 Chr.6: 39079075-39079075 on Build GRCh38
rs200118342 Chr.6: 39079133-39079133 on Build GRCh38
rs373025051 Chr.6: 39079229-39079229 on Build GRCh38
rs10305493 Chr.6: 39079155-39079155 on Build GRCh38
rs750191459 Chr.6: 39079323-39079323 on Build GRCh38
rs928305009 Chr.6: 39079070-39079070 on Build GRCh38
rs1022978694 Chr.6: 39079259-39079260 on Build GRCh38
rs1004455407 Chr.6: 39079104-39079104 on Build GRCh38
rs889296051 Chr.6: 39079471-39079471 on Build GRCh38
rs752432456 Chr.6: 39079115-39079115 on Build GRCh38
rs199517476 Chr.6: 39079059-39079059 on Build GRCh38
rs375636725 Chr.6: 39079126-39079126 on Build GRCh38
rs202171972 Chr.6: 39079189-39079189 on Build GRCh38
rs200682417 Chr.6: 39079107-39079107 on Build GRCh38
rs201235247 Chr.6: 39079148-39079148 on Build GRCh38
rs200053296 Chr.6: 39079097-39079097 on Build GRCh38
rs5875654 Chr.6: 39079260-39079261 on Build GRCh38
rs186153401 Chr.6: 39079225-39079225 on Build GRCh38
rs748428406 Chr.6: 39079168-39079168 on Build GRCh38
rs761386 Chr.6: 39079095-39079095 on Build GRCh38
rs772450698 Chr.6: 39079099-39079099 on Build GRCh38
rs928515569 Chr.6: 39079324-39079324 on Build GRCh38
rs542888272 Chr.6: 39079446-39079446 on Build GRCh38
rs900903329 Chr.6: 39079449-39079449 on Build GRCh38
rs1019126528 Chr.6: 39079152-39079152 on Build GRCh38
rs755813574 Chr.6: 39079154-39079154 on Build GRCh38
rs745477141 Chr.6: 39079094-39079094 on Build GRCh38
rs202103886 Chr.6: 39079170-39079170 on Build GRCh38
rs1040628195 Chr.6: 39079497-39079497 on Build GRCh38
rs914208528 Chr.6: 39079351-39079351 on Build GRCh38
rs374281194 Chr.6: 39079114-39079114 on Build GRCh38
rs760873769 Chr.6: 39079060-39079060 on Build GRCh38
rs766554045 Chr.6: 39079065-39079065 on Build GRCh38
rs371656852 Chr.6: 39079052-39079052 on Build GRCh38
rs969857406 Chr.6: 39079280-39079280 on Build GRCh38
rs775831860 Chr.6: 39079228-39079228 on Build GRCh38
rs981372539 Chr.6: 39079313-39079313 on Build GRCh38
rs767619542 Chr.6: 39079238-39079238 on Build GRCh38
rs939839577 Chr.6: 39079432-39079432 on Build GRCh38
rs767639322 Chr.6: 39079380-39079380 on Build GRCh38
rs934287858 Chr.6: 39079077-39079077 on Build GRCh38
rs368600696 Chr.6: 39079057-39079058 on Build GRCh38
rs199774152 Chr.6: 39079108-39079108 on Build GRCh38
rs1049466623 Chr.6: 39079457-39079457 on Build GRCh38
rs571893057 Chr.6: 39079078-39079078 on Build GRCh38
rs749947512 Chr.6: 39079244-39079244 on Build GRCh38
rs1045639934 Chr.6: 39079053-39079053 on Build GRCh38
rs758257648 Chr.6: 39079082-39079082 on Build GRCh38
rs200890494 Chr.6: 39079178-39079178 on Build GRCh38
rs772533032 Chr.6: 39079171-39079171 on Build GRCh38
rs147207725 Chr.6: 39079145-39079145 on Build GRCh38
rs909807014 Chr.6: 39079408-39079408 on Build GRCh38
rs528866272 Chr.6: 39079445-39079445 on Build GRCh38
rs752553316 Chr.6: 39079080-39079080 on Build GRCh38
rs978685338 Chr.6: 39079282-39079282 on Build GRCh38
rs984860594 Chr.6: 39079214-39079214 on Build GRCh38
rs747487240 Chr.6: 39079181-39079181 on Build GRCh38
rs777575741 Chr.6: 39079144-39079144 on Build GRCh38
rs780871870 Chr.6: 39079088-39079088 on Build GRCh38
rs202144978 Chr.6: 39079096-39079096 on Build GRCh38
rs755723418 Chr.6: 39079252-39079252 on Build GRCh38
rs560207479 Chr.6: 39079416-39079416 on Build GRCh38
rs990762891 Chr.6: 39079339-39079339 on Build GRCh38
rs148030795 Chr.6: 39079232-39079232 on Build GRCh38
rs754104238 Chr.6: 39079072-39079072 on Build GRCh38
rs777835186 Chr.6: 39079083-39079083 on Build GRCh38
rs147399881 Chr.6: 39079195-39079195 on Build GRCh38
rs767393758 Chr.6: 39079206-39079206 on Build GRCh38
rs746472258 Chr.6: 39079085-39079085 on Build GRCh38
rs35958702 Chr.6: 39079080-39079081 on Build GRCh38
rs753173778 Chr.6: 39079261-39079262 on Build GRCh38
rs369825346 Chr.6: 39079205-39079205 on Build GRCh38
rs766392467 Chr.6: 39079105-39079105 on Build GRCh38
rs947126009 Chr.6: 39079353-39079353 on Build GRCh38
rs199959395 Chr.6: 39079172-39079172 on Build GRCh38
rs943527724 Chr.6: 39079492-39079492 on Build GRCh38
rs1036893396 Chr.6: 39079435-39079435 on Build GRCh38
rs916614188 Chr.6: 39079342-39079342 on Build GRCh38
rs140519519 Chr.6: 39079147-39079147 on Build GRCh38
rs375971856 Chr.6: 39079230-39079230 on Build GRCh38
rs540508046 Chr.6: 39079348-39079348 on Build GRCh38

View Full Product Details