Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152351557-152352062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6336678 c.5873A>C p.Y1958S Chr.1: 152351913-152351913 on Build GRCh38
6239352 c.6060C>A p.S2020S Chr.1: 152351726-152351726 on Build GRCh38
275185 c.6107G>A p.S2036N Chr.1: 152351679-152351679 on Build GRCh38
7421911 c.6093T>A p.S2031R Chr.1: 152351693-152351693 on Build GRCh38
4553634 c.5976G>A p.E1992E Chr.1: 152351810-152351810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917704724 Chr.1: 152351765-152351765 on Build GRCh38
rs763868998 Chr.1: 152351797-152351797 on Build GRCh38
rs200728576 Chr.1: 152351781-152351781 on Build GRCh38
rs368870718 Chr.1: 152351747-152351747 on Build GRCh38
rs759741821 Chr.1: 152351897-152351897 on Build GRCh38
rs769587483 Chr.1: 152351965-152351965 on Build GRCh38
rs777733623 Chr.1: 152351695-152351695 on Build GRCh38
rs762787998 Chr.1: 152351954-152351954 on Build GRCh38
rs767790660 Chr.1: 152351973-152351987 on Build GRCh38
rs766645216 Chr.1: 152351787-152351787 on Build GRCh38
rs76395912 Chr.1: 152351906-152351906 on Build GRCh38
rs573208164 Chr.1: 152351635-152351635 on Build GRCh38
rs148541975 Chr.1: 152351857-152351857 on Build GRCh38
rs755816906 Chr.1: 152351688-152351688 on Build GRCh38
rs760715355 Chr.1: 152352024-152352025 on Build GRCh38
rs866710773 Chr.1: 152352037-152352037 on Build GRCh38
rs150834303 Chr.1: 152352036-152352036 on Build GRCh38
rs371924445 Chr.1: 152351822-152351822 on Build GRCh38
rs751285860 Chr.1: 152351888-152351888 on Build GRCh38
rs386635462 Chr.1: 152351839-152351840 on Build GRCh38
rs756823863 Chr.1: 152351704-152351704 on Build GRCh38
rs148224294 Chr.1: 152351803-152351803 on Build GRCh38
rs537172304 Chr.1: 152351579-152351579 on Build GRCh38
rs769689324 Chr.1: 152352031-152352031 on Build GRCh38
rs183469872 Chr.1: 152351988-152351988 on Build GRCh38
rs761649223 Chr.1: 152351926-152351926 on Build GRCh38
rs386635461 Chr.1: 152351835-152351836 on Build GRCh38
rs570972849 Chr.1: 152351786-152351786 on Build GRCh38
rs773074983 Chr.1: 152351834-152351835 on Build GRCh38
rs1858484 Chr.1: 152351810-152351810 on Build GRCh38
rs770564319 Chr.1: 152351879-152351880 on Build GRCh38
rs749750186 Chr.1: 152351902-152351902 on Build GRCh38
rs202060997 Chr.1: 152352032-152352032 on Build GRCh38
rs749826103 Chr.1: 152351586-152351586 on Build GRCh38
rs780299093 Chr.1: 152352019-152352019 on Build GRCh38
rs767327149 Chr.1: 152351983-152351983 on Build GRCh38
rs199701425 Chr.1: 152351719-152351719 on Build GRCh38
rs140669858 Chr.1: 152351895-152351895 on Build GRCh38
rs201561111 Chr.1: 152351706-152351706 on Build GRCh38
rs140302446 Chr.1: 152351763-152351763 on Build GRCh38
rs267598032 Chr.1: 152351933-152351933 on Build GRCh38
rs745312389 Chr.1: 152351712-152351712 on Build GRCh38
rs776417956 Chr.1: 152351854-152351854 on Build GRCh38
rs200489815 Chr.1: 152351804-152351804 on Build GRCh38
rs949284289 Chr.1: 152352024-152352024 on Build GRCh38
rs757707589 Chr.1: 152351993-152351993 on Build GRCh38
rs557443846 Chr.1: 152351840-152351840 on Build GRCh38
rs144875878 Chr.1: 152351872-152351872 on Build GRCh38
rs750652755 Chr.1: 152351597-152351597 on Build GRCh38
rs528776790 Chr.1: 152351733-152351733 on Build GRCh38
rs764453436 Chr.1: 152351585-152351585 on Build GRCh38
rs146074268 Chr.1: 152351639-152351639 on Build GRCh38
rs753741180 Chr.1: 152351698-152351698 on Build GRCh38
rs773268594 Chr.1: 152351949-152351949 on Build GRCh38
rs564244714 Chr.1: 152351669-152351669 on Build GRCh38
rs778383954 Chr.1: 152351900-152351900 on Build GRCh38
rs894704015 Chr.1: 152351982-152351982 on Build GRCh38
rs537890764 Chr.1: 152351839-152351839 on Build GRCh38
rs543978608 Chr.1: 152351663-152351663 on Build GRCh38
rs369780428 Chr.1: 152351812-152351812 on Build GRCh38
rs994130839 Chr.1: 152351899-152351899 on Build GRCh38
rs765245277 Chr.1: 152351863-152351863 on Build GRCh38
rs866170487 Chr.1: 152351964-152351964 on Build GRCh38
rs200946758 Chr.1: 152351956-152351956 on Build GRCh38
rs144916315 Chr.1: 152351792-152351792 on Build GRCh38
rs917834877 Chr.1: 152352009-152352009 on Build GRCh38
rs765378483 Chr.1: 152351673-152351673 on Build GRCh38
rs775512775 Chr.1: 152351999-152351999 on Build GRCh38
rs561751342 Chr.1: 152351762-152351762 on Build GRCh38
rs750743005 Chr.1: 152351681-152351681 on Build GRCh38
rs769322133 Chr.1: 152351667-152351667 on Build GRCh38
rs775096165 Chr.1: 152351981-152351981 on Build GRCh38
rs751143934 Chr.1: 152351975-152351975 on Build GRCh38
rs551007552 Chr.1: 152351595-152351595 on Build GRCh38
rs747227789 Chr.1: 152352020-152352020 on Build GRCh38
rs774144981 Chr.1: 152351891-152351891 on Build GRCh38
rs745972718 Chr.1: 152352002-152352002 on Build GRCh38
rs372992265 Chr.1: 152351770-152351770 on Build GRCh38
rs753976340 Chr.1: 152351588-152351588 on Build GRCh38
rs762900875 Chr.1: 152351766-152351766 on Build GRCh38
rs769741390 Chr.1: 152351714-152351721 on Build GRCh38
rs149671990 Chr.1: 152351898-152351898 on Build GRCh38
rs77345168 Chr.1: 152351623-152351623 on Build GRCh38
rs986580428 Chr.1: 152351672-152351672 on Build GRCh38
rs141276572 Chr.1: 152351670-152351670 on Build GRCh38
rs148579744 Chr.1: 152351836-152351836 on Build GRCh38
rs148270234 Chr.1: 152351638-152351638 on Build GRCh38
rs200175088 Chr.1: 152351622-152351622 on Build GRCh38
rs376358298 Chr.1: 152351590-152351590 on Build GRCh38
rs779438496 Chr.1: 152352001-152352001 on Build GRCh38
rs778573179 Chr.1: 152351640-152351640 on Build GRCh38
rs772315553 Chr.1: 152351919-152351919 on Build GRCh38
rs144929950 Chr.1: 152351633-152351633 on Build GRCh38
rs79983774 Chr.1: 152351921-152351921 on Build GRCh38
rs754771681 Chr.1: 152351601-152351601 on Build GRCh38
rs980613690 Chr.1: 152352028-152352028 on Build GRCh38
rs199899373 Chr.1: 152351835-152351835 on Build GRCh38
rs752366274 Chr.1: 152351984-152351984 on Build GRCh38
rs755402294 Chr.1: 152351817-152351817 on Build GRCh38
rs535751082 Chr.1: 152351824-152351824 on Build GRCh38
rs887305255 Chr.1: 152352035-152352035 on Build GRCh38
rs772121360 Chr.1: 152352004-152352004 on Build GRCh38
rs148279391 Chr.1: 152351986-152351986 on Build GRCh38
rs752567643 Chr.1: 152351686-152351686 on Build GRCh38
rs761559123 Chr.1: 152351858-152351858 on Build GRCh38
rs749984830 Chr.1: 152351808-152351808 on Build GRCh38
rs745883798 Chr.1: 152351913-152351913 on Build GRCh38
rs539533778 Chr.1: 152351799-152351799 on Build GRCh38
rs987021120 Chr.1: 152351749-152351749 on Build GRCh38
rs376795828 Chr.1: 152351881-152351881 on Build GRCh38
rs575262399 Chr.1: 152351662-152351662 on Build GRCh38
rs372918296 Chr.1: 152351583-152351583 on Build GRCh38
rs140460426 Chr.1: 152351630-152351630 on Build GRCh38
rs138683630 Chr.1: 152351849-152351849 on Build GRCh38
rs1017682353 Chr.1: 152351943-152351943 on Build GRCh38
rs774306057 Chr.1: 152351908-152351908 on Build GRCh38
rs372778870 Chr.1: 152351904-152351904 on Build GRCh38
rs559508459 Chr.1: 152351726-152351726 on Build GRCh38
rs772380958 Chr.1: 152351643-152351643 on Build GRCh38
rs898090743 Chr.1: 152351887-152351887 on Build GRCh38
rs781682216 Chr.1: 152351825-152351825 on Build GRCh38
rs201174892 Chr.1: 152351737-152351737 on Build GRCh38
rs749278143 Chr.1: 152351858-152351861 on Build GRCh38
rs766443752 Chr.1: 152351684-152351684 on Build GRCh38
rs769444603 Chr.1: 152351928-152351928 on Build GRCh38
rs559545444 Chr.1: 152351879-152351879 on Build GRCh38
rs574531606 Chr.1: 152351992-152351992 on Build GRCh38
rs748455525 Chr.1: 152351828-152351828 on Build GRCh38
rs777318493 Chr.1: 152351759-152351759 on Build GRCh38

View Full Product Details