Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153983138-153983636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB13
Entrez Gene ID
5872
Gene Name
RAB13, member RAS oncogene family
Gene Aliases
GIG4
UniGene
Hs.151536
Gene Chromosome Location
Chr.1: 153981617-153990644 on Build GRCh38
Transcripts
NM_001272038.1 NM_002870.3 NR_073553.1 XM_017001959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6236999 c.272C>A p.T91K Chr.1: 153983271-153983271 on Build GRCh38
896484 c.264C>A p.Y88* Chr.1: 153983279-153983279 on Build GRCh38
4022712 c.319A>G p.K107E Chr.1: 153983224-153983224 on Build GRCh38
5509717 c.191C>T p.T64M Chr.1: 153983576-153983576 on Build GRCh38
5652582 c.186-1G>A p.? Chr.1: 153983582-153983582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026653941 Chr.1: 153983393-153983393 on Build GRCh38
rs762474093 Chr.1: 153983313-153983313 on Build GRCh38
rs985567775 Chr.1: 153983455-153983455 on Build GRCh38
rs1053880370 Chr.1: 153983567-153983567 on Build GRCh38
rs770138791 Chr.1: 153983219-153983219 on Build GRCh38
rs997690737 Chr.1: 153983442-153983442 on Build GRCh38
rs559940372 Chr.1: 153983173-153983173 on Build GRCh38
rs372931300 Chr.1: 153983366-153983366 on Build GRCh38
rs750132012 Chr.1: 153983303-153983303 on Build GRCh38
rs778220242 Chr.1: 153983562-153983562 on Build GRCh38
rs776709028 Chr.1: 153983270-153983270 on Build GRCh38
rs765938779 Chr.1: 153983320-153983320 on Build GRCh38
rs769662617 Chr.1: 153983508-153983508 on Build GRCh38
rs779339153 Chr.1: 153983603-153983603 on Build GRCh38
rs902331564 Chr.1: 153983214-153983214 on Build GRCh38
rs754086932 Chr.1: 153983583-153983583 on Build GRCh38
rs781712765 Chr.1: 153983332-153983332 on Build GRCh38
rs747583449 Chr.1: 153983175-153983175 on Build GRCh38
rs746014186 Chr.1: 153983486-153983486 on Build GRCh38
rs777965672 Chr.1: 153983348-153983348 on Build GRCh38
rs751010727 Chr.1: 153983326-153983326 on Build GRCh38
rs781547668 Chr.1: 153983192-153983192 on Build GRCh38
rs773537869 Chr.1: 153983243-153983243 on Build GRCh38
rs749553096 Chr.1: 153983466-153983466 on Build GRCh38
rs765879703 Chr.1: 153983522-153983522 on Build GRCh38
rs748444413 Chr.1: 153983206-153983206 on Build GRCh38
rs757640668 Chr.1: 153983590-153983590 on Build GRCh38
rs753189443 Chr.1: 153983335-153983335 on Build GRCh38
rs531234487 Chr.1: 153983453-153983453 on Build GRCh38
rs774252826 Chr.1: 153983255-153983255 on Build GRCh38
rs757976221 Chr.1: 153983169-153983169 on Build GRCh38
rs761811925 Chr.1: 153983498-153983498 on Build GRCh38
rs868247308 Chr.1: 153983278-153983278 on Build GRCh38
rs756547121 Chr.1: 153983346-153983346 on Build GRCh38
rs952454382 Chr.1: 153983437-153983437 on Build GRCh38
rs771007895 Chr.1: 153983477-153983477 on Build GRCh38
rs866297319 Chr.1: 153983194-153983194 on Build GRCh38
rs751207948 Chr.1: 153983531-153983531 on Build GRCh38
rs761761199 Chr.1: 153983279-153983279 on Build GRCh38
rs368998921 Chr.1: 153983287-153983287 on Build GRCh38
rs773980856 Chr.1: 153983448-153983448 on Build GRCh38
rs186620971 Chr.1: 153983493-153983493 on Build GRCh38
rs376027047 Chr.1: 153983575-153983575 on Build GRCh38
rs967850994 Chr.1: 153983576-153983576 on Build GRCh38
rs144395961 Chr.1: 153983550-153983550 on Build GRCh38
rs377318766 Chr.1: 153983269-153983269 on Build GRCh38
rs772268363 Chr.1: 153983492-153983492 on Build GRCh38
rs563957950 Chr.1: 153983417-153983417 on Build GRCh38
rs1015538450 Chr.1: 153983305-153983305 on Build GRCh38
rs1035655806 Chr.1: 153983613-153983613 on Build GRCh38
rs755479218 Chr.1: 153983327-153983327 on Build GRCh38
rs759438703 Chr.1: 153983445-153983445 on Build GRCh38
rs753238807 Chr.1: 153983551-153983551 on Build GRCh38
rs144423974 Chr.1: 153983254-153983254 on Build GRCh38
rs756535536 Chr.1: 153983554-153983554 on Build GRCh38
rs549744601 Chr.1: 153983482-153983482 on Build GRCh38
rs760704492 Chr.1: 153983258-153983258 on Build GRCh38
rs552070663 Chr.1: 153983370-153983370 on Build GRCh38
rs977469670 Chr.1: 153983577-153983577 on Build GRCh38
rs762679129 Chr.1: 153983511-153983511 on Build GRCh38
rs779137345 Chr.1: 153983468-153983468 on Build GRCh38
rs745899854 Chr.1: 153983589-153983591 on Build GRCh38
rs755428042 Chr.1: 153983177-153983177 on Build GRCh38
rs759104422 Chr.1: 153983532-153983532 on Build GRCh38
rs939726459 Chr.1: 153983325-153983325 on Build GRCh38

View Full Product Details