Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43520533-43521027 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1C
Entrez Gene ID
9533
Gene Name
RNA polymerase I subunit C
Gene Aliases
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3
UniGene
Hs.743509
Gene Chromosome Location
Chr.6: 43517024-43562404 on Build GRCh38
Transcripts
NM_001318876.1 NM_203290.3 XM_011515000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4613692 c.751_753delGAG p.E252delE Chr.6: 43520720-43520722 on Build GRCh38
7081419 c.882G>C p.K294N Chr.6: 43521008-43521008 on Build GRCh38
4856097 c.695G>T p.S232I Chr.6: 43520664-43520664 on Build GRCh38
7181131 c.873G>C p.E291D Chr.6: 43520999-43520999 on Build GRCh38
3874366 c.768C>A p.C256* Chr.6: 43520737-43520737 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79826982 Chr.6: 43520657-43520657 on Build GRCh38
rs746932251 Chr.6: 43520710-43520710 on Build GRCh38
rs745835548 Chr.6: 43520935-43520935 on Build GRCh38
rs764460811 Chr.6: 43520926-43520926 on Build GRCh38
rs929224613 Chr.6: 43520880-43520880 on Build GRCh38
rs374207903 Chr.6: 43520638-43520638 on Build GRCh38
rs754188352 Chr.6: 43520688-43520688 on Build GRCh38
rs371479150 Chr.6: 43520971-43520971 on Build GRCh38
rs779412597 Chr.6: 43520776-43520776 on Build GRCh38
rs763593155 Chr.6: 43520668-43520668 on Build GRCh38
rs779858907 Chr.6: 43520954-43520954 on Build GRCh38
rs907156179 Chr.6: 43520839-43520839 on Build GRCh38
rs779322593 Chr.6: 43520699-43520699 on Build GRCh38
rs574803059 Chr.6: 43520777-43520777 on Build GRCh38
rs765709261 Chr.6: 43520901-43520901 on Build GRCh38
rs201455719 Chr.6: 43520785-43520785 on Build GRCh38
rs1012780248 Chr.6: 43520812-43520812 on Build GRCh38
rs375012958 Chr.6: 43520789-43520789 on Build GRCh38
rs375673819 Chr.6: 43520592-43520592 on Build GRCh38
rs746343275 Chr.6: 43520583-43520583 on Build GRCh38
rs543605130 Chr.6: 43520846-43520846 on Build GRCh38
rs374637200 Chr.6: 43520569-43520569 on Build GRCh38
rs747749279 Chr.6: 43520968-43520968 on Build GRCh38
rs191582628 Chr.6: 43520962-43520962 on Build GRCh38
rs200549821 Chr.6: 43520853-43520853 on Build GRCh38
rs760123164 Chr.6: 43520990-43520990 on Build GRCh38
rs758724481 Chr.6: 43520924-43520924 on Build GRCh38
rs764896977 Chr.6: 43520648-43520648 on Build GRCh38
rs368111931 Chr.6: 43520778-43520778 on Build GRCh38
rs371057107 Chr.6: 43520566-43520568 on Build GRCh38
rs910709453 Chr.6: 43520917-43520917 on Build GRCh38
rs953781624 Chr.6: 43520910-43520910 on Build GRCh38
rs147793518 Chr.6: 43520731-43520731 on Build GRCh38
rs773318501 Chr.6: 43520617-43520617 on Build GRCh38
rs770834639 Chr.6: 43520715-43520715 on Build GRCh38
rs759946050 Chr.6: 43520893-43520893 on Build GRCh38
rs762601060 Chr.6: 43520815-43520815 on Build GRCh38
rs781119795 Chr.6: 43520932-43520932 on Build GRCh38
rs371158834 Chr.6: 43520598-43520598 on Build GRCh38
rs1000346057 Chr.6: 43520630-43520630 on Build GRCh38
rs751006626 Chr.6: 43520754-43520754 on Build GRCh38
rs751702117 Chr.6: 43520929-43520929 on Build GRCh38
rs748345433 Chr.6: 43520701-43520701 on Build GRCh38
rs924649314 Chr.6: 43520952-43520952 on Build GRCh38
rs200121299 Chr.6: 43520756-43520756 on Build GRCh38
rs772343871 Chr.6: 43520702-43520702 on Build GRCh38
rs952189913 Chr.6: 43520747-43520747 on Build GRCh38
rs766143367 Chr.6: 43520634-43520634 on Build GRCh38
rs760685164 Chr.6: 43520631-43520631 on Build GRCh38
rs776119212 Chr.6: 43520992-43520992 on Build GRCh38
rs756505140 Chr.6: 43520585-43520585 on Build GRCh38
rs200780238 Chr.6: 43520817-43520817 on Build GRCh38
rs963134845 Chr.6: 43520826-43520826 on Build GRCh38
rs530969798 Chr.6: 43520808-43520808 on Build GRCh38
rs775455943 Chr.6: 43520917-43520920 on Build GRCh38
rs773752398 Chr.6: 43520816-43520816 on Build GRCh38
rs535176929 Chr.6: 43520949-43520949 on Build GRCh38
rs780289892 Chr.6: 43520684-43520684 on Build GRCh38
rs745790012 Chr.6: 43520794-43520794 on Build GRCh38
rs773684449 Chr.6: 43520605-43520605 on Build GRCh38
rs892012657 Chr.6: 43520674-43520674 on Build GRCh38
rs533051491 Chr.6: 43520761-43520761 on Build GRCh38
rs113383614 Chr.6: 43520616-43520616 on Build GRCh38
rs1033608704 Chr.6: 43520664-43520664 on Build GRCh38
rs995232530 Chr.6: 43520862-43520862 on Build GRCh38
rs762491554 Chr.6: 43520653-43520653 on Build GRCh38
rs766857713 Chr.6: 43520758-43520758 on Build GRCh38
rs757786256 Chr.6: 43520581-43520581 on Build GRCh38
rs1026712855 Chr.6: 43520879-43520879 on Build GRCh38
rs111455262 Chr.6: 43520798-43520798 on Build GRCh38
rs759144195 Chr.6: 43520639-43520639 on Build GRCh38
rs971703418 Chr.6: 43520687-43520687 on Build GRCh38
rs999345252 Chr.6: 43520578-43520578 on Build GRCh38
rs759426384 Chr.6: 43520735-43520735 on Build GRCh38
rs755407076 Chr.6: 43520768-43520768 on Build GRCh38
rs765798937 Chr.6: 43520991-43520991 on Build GRCh38
rs200712637 Chr.6: 43520606-43520606 on Build GRCh38
rs748187791 Chr.6: 43520611-43520611 on Build GRCh38
rs752324853 Chr.6: 43520649-43520649 on Build GRCh38
rs141156009 Chr.6: 43520961-43520961 on Build GRCh38
rs749037138 Chr.6: 43520956-43520956 on Build GRCh38
rs372836799 Chr.6: 43520999-43520999 on Build GRCh38
rs868788788 Chr.6: 43520848-43520848 on Build GRCh38
rs766901419 Chr.6: 43520822-43520822 on Build GRCh38
rs749203403 Chr.6: 43520956-43520957 on Build GRCh38
rs1003164445 Chr.6: 43520841-43520843 on Build GRCh38
rs961302923 Chr.6: 43520858-43520858 on Build GRCh38
rs903303034 Chr.6: 43520566-43520566 on Build GRCh38
rs374854435 Chr.6: 43521001-43521001 on Build GRCh38
rs753042694 Chr.6: 43520914-43520914 on Build GRCh38
rs563511563 Chr.6: 43520802-43520802 on Build GRCh38
rs762584743 Chr.6: 43520745-43520745 on Build GRCh38
rs755175264 Chr.6: 43520696-43520696 on Build GRCh38
rs376857047 Chr.6: 43520561-43520563 on Build GRCh38
rs373166712 Chr.6: 43520588-43520588 on Build GRCh38
rs910836025 Chr.6: 43520764-43520764 on Build GRCh38
rs756588367 Chr.6: 43520671-43520671 on Build GRCh38
rs146936712 Chr.6: 43520989-43520989 on Build GRCh38
rs777986693 Chr.6: 43520703-43520703 on Build GRCh38
rs1022865475 Chr.6: 43520683-43520683 on Build GRCh38
rs865979957 Chr.6: 43520977-43520977 on Build GRCh38
rs776376952 Chr.6: 43520740-43520740 on Build GRCh38

View Full Product Details