Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43520846-43521345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR1C
Entrez Gene ID
9533
Gene Name
RNA polymerase I subunit C
Gene Aliases
AC40, HLD11, RPA39, RPA40, RPA5, RPAC1, RPC40, TCS3
UniGene
Hs.743509
Gene Chromosome Location
Chr.6: 43517024-43562404 on Build GRCh38
Transcripts
NM_001318876.1 NM_203290.3 XM_011515000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7423733 c.902G>T p.R301L Chr.6: 43521028-43521028 on Build GRCh38
7081419 c.882G>C p.K294N Chr.6: 43521008-43521008 on Build GRCh38
6033655 c.1026G>A p.A342A Chr.6: 43521285-43521285 on Build GRCh38
1754829 c.1032G>A p.Q344Q Chr.6: 43521291-43521291 on Build GRCh38
7181131 c.873G>C p.E291D Chr.6: 43520999-43520999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745835548 Chr.6: 43520935-43520935 on Build GRCh38
rs764460811 Chr.6: 43520926-43520926 on Build GRCh38
rs929224613 Chr.6: 43520880-43520880 on Build GRCh38
rs371041894 Chr.6: 43521207-43521207 on Build GRCh38
rs748680988 Chr.6: 43521286-43521286 on Build GRCh38
rs387907020 Chr.6: 43521238-43521238 on Build GRCh38
rs886061428 Chr.6: 43521039-43521039 on Build GRCh38
rs372409106 Chr.6: 43521049-43521049 on Build GRCh38
rs113321982 Chr.6: 43521156-43521156 on Build GRCh38
rs571910017 Chr.6: 43521025-43521025 on Build GRCh38
rs371479150 Chr.6: 43520971-43520971 on Build GRCh38
rs891995003 Chr.6: 43521007-43521007 on Build GRCh38
rs779858907 Chr.6: 43520954-43520954 on Build GRCh38
rs762136709 Chr.6: 43521202-43521202 on Build GRCh38
rs762129277 Chr.6: 43521131-43521131 on Build GRCh38
rs767589991 Chr.6: 43521132-43521132 on Build GRCh38
rs767639108 Chr.6: 43521040-43521044 on Build GRCh38
rs950580550 Chr.6: 43521119-43521119 on Build GRCh38
rs771555921 Chr.6: 43521060-43521060 on Build GRCh38
rs765709261 Chr.6: 43520901-43520901 on Build GRCh38
rs770534885 Chr.6: 43521070-43521070 on Build GRCh38
rs747233730 Chr.6: 43521301-43521301 on Build GRCh38
rs770624952 Chr.6: 43521196-43521196 on Build GRCh38
rs778556895 Chr.6: 43521050-43521050 on Build GRCh38
rs774805301 Chr.6: 43521201-43521201 on Build GRCh38
rs184144591 Chr.6: 43521151-43521151 on Build GRCh38
rs996036811 Chr.6: 43521092-43521092 on Build GRCh38
rs897110459 Chr.6: 43521271-43521271 on Build GRCh38
rs749820918 Chr.6: 43521198-43521198 on Build GRCh38
rs747749279 Chr.6: 43520968-43520968 on Build GRCh38
rs191582628 Chr.6: 43520962-43520962 on Build GRCh38
rs754344123 Chr.6: 43521248-43521249 on Build GRCh38
rs760123164 Chr.6: 43520990-43520990 on Build GRCh38
rs758724481 Chr.6: 43520924-43520924 on Build GRCh38
rs910709453 Chr.6: 43520917-43520917 on Build GRCh38
rs953781624 Chr.6: 43520910-43520910 on Build GRCh38
rs769080013 Chr.6: 43521086-43521086 on Build GRCh38
rs113209557 Chr.6: 43521098-43521098 on Build GRCh38
rs746655084 Chr.6: 43521195-43521195 on Build GRCh38
rs759946050 Chr.6: 43520893-43520893 on Build GRCh38
rs983311637 Chr.6: 43521120-43521120 on Build GRCh38
rs781119795 Chr.6: 43520932-43520932 on Build GRCh38
rs1010480660 Chr.6: 43521012-43521012 on Build GRCh38
rs766457547 Chr.6: 43521245-43521245 on Build GRCh38
rs764550766 Chr.6: 43521009-43521009 on Build GRCh38
rs751702117 Chr.6: 43520929-43520929 on Build GRCh38
rs924649314 Chr.6: 43520952-43520952 on Build GRCh38
rs780884071 Chr.6: 43521283-43521283 on Build GRCh38
rs758194396 Chr.6: 43521262-43521262 on Build GRCh38
rs776119212 Chr.6: 43520992-43520992 on Build GRCh38
rs758090769 Chr.6: 43521187-43521187 on Build GRCh38
rs750625332 Chr.6: 43521135-43521135 on Build GRCh38
rs763542809 Chr.6: 43521076-43521076 on Build GRCh38
rs777698504 Chr.6: 43521263-43521263 on Build GRCh38
rs749189234 Chr.6: 43521034-43521034 on Build GRCh38
rs775455943 Chr.6: 43520917-43520920 on Build GRCh38
rs753847505 Chr.6: 43521252-43521252 on Build GRCh38
rs201731980 Chr.6: 43521064-43521064 on Build GRCh38
rs535176929 Chr.6: 43520949-43520949 on Build GRCh38
rs376732667 Chr.6: 43521259-43521259 on Build GRCh38
rs751272699 Chr.6: 43521270-43521270 on Build GRCh38
rs375077426 Chr.6: 43521156-43521160 on Build GRCh38
rs754848781 Chr.6: 43521177-43521177 on Build GRCh38
rs112057763 Chr.6: 43521303-43521303 on Build GRCh38
rs767791371 Chr.6: 43521133-43521134 on Build GRCh38
rs780012379 Chr.6: 43521028-43521028 on Build GRCh38
rs563473186 Chr.6: 43521033-43521033 on Build GRCh38
rs875989826 Chr.6: 43521009-43521011 on Build GRCh38
rs772375161 Chr.6: 43521289-43521289 on Build GRCh38
rs756072030 Chr.6: 43521027-43521027 on Build GRCh38
rs921596766 Chr.6: 43521199-43521199 on Build GRCh38
rs773485565 Chr.6: 43521213-43521213 on Build GRCh38
rs1026712855 Chr.6: 43520879-43520879 on Build GRCh38
rs181576159 Chr.6: 43521197-43521197 on Build GRCh38
rs774083211 Chr.6: 43521040-43521042 on Build GRCh38
rs765798937 Chr.6: 43520991-43520991 on Build GRCh38
rs141156009 Chr.6: 43520961-43520961 on Build GRCh38
rs769006912 Chr.6: 43521200-43521200 on Build GRCh38
rs757367771 Chr.6: 43521020-43521020 on Build GRCh38
rs376488921 Chr.6: 43521176-43521176 on Build GRCh38
rs749037138 Chr.6: 43520956-43520956 on Build GRCh38
rs564038848 Chr.6: 43521285-43521285 on Build GRCh38
rs372836799 Chr.6: 43520999-43520999 on Build GRCh38
rs386352351 Chr.6: 43521008-43521008 on Build GRCh38
rs776205610 Chr.6: 43521073-43521073 on Build GRCh38
rs749203403 Chr.6: 43520956-43520957 on Build GRCh38
rs752393230 Chr.6: 43521185-43521185 on Build GRCh38
rs543729152 Chr.6: 43521192-43521192 on Build GRCh38
rs777338518 Chr.6: 43521188-43521188 on Build GRCh38
rs778772237 Chr.6: 43521183-43521183 on Build GRCh38
rs760799818 Chr.6: 43521160-43521160 on Build GRCh38
rs374854435 Chr.6: 43521001-43521001 on Build GRCh38
rs1017782101 Chr.6: 43521126-43521129 on Build GRCh38
rs753042694 Chr.6: 43520914-43520914 on Build GRCh38
rs760780461 Chr.6: 43521139-43521139 on Build GRCh38
rs760870530 Chr.6: 43521242-43521242 on Build GRCh38
rs777373707 Chr.6: 43521063-43521063 on Build GRCh38
rs137871425 Chr.6: 43521043-43521043 on Build GRCh38
rs148998616 Chr.6: 43521153-43521157 on Build GRCh38
rs766394808 Chr.6: 43521160-43521161 on Build GRCh38
rs889417996 Chr.6: 43521299-43521299 on Build GRCh38
rs747848622 Chr.6: 43521055-43521055 on Build GRCh38
rs759546887 Chr.6: 43521253-43521253 on Build GRCh38
rs767642581 Chr.6: 43521021-43521021 on Build GRCh38
rs370935238 Chr.6: 43521305-43521305 on Build GRCh38
rs140888409 Chr.6: 43521284-43521284 on Build GRCh38
rs773505724 Chr.6: 43521291-43521291 on Build GRCh38
rs146936712 Chr.6: 43520989-43520989 on Build GRCh38
rs557660342 Chr.6: 43521115-43521115 on Build GRCh38
rs769188761 Chr.6: 43521174-43521174 on Build GRCh38
rs751867271 Chr.6: 43521013-43521013 on Build GRCh38
rs752657369 Chr.6: 43521260-43521260 on Build GRCh38
rs865979957 Chr.6: 43520977-43520977 on Build GRCh38
rs756904557 Chr.6: 43521277-43521277 on Build GRCh38

View Full Product Details