Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44826609-44827113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUPT3H
Entrez Gene ID
8464
Gene Name
SPT3 homolog, SAGA and STAGA complex component
Gene Aliases
SPT3, SPT3L
UniGene
Hs.368325
Gene Chromosome Location
Chr.6: 44808844-45378051 on Build GRCh38
Transcripts
NM_001261823.1 NM_003599.3 NM_181356.2

SNP Detail

SNP ID Chromosome Location
rs1046498228 Chr.6: 44826710-44826710 on Build GRCh38
rs548463280 Chr.6: 44826672-44826672 on Build GRCh38
rs554395192 Chr.6: 44827021-44827021 on Build GRCh38
rs1019741464 Chr.6: 44826950-44826950 on Build GRCh38
rs1020215348 Chr.6: 44826687-44826687 on Build GRCh38
rs751146085 Chr.6: 44826953-44826953 on Build GRCh38
rs756797996 Chr.6: 44827019-44827019 on Build GRCh38
rs764650895 Chr.6: 44826871-44826871 on Build GRCh38
rs192809103 Chr.6: 44826993-44826993 on Build GRCh38
rs560094064 Chr.6: 44826704-44826704 on Build GRCh38
rs1007939816 Chr.6: 44826944-44826944 on Build GRCh38
rs939912998 Chr.6: 44826803-44826803 on Build GRCh38
rs907993157 Chr.6: 44827060-44827060 on Build GRCh38
rs1046295073 Chr.6: 44826798-44826798 on Build GRCh38
rs890608070 Chr.6: 44826898-44826898 on Build GRCh38
rs899372882 Chr.6: 44826855-44826855 on Build GRCh38
rs970770458 Chr.6: 44827014-44827015 on Build GRCh38
rs993687144 Chr.6: 44826862-44826862 on Build GRCh38
rs907828267 Chr.6: 44826802-44826802 on Build GRCh38
rs767155769 Chr.6: 44827013-44827014 on Build GRCh38
rs958018403 Chr.6: 44826716-44826716 on Build GRCh38
rs543982097 Chr.6: 44826796-44826799 on Build GRCh38
rs149525797 Chr.6: 44826956-44826956 on Build GRCh38
rs1037972541 Chr.6: 44826821-44826821 on Build GRCh38
rs925839400 Chr.6: 44826660-44826660 on Build GRCh38
rs758991244 Chr.6: 44826671-44826671 on Build GRCh38
rs898451809 Chr.6: 44826827-44826827 on Build GRCh38
rs978662264 Chr.6: 44826642-44826642 on Build GRCh38
rs549796554 Chr.6: 44826948-44826948 on Build GRCh38
rs1008633105 Chr.6: 44826909-44826909 on Build GRCh38
rs1037687313 Chr.6: 44826817-44826817 on Build GRCh38
rs1023656733 Chr.6: 44826999-44826999 on Build GRCh38
rs900196184 Chr.6: 44826928-44826928 on Build GRCh38
rs535912102 Chr.6: 44826888-44826888 on Build GRCh38
rs949486506 Chr.6: 44826686-44826686 on Build GRCh38
rs537926060 Chr.6: 44826787-44826787 on Build GRCh38
rs978754098 Chr.6: 44826709-44826709 on Build GRCh38
rs902680106 Chr.6: 44826952-44826952 on Build GRCh38
rs889561063 Chr.6: 44826679-44826679 on Build GRCh38
rs926637975 Chr.6: 44826738-44826738 on Build GRCh38
rs958297346 Chr.6: 44826962-44826962 on Build GRCh38
rs917080671 Chr.6: 44826680-44826680 on Build GRCh38
rs964335512 Chr.6: 44826638-44826638 on Build GRCh38
rs527441240 Chr.6: 44826722-44826722 on Build GRCh38
rs138089480 Chr.6: 44826771-44826771 on Build GRCh38
rs1049408717 Chr.6: 44826896-44826896 on Build GRCh38
rs565656672 Chr.6: 44826701-44826701 on Build GRCh38
rs563364937 Chr.6: 44826866-44826866 on Build GRCh38
rs940669608 Chr.6: 44826810-44826810 on Build GRCh38
rs531246328 Chr.6: 44826875-44826875 on Build GRCh38
rs552406269 Chr.6: 44826764-44826764 on Build GRCh38
rs760732448 Chr.6: 44826795-44826796 on Build GRCh38
rs147216526 Chr.6: 44827055-44827055 on Build GRCh38
rs1017280654 Chr.6: 44826915-44826915 on Build GRCh38
rs371454105 Chr.6: 44826673-44826673 on Build GRCh38
rs916651889 Chr.6: 44826736-44826736 on Build GRCh38
rs369036274 Chr.6: 44826773-44826773 on Build GRCh38
rs780948507 Chr.6: 44827028-44827028 on Build GRCh38

View Full Product Details