Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52404183-52404693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAQR8
Entrez Gene ID
85315
Gene Name
progestin and adipoQ receptor family member 8
Gene Aliases
C6orf33, LMPB1, MPRB
UniGene
Hs.239388
Gene Chromosome Location
Chr.6: 52362128-52407777 on Build GRCh38
Transcripts
NM_133367.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3874772 c.1012G>A p.A338T Chr.6: 52404225-52404225 on Build GRCh38
1080239 c.1039G>A p.A347T Chr.6: 52404252-52404252 on Build GRCh38
1634849 c.1035C>G p.V345V Chr.6: 52404248-52404248 on Build GRCh38
3874771 c.976T>G p.S326A Chr.6: 52404189-52404189 on Build GRCh38
1445161 c.978_980delCTT p.F329delF Chr.6: 52404191-52404193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575802052 Chr.6: 52404614-52404614 on Build GRCh38
rs775236580 Chr.6: 52404284-52404284 on Build GRCh38
rs1042597060 Chr.6: 52404252-52404252 on Build GRCh38
rs764000675 Chr.6: 52404280-52404280 on Build GRCh38
rs573184173 Chr.6: 52404400-52404400 on Build GRCh38
rs655113 Chr.6: 52404353-52404353 on Build GRCh38
rs530300909 Chr.6: 52404654-52404654 on Build GRCh38
rs1051256732 Chr.6: 52404370-52404370 on Build GRCh38
rs750581614 Chr.6: 52404309-52404309 on Build GRCh38
rs753676972 Chr.6: 52404299-52404299 on Build GRCh38
rs556884940 Chr.6: 52404270-52404270 on Build GRCh38
rs932687417 Chr.6: 52404365-52404365 on Build GRCh38
rs767424580 Chr.6: 52404214-52404214 on Build GRCh38
rs889993113 Chr.6: 52404373-52404373 on Build GRCh38
rs1000346639 Chr.6: 52404558-52404558 on Build GRCh38
rs1034852882 Chr.6: 52404578-52404578 on Build GRCh38
rs915344992 Chr.6: 52404632-52404632 on Build GRCh38
rs575760055 Chr.6: 52404539-52404539 on Build GRCh38
rs940310494 Chr.6: 52404440-52404440 on Build GRCh38
rs765196798 Chr.6: 52404614-52404615 on Build GRCh38
rs763959251 Chr.6: 52404297-52404297 on Build GRCh38
rs990825157 Chr.6: 52404587-52404587 on Build GRCh38
rs747152836 Chr.6: 52404224-52404224 on Build GRCh38
rs552630869 Chr.6: 52404464-52404464 on Build GRCh38
rs3799272 Chr.6: 52404401-52404401 on Build GRCh38
rs898806896 Chr.6: 52404443-52404443 on Build GRCh38
rs533009816 Chr.6: 52404574-52404574 on Build GRCh38
rs758736954 Chr.6: 52404207-52404207 on Build GRCh38
rs1003908283 Chr.6: 52404549-52404549 on Build GRCh38
rs1044506565 Chr.6: 52404477-52404477 on Build GRCh38
rs562307065 Chr.6: 52404335-52404335 on Build GRCh38
rs558534530 Chr.6: 52404545-52404545 on Build GRCh38
rs989120235 Chr.6: 52404244-52404244 on Build GRCh38
rs959475445 Chr.6: 52404583-52404583 on Build GRCh38
rs572917203 Chr.6: 52404481-52404481 on Build GRCh38
rs561607850 Chr.6: 52404626-52404626 on Build GRCh38
rs75489244 Chr.6: 52404509-52404509 on Build GRCh38
rs761912288 Chr.6: 52404303-52404303 on Build GRCh38
rs773890042 Chr.6: 52404262-52404262 on Build GRCh38
rs760174888 Chr.6: 52404292-52404292 on Build GRCh38
rs994516593 Chr.6: 52404446-52404446 on Build GRCh38
rs780302848 Chr.6: 52404218-52404218 on Build GRCh38
rs544884854 Chr.6: 52404623-52404623 on Build GRCh38
rs769096577 Chr.6: 52404225-52404225 on Build GRCh38
rs3799274 Chr.6: 52404248-52404248 on Build GRCh38
rs759022615 Chr.6: 52404266-52404266 on Build GRCh38
rs112982203 Chr.6: 52404451-52404451 on Build GRCh38
rs1032705911 Chr.6: 52404572-52404572 on Build GRCh38
rs1026012577 Chr.6: 52404505-52404505 on Build GRCh38

View Full Product Details