Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 53128137-53128643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM1
Entrez Gene ID
8521
Gene Name
glial cells missing homolog 1
Gene Aliases
GCMA, hGCMa
UniGene
Hs.23589
Gene Chromosome Location
Chr.6: 53125644-53262803 on Build GRCh38
Transcripts
NM_003643.3 XM_017011390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
742516 c.1195C>T p.Q399* Chr.6: 53128322-53128322 on Build GRCh38
4686646 c.1061delC p.P354fs*63 Chr.6: 53128456-53128456 on Build GRCh38
742517 c.1267C>A p.H423N Chr.6: 53128250-53128250 on Build GRCh38
3430756 c.1125C>T p.Y375Y Chr.6: 53128392-53128392 on Build GRCh38
742514 c.1075C>T p.P359S Chr.6: 53128442-53128442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754318983 Chr.6: 53128201-53128201 on Build GRCh38
rs565567636 Chr.6: 53128362-53128362 on Build GRCh38
rs936393185 Chr.6: 53128396-53128396 on Build GRCh38
rs12207430 Chr.6: 53128333-53128333 on Build GRCh38
rs201788899 Chr.6: 53128325-53128325 on Build GRCh38
rs757083741 Chr.6: 53128502-53128502 on Build GRCh38
rs760991885 Chr.6: 53128317-53128317 on Build GRCh38
rs762117591 Chr.6: 53128465-53128465 on Build GRCh38
rs769049481 Chr.6: 53128337-53128337 on Build GRCh38
rs764545605 Chr.6: 53128189-53128189 on Build GRCh38
rs772167468 Chr.6: 53128432-53128432 on Build GRCh38
rs867455356 Chr.6: 53128274-53128274 on Build GRCh38
rs962286314 Chr.6: 53128316-53128316 on Build GRCh38
rs781353031 Chr.6: 53128551-53128551 on Build GRCh38
rs139786380 Chr.6: 53128464-53128464 on Build GRCh38
rs200632700 Chr.6: 53128306-53128306 on Build GRCh38
rs147283338 Chr.6: 53128411-53128411 on Build GRCh38
rs773767712 Chr.6: 53128594-53128594 on Build GRCh38
rs762311742 Chr.6: 53128352-53128352 on Build GRCh38
rs376111326 Chr.6: 53128597-53128597 on Build GRCh38
rs12207437 Chr.6: 53128384-53128384 on Build GRCh38
rs759307890 Chr.6: 53128232-53128232 on Build GRCh38
rs753713611 Chr.6: 53128496-53128496 on Build GRCh38
rs755215513 Chr.6: 53128547-53128547 on Build GRCh38
rs760060174 Chr.6: 53128612-53128612 on Build GRCh38
rs1028293636 Chr.6: 53128488-53128488 on Build GRCh38
rs370422081 Chr.6: 53128437-53128437 on Build GRCh38
rs145721869 Chr.6: 53128539-53128539 on Build GRCh38
rs1028445706 Chr.6: 53128223-53128223 on Build GRCh38
rs367768924 Chr.6: 53128410-53128410 on Build GRCh38
rs765868524 Chr.6: 53128218-53128218 on Build GRCh38
rs760382550 Chr.6: 53128491-53128491 on Build GRCh38
rs888545500 Chr.6: 53128490-53128490 on Build GRCh38
rs759019503 Chr.6: 53128475-53128475 on Build GRCh38
rs765311065 Chr.6: 53128511-53128511 on Build GRCh38
rs961931855 Chr.6: 53128552-53128552 on Build GRCh38
rs956295097 Chr.6: 53128420-53128420 on Build GRCh38
rs747001987 Chr.6: 53128546-53128546 on Build GRCh38
rs1023368921 Chr.6: 53128336-53128336 on Build GRCh38
rs758571009 Chr.6: 53128598-53128598 on Build GRCh38
rs12207375 Chr.6: 53128198-53128198 on Build GRCh38
rs777710650 Chr.6: 53128397-53128397 on Build GRCh38
rs575787885 Chr.6: 53128392-53128392 on Build GRCh38
rs779142740 Chr.6: 53128280-53128280 on Build GRCh38
rs767117665 Chr.6: 53128373-53128373 on Build GRCh38
rs552574680 Chr.6: 53128302-53128302 on Build GRCh38
rs879100265 Chr.6: 53128391-53128391 on Build GRCh38
rs775083311 Chr.6: 53128601-53128601 on Build GRCh38
rs750359033 Chr.6: 53128534-53128534 on Build GRCh38
rs149786327 Chr.6: 53128476-53128476 on Build GRCh38
rs1010132815 Chr.6: 53128431-53128431 on Build GRCh38
rs768694108 Chr.6: 53128463-53128463 on Build GRCh38
rs952771233 Chr.6: 53128259-53128259 on Build GRCh38
rs13200319 Chr.6: 53128204-53128204 on Build GRCh38
rs372692867 Chr.6: 53128166-53128166 on Build GRCh38
rs139699761 Chr.6: 53128249-53128249 on Build GRCh38
rs749226935 Chr.6: 53128269-53128269 on Build GRCh38
rs377142806 Chr.6: 53128355-53128355 on Build GRCh38
rs777623767 Chr.6: 53128266-53128266 on Build GRCh38
rs754600067 Chr.6: 53128221-53128221 on Build GRCh38
rs752375447 Chr.6: 53128375-53128375 on Build GRCh38
rs551536061 Chr.6: 53128354-53128354 on Build GRCh38
rs771205459 Chr.6: 53128277-53128277 on Build GRCh38
rs748552826 Chr.6: 53128589-53128589 on Build GRCh38
rs780186014 Chr.6: 53128542-53128542 on Build GRCh38
rs930646909 Chr.6: 53128617-53128617 on Build GRCh38
rs145233025 Chr.6: 53128265-53128265 on Build GRCh38
rs745971206 Chr.6: 53128414-53128414 on Build GRCh38
rs866753184 Chr.6: 53128442-53128442 on Build GRCh38
rs200433538 Chr.6: 53128438-53128438 on Build GRCh38
rs200656203 Chr.6: 53128469-53128469 on Build GRCh38
rs775075424 Chr.6: 53128487-53128487 on Build GRCh38
rs1005615503 Chr.6: 53128505-53128505 on Build GRCh38
rs972299340 Chr.6: 53128576-53128576 on Build GRCh38
rs770122746 Chr.6: 53128590-53128590 on Build GRCh38
rs1129069 Chr.6: 53128360-53128360 on Build GRCh38
rs12207434 Chr.6: 53128347-53128347 on Build GRCh38
rs372691544 Chr.6: 53128308-53128308 on Build GRCh38
rs761058673 Chr.6: 53128178-53128178 on Build GRCh38
rs763785660 Chr.6: 53128493-53128493 on Build GRCh38
rs751004414 Chr.6: 53128220-53128220 on Build GRCh38
rs777186773 Chr.6: 53128338-53128338 on Build GRCh38

View Full Product Details