Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 69701162-69701709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMBRD1
Entrez Gene ID
55788
Gene Name
LMBR1 domain containing 1
Gene Aliases
C6orf209, LMBD1, MAHCF, NESI
UniGene
Hs.271643
Gene Chromosome Location
Chr.6: 69675749-69797157 on Build GRCh38
Transcripts
NM_018368.3 XM_011535941.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586878 c.1056delG p.N353fs*18 Chr.6: 69701470-69701470 on Build GRCh38
321408 c.1045A>T p.S349C Chr.6: 69701481-69701481 on Build GRCh38
6107184 c.1033G>A p.G345R Chr.6: 69701493-69701493 on Build GRCh38
1256537 c.985G>C p.D329H Chr.6: 69701541-69701541 on Build GRCh38
5351325 c.981-2_981-1insT p.? Chr.6: 69701546-69701547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749272546 Chr.6: 69701470-69701470 on Build GRCh38
rs2445969 Chr.6: 69701265-69701265 on Build GRCh38
rs963856458 Chr.6: 69701193-69701193 on Build GRCh38
rs779824573 Chr.6: 69701588-69701588 on Build GRCh38
rs753360068 Chr.6: 69701561-69701561 on Build GRCh38
rs558926049 Chr.6: 69701509-69701509 on Build GRCh38
rs1057244740 Chr.6: 69701390-69701390 on Build GRCh38
rs921039250 Chr.6: 69701509-69701509 on Build GRCh38
rs377103194 Chr.6: 69701575-69701575 on Build GRCh38
rs760419494 Chr.6: 69701553-69701553 on Build GRCh38
rs751906360 Chr.6: 69701543-69701543 on Build GRCh38
rs758289173 Chr.6: 69701585-69701585 on Build GRCh38
rs912202285 Chr.6: 69701239-69701239 on Build GRCh38
rs1019380464 Chr.6: 69701641-69701641 on Build GRCh38
rs982512804 Chr.6: 69701617-69701617 on Build GRCh38
rs181787663 Chr.6: 69701475-69701475 on Build GRCh38
rs1045212189 Chr.6: 69701334-69701334 on Build GRCh38
rs556720958 Chr.6: 69701332-69701332 on Build GRCh38
rs951109210 Chr.6: 69701207-69701207 on Build GRCh38
rs149269326 Chr.6: 69701237-69701237 on Build GRCh38
rs771835523 Chr.6: 69701472-69701472 on Build GRCh38
rs934735779 Chr.6: 69701388-69701388 on Build GRCh38
rs778769611 Chr.6: 69701563-69701563 on Build GRCh38
rs975714665 Chr.6: 69701262-69701262 on Build GRCh38
rs903062004 Chr.6: 69701356-69701356 on Build GRCh38
rs1011399688 Chr.6: 69701312-69701312 on Build GRCh38
rs750386636 Chr.6: 69701580-69701580 on Build GRCh38
rs943742767 Chr.6: 69701248-69701248 on Build GRCh38
rs772467119 Chr.6: 69701573-69701574 on Build GRCh38
rs965391220 Chr.6: 69701653-69701653 on Build GRCh38
rs189820560 Chr.6: 69701336-69701336 on Build GRCh38
rs765215874 Chr.6: 69701503-69701503 on Build GRCh38
rs766762873 Chr.6: 69701235-69701235 on Build GRCh38
rs202207965 Chr.6: 69701547-69701548 on Build GRCh38
rs766881860 Chr.6: 69701537-69701537 on Build GRCh38
rs753333905 Chr.6: 69701417-69701417 on Build GRCh38
rs757203257 Chr.6: 69701419-69701419 on Build GRCh38
rs768797474 Chr.6: 69701485-69701485 on Build GRCh38
rs755788958 Chr.6: 69701400-69701400 on Build GRCh38
rs745694228 Chr.6: 69701451-69701451 on Build GRCh38
rs915391643 Chr.6: 69701605-69701606 on Build GRCh38
rs756762965 Chr.6: 69701562-69701562 on Build GRCh38
rs376835097 Chr.6: 69701469-69701469 on Build GRCh38
rs779771467 Chr.6: 69701477-69701477 on Build GRCh38
rs75847296 Chr.6: 69701556-69701556 on Build GRCh38
rs973576681 Chr.6: 69701200-69701200 on Build GRCh38
rs752884512 Chr.6: 69701613-69701613 on Build GRCh38
rs1034907686 Chr.6: 69701383-69701383 on Build GRCh38
rs1003574625 Chr.6: 69701382-69701382 on Build GRCh38
rs763459076 Chr.6: 69701534-69701534 on Build GRCh38
rs778902430 Chr.6: 69701548-69701548 on Build GRCh38
rs770269966 Chr.6: 69701514-69701514 on Build GRCh38
rs747258498 Chr.6: 69701478-69701478 on Build GRCh38
rs947185670 Chr.6: 69701318-69701318 on Build GRCh38
rs777152526 Chr.6: 69701404-69701404 on Build GRCh38
rs749573257 Chr.6: 69701510-69701511 on Build GRCh38
rs778907355 Chr.6: 69701427-69701427 on Build GRCh38
rs773738182 Chr.6: 69701529-69701529 on Build GRCh38
rs756288021 Chr.6: 69701193-69701194 on Build GRCh38
rs919636224 Chr.6: 69701203-69701203 on Build GRCh38
rs912156834 Chr.6: 69701220-69701220 on Build GRCh38

View Full Product Details