Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 88611310-88611821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNGTT
Entrez Gene ID
8732
Gene Name
RNA guanylyltransferase and 5'-phosphatase
Gene Aliases
CAP1A, HCE, HCE1, hCAP
UniGene
Hs.567378
Gene Chromosome Location
Chr.6: 88609897-88963716 on Build GRCh38
Transcripts
NM_001286426.1 NM_001286428.1 NM_003800.4 XM_017011401.1

SNP Detail

SNP ID Chromosome Location
rs536889713 Chr.6: 88611696-88611696 on Build GRCh38
rs989114327 Chr.6: 88611667-88611667 on Build GRCh38
rs994298617 Chr.6: 88611719-88611719 on Build GRCh38
rs548279639 Chr.6: 88611624-88611624 on Build GRCh38
rs928597719 Chr.6: 88611598-88611598 on Build GRCh38
rs12662216 Chr.6: 88611524-88611524 on Build GRCh38
rs1026035436 Chr.6: 88611336-88611336 on Build GRCh38
rs978352058 Chr.6: 88611686-88611686 on Build GRCh38
rs1016040805 Chr.6: 88611797-88611797 on Build GRCh38
rs984451421 Chr.6: 88611410-88611410 on Build GRCh38
rs984935477 Chr.6: 88611796-88611796 on Build GRCh38
rs938512922 Chr.6: 88611604-88611604 on Build GRCh38
rs950419596 Chr.6: 88611729-88611729 on Build GRCh38
rs908884760 Chr.6: 88611486-88611486 on Build GRCh38
rs375674612 Chr.6: 88611529-88611548 on Build GRCh38
rs1043174855 Chr.6: 88611625-88611625 on Build GRCh38
rs1005480747 Chr.6: 88611509-88611509 on Build GRCh38
rs939251821 Chr.6: 88611392-88611392 on Build GRCh38
rs148641569 Chr.6: 88611674-88611674 on Build GRCh38
rs1049615191 Chr.6: 88611426-88611426 on Build GRCh38
rs902126896 Chr.6: 88611589-88611589 on Build GRCh38
rs953697741 Chr.6: 88611641-88611641 on Build GRCh38
rs566044394 Chr.6: 88611532-88611532 on Build GRCh38
rs1022473597 Chr.6: 88611673-88611673 on Build GRCh38
rs879101807 Chr.6: 88611699-88611699 on Build GRCh38
rs768557539 Chr.6: 88611531-88611531 on Build GRCh38
rs144934991 Chr.6: 88611410-88611411 on Build GRCh38
rs1052884588 Chr.6: 88611608-88611608 on Build GRCh38
rs950450227 Chr.6: 88611393-88611393 on Build GRCh38
rs889765358 Chr.6: 88611635-88611635 on Build GRCh38
rs916459026 Chr.6: 88611750-88611750 on Build GRCh38
rs948443425 Chr.6: 88611550-88611550 on Build GRCh38
rs140351304 Chr.6: 88611520-88611539 on Build GRCh38
rs930767591 Chr.6: 88611417-88611417 on Build GRCh38
rs186220225 Chr.6: 88611785-88611785 on Build GRCh38
rs66919167 Chr.6: 88611524-88611543 on Build GRCh38
rs188989261 Chr.6: 88611595-88611595 on Build GRCh38
rs113597955 Chr.6: 88611411-88611412 on Build GRCh38
rs927805784 Chr.6: 88611700-88611700 on Build GRCh38
rs891855542 Chr.6: 88611615-88611615 on Build GRCh38
rs901431747 Chr.6: 88611687-88611687 on Build GRCh38
rs1019870206 Chr.6: 88611680-88611680 on Build GRCh38
rs866458820 Chr.6: 88611404-88611404 on Build GRCh38
rs144363758 Chr.6: 88611723-88611723 on Build GRCh38
rs1049033991 Chr.6: 88611792-88611792 on Build GRCh38
rs181406823 Chr.6: 88611597-88611597 on Build GRCh38

View Full Product Details