Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96897585-96898086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL32
Entrez Gene ID
114792
Gene Name
kelch like family member 32
Gene Aliases
BKLHD5, KIAA1900, UG0030H05, dJ21F7.1
UniGene
Hs.45056
Gene Chromosome Location
Chr.6: 96898083-97142949 on Build GRCh38
Transcripts
NM_001323258.1 NM_014165.3

Gene Symbol
NDUFAF4
Entrez Gene ID
29078
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 4
Gene Aliases
C6orf66, HRPAP20, HSPC125, My013, bA22L21.1
UniGene
Hs.512144
Gene Chromosome Location
Chr.6: 96889311-96897891 on Build GRCh38
Transcripts
NM_001323258.1 NM_014165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056351 c.83C>A p.S28Y Chr.6: 96897719-96897719 on Build GRCh38
1082733 c.3G>A p.M1I Chr.6: 96897799-96897799 on Build GRCh38
6723729 c.115C>A p.L39I Chr.6: 96897687-96897687 on Build GRCh38
7401884 c.43G>C p.E15Q Chr.6: 96897759-96897759 on Build GRCh38
6723730 c.54G>A p.A18A Chr.6: 96897748-96897748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758930066 Chr.6: 96897711-96897711 on Build GRCh38
rs978245262 Chr.6: 96898045-96898045 on Build GRCh38
rs749427985 Chr.6: 96897841-96897841 on Build GRCh38
rs558262843 Chr.6: 96897801-96897801 on Build GRCh38
rs540670288 Chr.6: 96897888-96897888 on Build GRCh38
rs545609423 Chr.6: 96898014-96898014 on Build GRCh38
rs765933181 Chr.6: 96897820-96897820 on Build GRCh38
rs931710746 Chr.6: 96897858-96897858 on Build GRCh38
rs567620057 Chr.6: 96898046-96898046 on Build GRCh38
rs747350041 Chr.6: 96897718-96897718 on Build GRCh38
rs1015181678 Chr.6: 96897977-96897977 on Build GRCh38
rs921570712 Chr.6: 96897992-96897992 on Build GRCh38
rs755717389 Chr.6: 96897648-96897653 on Build GRCh38
rs571796525 Chr.6: 96897812-96897812 on Build GRCh38
rs112722560 Chr.6: 96897738-96897738 on Build GRCh38
rs766147748 Chr.6: 96897966-96897966 on Build GRCh38
rs554094360 Chr.6: 96897896-96897896 on Build GRCh38
rs748262976 Chr.6: 96897724-96897724 on Build GRCh38
rs989798451 Chr.6: 96898023-96898023 on Build GRCh38
rs752441413 Chr.6: 96897670-96897670 on Build GRCh38
rs752120764 Chr.6: 96897823-96897823 on Build GRCh38
rs201495784 Chr.6: 96897629-96897629 on Build GRCh38
rs863224091 Chr.6: 96897781-96897782 on Build GRCh38
rs960782124 Chr.6: 96897872-96897872 on Build GRCh38
rs939589755 Chr.6: 96898053-96898053 on Build GRCh38
rs1042989076 Chr.6: 96897901-96897901 on Build GRCh38
rs760457567 Chr.6: 96897744-96897744 on Build GRCh38
rs983712762 Chr.6: 96897809-96897809 on Build GRCh38
rs749336430 Chr.6: 96897793-96897793 on Build GRCh38
rs543270459 Chr.6: 96897909-96897909 on Build GRCh38
rs771976184 Chr.6: 96897739-96897739 on Build GRCh38
rs780496036 Chr.6: 96897623-96897623 on Build GRCh38
rs762543854 Chr.6: 96897818-96897818 on Build GRCh38
rs1049846948 Chr.6: 96897717-96897717 on Build GRCh38
rs761952170 Chr.6: 96897635-96897635 on Build GRCh38
rs370052081 Chr.6: 96897826-96897826 on Build GRCh38
rs920427894 Chr.6: 96897606-96897606 on Build GRCh38
rs774362134 Chr.6: 96897637-96897637 on Build GRCh38
rs768499691 Chr.6: 96897804-96897804 on Build GRCh38
rs200882759 Chr.6: 96897893-96897894 on Build GRCh38
rs781347640 Chr.6: 96897789-96897789 on Build GRCh38
rs944367172 Chr.6: 96897630-96897630 on Build GRCh38
rs985481 Chr.6: 96898025-96898025 on Build GRCh38
rs970324494 Chr.6: 96897857-96897857 on Build GRCh38
rs200056339 Chr.6: 96897859-96897860 on Build GRCh38
rs750271319 Chr.6: 96897879-96897879 on Build GRCh38
rs1052748213 Chr.6: 96898039-96898039 on Build GRCh38
rs752031326 Chr.6: 96897787-96897787 on Build GRCh38
rs767332980 Chr.6: 96897668-96897668 on Build GRCh38
rs144627695 Chr.6: 96897709-96897709 on Build GRCh38
rs767134572 Chr.6: 96897833-96897833 on Build GRCh38
rs547822298 Chr.6: 96898040-96898040 on Build GRCh38
rs1011479272 Chr.6: 96898057-96898057 on Build GRCh38
rs62415287 Chr.6: 96898016-96898016 on Build GRCh38
rs747262076 Chr.6: 96897626-96897626 on Build GRCh38
rs1027720456 Chr.6: 96897961-96897961 on Build GRCh38
rs748435560 Chr.6: 96897790-96897790 on Build GRCh38
rs965951024 Chr.6: 96897999-96897999 on Build GRCh38
rs372137098 Chr.6: 96897721-96897721 on Build GRCh38
rs773015545 Chr.6: 96897851-96897851 on Build GRCh38
rs1054148080 Chr.6: 96897875-96897875 on Build GRCh38
rs759404617 Chr.6: 96897646-96897646 on Build GRCh38
rs776618475 Chr.6: 96897631-96897631 on Build GRCh38
rs946869726 Chr.6: 96897897-96897897 on Build GRCh38
rs531960179 Chr.6: 96897976-96897976 on Build GRCh38
rs1022779566 Chr.6: 96897854-96897854 on Build GRCh38
rs943564832 Chr.6: 96897933-96897934 on Build GRCh38
rs1057104490 Chr.6: 96898055-96898055 on Build GRCh38
rs907779757 Chr.6: 96897822-96897822 on Build GRCh38
rs757340492 Chr.6: 96897842-96897842 on Build GRCh38
rs780583714 Chr.6: 96897714-96897714 on Build GRCh38
rs562962419 Chr.6: 96897920-96897920 on Build GRCh38
rs764771728 Chr.6: 96897815-96897815 on Build GRCh38
rs755401964 Chr.6: 96897824-96897824 on Build GRCh38
rs746995133 Chr.6: 96897847-96897847 on Build GRCh38
rs863224092 Chr.6: 96897732-96897732 on Build GRCh38
rs886061830 Chr.6: 96897825-96897825 on Build GRCh38
rs147891467 Chr.6: 96897743-96897743 on Build GRCh38
rs779751481 Chr.6: 96897779-96897779 on Build GRCh38
rs755386760 Chr.6: 96897788-96897788 on Build GRCh38
rs149192863 Chr.6: 96897902-96897902 on Build GRCh38
rs768426462 Chr.6: 96897849-96897849 on Build GRCh38
rs759018209 Chr.6: 96897764-96897764 on Build GRCh38
rs766993935 Chr.6: 96897783-96897783 on Build GRCh38
rs939600347 Chr.6: 96897905-96897905 on Build GRCh38
rs886061832 Chr.6: 96897834-96897834 on Build GRCh38
rs895161859 Chr.6: 96897944-96897944 on Build GRCh38
rs1003544056 Chr.6: 96897754-96897754 on Build GRCh38
rs776356978 Chr.6: 96897753-96897753 on Build GRCh38
rs756400083 Chr.6: 96897830-96897830 on Build GRCh38
rs752982123 Chr.6: 96897829-96897829 on Build GRCh38
rs983379029 Chr.6: 96897628-96897628 on Build GRCh38
rs1040143236 Chr.6: 96897934-96897934 on Build GRCh38
rs756243354 Chr.6: 96897792-96897792 on Build GRCh38
rs372549179 Chr.6: 96897845-96897845 on Build GRCh38
rs1056446061 Chr.6: 96897923-96897923 on Build GRCh38
rs760369602 Chr.6: 96897678-96897678 on Build GRCh38
rs139675421 Chr.6: 96897691-96897691 on Build GRCh38
rs1012281047 Chr.6: 96897954-96897954 on Build GRCh38
rs761302733 Chr.6: 96897757-96897757 on Build GRCh38
rs886061831 Chr.6: 96897833-96897833 on Build GRCh38
rs887899460 Chr.6: 96897964-96897964 on Build GRCh38
rs145481564 Chr.6: 96897802-96897802 on Build GRCh38
rs1027460280 Chr.6: 96897800-96897800 on Build GRCh38
rs763710545 Chr.6: 96897749-96897749 on Build GRCh38
rs772927541 Chr.6: 96897817-96897817 on Build GRCh38
rs372635399 Chr.6: 96897850-96897850 on Build GRCh38
rs765102960 Chr.6: 96897986-96897986 on Build GRCh38
rs762629470 Chr.6: 96897853-96897853 on Build GRCh38
rs772575575 Chr.6: 96897615-96897615 on Build GRCh38
rs1035109174 Chr.6: 96897860-96897860 on Build GRCh38
rs976425755 Chr.6: 96898002-96898002 on Build GRCh38
rs769923399 Chr.6: 96897730-96897730 on Build GRCh38
rs117908219 Chr.6: 96897682-96897682 on Build GRCh38
rs773237522 Chr.6: 96897736-96897736 on Build GRCh38
rs145392673 Chr.6: 96897762-96897762 on Build GRCh38

View Full Product Details