Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118816075-118816589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM9
Entrez Gene ID
254394
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Gene Aliases
C6orf61, MCMDC1, ODG4, dJ329L24.1, dJ329L24.3
UniGene
Hs.279008
Gene Chromosome Location
Chr.6: 118813449-118935162 on Build GRCh38
Transcripts
NM_017696.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043240 c.2155C>A p.H719N Chr.6: 118816101-118816101 on Build GRCh38
5606717 c.2027C>T p.S676F Chr.6: 118816229-118816229 on Build GRCh38
6871925 c.2066C>T p.T689I Chr.6: 118816190-118816190 on Build GRCh38
7192905 c.2050G>A p.E684K Chr.6: 118816206-118816206 on Build GRCh38
7280656 c.2167A>G p.N723D Chr.6: 118816089-118816089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039957911 Chr.6: 118816541-118816541 on Build GRCh38
rs75607191 Chr.6: 118816260-118816260 on Build GRCh38
rs546929886 Chr.6: 118816178-118816178 on Build GRCh38
rs143163834 Chr.6: 118816186-118816186 on Build GRCh38
rs914925149 Chr.6: 118816515-118816515 on Build GRCh38
rs368975394 Chr.6: 118816272-118816272 on Build GRCh38
rs529929907 Chr.6: 118816483-118816483 on Build GRCh38
rs771346865 Chr.6: 118816254-118816254 on Build GRCh38
rs1054026612 Chr.6: 118816252-118816252 on Build GRCh38
rs11961865 Chr.6: 118816493-118816493 on Build GRCh38
rs145511219 Chr.6: 118816467-118816467 on Build GRCh38
rs970571824 Chr.6: 118816471-118816471 on Build GRCh38
rs768933770 Chr.6: 118816119-118816119 on Build GRCh38
rs188095623 Chr.6: 118816548-118816548 on Build GRCh38
rs190219997 Chr.6: 118816132-118816132 on Build GRCh38
rs759190028 Chr.6: 118816275-118816275 on Build GRCh38
rs909151147 Chr.6: 118816122-118816122 on Build GRCh38
rs981816142 Chr.6: 118816486-118816486 on Build GRCh38
rs941856952 Chr.6: 118816123-118816123 on Build GRCh38
rs916231662 Chr.6: 118816410-118816410 on Build GRCh38
rs990761833 Chr.6: 118816454-118816454 on Build GRCh38
rs77045701 Chr.6: 118816321-118816321 on Build GRCh38
rs887168095 Chr.6: 118816555-118816555 on Build GRCh38
rs564175402 Chr.6: 118816163-118816163 on Build GRCh38
rs942912425 Chr.6: 118816536-118816536 on Build GRCh38
rs192847060 Chr.6: 118816102-118816102 on Build GRCh38
rs78791427 Chr.6: 118816282-118816282 on Build GRCh38
rs184468109 Chr.6: 118816445-118816445 on Build GRCh38
rs200423526 Chr.6: 118816125-118816125 on Build GRCh38
rs762459633 Chr.6: 118816484-118816484 on Build GRCh38
rs759280235 Chr.6: 118816259-118816259 on Build GRCh38
rs923750811 Chr.6: 118816362-118816362 on Build GRCh38
rs369668984 Chr.6: 118816363-118816363 on Build GRCh38
rs1005590070 Chr.6: 118816231-118816231 on Build GRCh38
rs763498265 Chr.6: 118816196-118816196 on Build GRCh38
rs185696063 Chr.6: 118816112-118816112 on Build GRCh38
rs902182209 Chr.6: 118816211-118816211 on Build GRCh38
rs951399830 Chr.6: 118816106-118816106 on Build GRCh38
rs1047161806 Chr.6: 118816554-118816554 on Build GRCh38
rs763995379 Chr.6: 118816499-118816499 on Build GRCh38
rs928145098 Chr.6: 118816473-118816473 on Build GRCh38
rs17080673 Chr.6: 118816391-118816391 on Build GRCh38
rs989192693 Chr.6: 118816392-118816392 on Build GRCh38
rs575196712 Chr.6: 118816559-118816559 on Build GRCh38
rs533227061 Chr.6: 118816228-118816228 on Build GRCh38
rs888572781 Chr.6: 118816527-118816530 on Build GRCh38
rs17080674 Chr.6: 118816495-118816495 on Build GRCh38
rs35386524 Chr.6: 118816557-118816557 on Build GRCh38
rs781436076 Chr.6: 118816131-118816131 on Build GRCh38
rs954493053 Chr.6: 118816350-118816350 on Build GRCh38
rs1046135510 Chr.6: 118816197-118816197 on Build GRCh38
rs994227365 Chr.6: 118816124-118816124 on Build GRCh38
rs763780278 Chr.6: 118816542-118816542 on Build GRCh38
rs193242732 Chr.6: 118816166-118816166 on Build GRCh38
rs935030275 Chr.6: 118816215-118816215 on Build GRCh38
rs963598394 Chr.6: 118816453-118816453 on Build GRCh38
rs775152084 Chr.6: 118816348-118816348 on Build GRCh38
rs1005991078 Chr.6: 118816402-118816402 on Build GRCh38
rs529304688 Chr.6: 118816202-118816202 on Build GRCh38
rs998268482 Chr.6: 118816335-118816335 on Build GRCh38
rs778828006 Chr.6: 118816103-118816103 on Build GRCh38
rs7453731 Chr.6: 118816407-118816407 on Build GRCh38
rs562476412 Chr.6: 118816187-118816187 on Build GRCh38
rs140415491 Chr.6: 118816351-118816351 on Build GRCh38
rs1007159742 Chr.6: 118816262-118816262 on Build GRCh38

View Full Product Details