Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154571862-154572399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
UniGene
Hs.2306
Gene Chromosome Location
Chr.1: 154567781-154580026 on Build GRCh38
Transcripts
NM_000748.2 XM_017000180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022831 c.1137C>A p.R379R Chr.1: 154571960-154571960 on Build GRCh38
4022830 c.1116C>T p.G372G Chr.1: 154571939-154571939 on Build GRCh38
4140850 c.1151C>T p.A384V Chr.1: 154571974-154571974 on Build GRCh38
3385218 c.1096C>T p.R366C Chr.1: 154571919-154571919 on Build GRCh38
6121523 c.1049A>T p.Q350L Chr.1: 154571872-154571872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770055798 Chr.1: 154571994-154571994 on Build GRCh38
rs762237464 Chr.1: 154571891-154571891 on Build GRCh38
rs746495548 Chr.1: 154571975-154571975 on Build GRCh38
rs201462348 Chr.1: 154572161-154572161 on Build GRCh38
rs758068217 Chr.1: 154571965-154571965 on Build GRCh38
rs1029013963 Chr.1: 154572243-154572243 on Build GRCh38
rs201793956 Chr.1: 154571884-154571884 on Build GRCh38
rs771273497 Chr.1: 154572139-154572139 on Build GRCh38
rs547569557 Chr.1: 154572302-154572302 on Build GRCh38
rs951894685 Chr.1: 154572362-154572362 on Build GRCh38
rs893344333 Chr.1: 154572262-154572262 on Build GRCh38
rs56146528 Chr.1: 154572186-154572186 on Build GRCh38
rs990542717 Chr.1: 154572133-154572133 on Build GRCh38
rs200910048 Chr.1: 154572137-154572137 on Build GRCh38
rs930562574 Chr.1: 154571912-154571912 on Build GRCh38
rs1020953003 Chr.1: 154572022-154572022 on Build GRCh38
rs1020120934 Chr.1: 154572023-154572023 on Build GRCh38
rs55857552 Chr.1: 154572056-154572056 on Build GRCh38
rs747248135 Chr.1: 154572118-154572118 on Build GRCh38
rs1019064351 Chr.1: 154571958-154571958 on Build GRCh38
rs901932970 Chr.1: 154571967-154571967 on Build GRCh38
rs982967837 Chr.1: 154571925-154571925 on Build GRCh38
rs55685423 Chr.1: 154572014-154572014 on Build GRCh38
rs202114705 Chr.1: 154571926-154571926 on Build GRCh38
rs987019930 Chr.1: 154572349-154572349 on Build GRCh38
rs4845378 Chr.1: 154572175-154572175 on Build GRCh38
rs113116986 Chr.1: 154571951-154571951 on Build GRCh38
rs542041934 Chr.1: 154571980-154571980 on Build GRCh38
rs960350051 Chr.1: 154572346-154572346 on Build GRCh38
rs1049048240 Chr.1: 154572143-154572143 on Build GRCh38
rs113488072 Chr.1: 154571956-154571956 on Build GRCh38
rs199743038 Chr.1: 154572080-154572080 on Build GRCh38
rs1011691290 Chr.1: 154572274-154572274 on Build GRCh38
rs199928622 Chr.1: 154571907-154571907 on Build GRCh38
rs1015742850 Chr.1: 154572260-154572260 on Build GRCh38
rs1032924289 Chr.1: 154572182-154572182 on Build GRCh38
rs982906227 Chr.1: 154572077-154572077 on Build GRCh38
rs112744508 Chr.1: 154571957-154571957 on Build GRCh38
rs796052330 Chr.1: 154571955-154571957 on Build GRCh38
rs903039235 Chr.1: 154572171-154572171 on Build GRCh38
rs143990513 Chr.1: 154572309-154572309 on Build GRCh38
rs777049407 Chr.1: 154572150-154572150 on Build GRCh38
rs767533378 Chr.1: 154572059-154572059 on Build GRCh38
rs753731408 Chr.1: 154571937-154571937 on Build GRCh38
rs35834206 Chr.1: 154572013-154572014 on Build GRCh38
rs372757638 Chr.1: 154572062-154572062 on Build GRCh38
rs1040780008 Chr.1: 154572185-154572185 on Build GRCh38
rs981692284 Chr.1: 154572313-154572313 on Build GRCh38
rs796052331 Chr.1: 154571994-154572003 on Build GRCh38
rs763983504 Chr.1: 154571933-154571933 on Build GRCh38
rs912580588 Chr.1: 154572372-154572372 on Build GRCh38
rs1023981720 Chr.1: 154572215-154572215 on Build GRCh38
rs751625142 Chr.1: 154571914-154571914 on Build GRCh38
rs748619028 Chr.1: 154571983-154571983 on Build GRCh38
rs560427247 Chr.1: 154571999-154571999 on Build GRCh38
rs535787176 Chr.1: 154572228-154572228 on Build GRCh38
rs757023317 Chr.1: 154571938-154571938 on Build GRCh38
rs778714852 Chr.1: 154571940-154571940 on Build GRCh38
rs763142542 Chr.1: 154572055-154572055 on Build GRCh38
rs77710036 Chr.1: 154572096-154572096 on Build GRCh38
rs902984484 Chr.1: 154572024-154572024 on Build GRCh38
rs774449832 Chr.1: 154572057-154572057 on Build GRCh38
rs971173003 Chr.1: 154572067-154572067 on Build GRCh38
rs200924076 Chr.1: 154571881-154571881 on Build GRCh38
rs777185375 Chr.1: 154571885-154571885 on Build GRCh38
rs1038602573 Chr.1: 154571960-154571960 on Build GRCh38
rs781654245 Chr.1: 154571899-154571900 on Build GRCh38
rs1024048381 Chr.1: 154572063-154572063 on Build GRCh38
rs974346112 Chr.1: 154572081-154572081 on Build GRCh38
rs996179072 Chr.1: 154571995-154571995 on Build GRCh38
rs199658508 Chr.1: 154572027-154572027 on Build GRCh38
rs773331007 Chr.1: 154571897-154571897 on Build GRCh38
rs200441832 Chr.1: 154572013-154572013 on Build GRCh38
rs942920445 Chr.1: 154572037-154572048 on Build GRCh38
rs76126473 Chr.1: 154572089-154572089 on Build GRCh38
rs201040879 Chr.1: 154571910-154571910 on Build GRCh38
rs990593584 Chr.1: 154572348-154572348 on Build GRCh38
rs112585933 Chr.1: 154572058-154572058 on Build GRCh38
rs200483865 Chr.1: 154571932-154571932 on Build GRCh38
rs886348487 Chr.1: 154572164-154572164 on Build GRCh38
rs908723251 Chr.1: 154572079-154572079 on Build GRCh38
rs1035113615 Chr.1: 154572323-154572323 on Build GRCh38
rs916332689 Chr.1: 154572361-154572361 on Build GRCh38
rs201986342 Chr.1: 154571969-154571969 on Build GRCh38
rs932622161 Chr.1: 154571976-154571976 on Build GRCh38
rs527768017 Chr.1: 154572001-154572001 on Build GRCh38
rs996244767 Chr.1: 154572201-154572201 on Build GRCh38
rs753784853 Chr.1: 154572122-154572122 on Build GRCh38
rs779771980 Chr.1: 154571972-154571972 on Build GRCh38
rs760653937 Chr.1: 154571917-154571917 on Build GRCh38
rs945492622 Chr.1: 154572138-154572138 on Build GRCh38
rs770156397 Chr.1: 154571896-154571896 on Build GRCh38
rs916448473 Chr.1: 154572135-154572135 on Build GRCh38

View Full Product Details