Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154577653-154578155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
UniGene
Hs.2306
Gene Chromosome Location
Chr.1: 154567781-154580026 on Build GRCh38
Transcripts
NM_000748.2 XM_017000180.1

SNP Detail

SNP ID Chromosome Location
rs982431783 Chr.1: 154577751-154577751 on Build GRCh38
rs201569830 Chr.1: 154577953-154577953 on Build GRCh38
rs200757123 Chr.1: 154577934-154577934 on Build GRCh38
rs983653795 Chr.1: 154578033-154578033 on Build GRCh38
rs887104385 Chr.1: 154577798-154577798 on Build GRCh38
rs199694457 Chr.1: 154577866-154577866 on Build GRCh38
rs908188405 Chr.1: 154577832-154577832 on Build GRCh38
rs920850105 Chr.1: 154577725-154577725 on Build GRCh38
rs967008538 Chr.1: 154577912-154577912 on Build GRCh38
rs951046762 Chr.1: 154578020-154578020 on Build GRCh38
rs200468468 Chr.1: 154577829-154577829 on Build GRCh38
rs200050160 Chr.1: 154577971-154577971 on Build GRCh38
rs867723116 Chr.1: 154577880-154577880 on Build GRCh38
rs201303323 Chr.1: 154578008-154578008 on Build GRCh38
rs954450305 Chr.1: 154577892-154577892 on Build GRCh38
rs565202069 Chr.1: 154577941-154577941 on Build GRCh38
rs201431543 Chr.1: 154578079-154578079 on Build GRCh38
rs200351372 Chr.1: 154578063-154578063 on Build GRCh38
rs950122279 Chr.1: 154577783-154577783 on Build GRCh38
rs760738801 Chr.1: 154577995-154577995 on Build GRCh38
rs199960573 Chr.1: 154577695-154577695 on Build GRCh38
rs200469143 Chr.1: 154577748-154577748 on Build GRCh38
rs199748893 Chr.1: 154577885-154577885 on Build GRCh38
rs926231544 Chr.1: 154577732-154577732 on Build GRCh38
rs151180783 Chr.1: 154577897-154577897 on Build GRCh38
rs865900398 Chr.1: 154577720-154577720 on Build GRCh38
rs959229019 Chr.1: 154577950-154577950 on Build GRCh38
rs369265710 Chr.1: 154577681-154577681 on Build GRCh38
rs545656402 Chr.1: 154577686-154577686 on Build GRCh38
rs548589333 Chr.1: 154578078-154578078 on Build GRCh38
rs200738385 Chr.1: 154577697-154577697 on Build GRCh38
rs201698776 Chr.1: 154577913-154577913 on Build GRCh38
rs78614862 Chr.1: 154577694-154577694 on Build GRCh38
rs1008716767 Chr.1: 154578132-154578132 on Build GRCh38
rs908241478 Chr.1: 154578034-154578034 on Build GRCh38
rs569372918 Chr.1: 154578044-154578044 on Build GRCh38
rs200152526 Chr.1: 154577994-154577994 on Build GRCh38
rs1008603696 Chr.1: 154577894-154577894 on Build GRCh38
rs200037268 Chr.1: 154577937-154577937 on Build GRCh38
rs202143953 Chr.1: 154577738-154577738 on Build GRCh38
rs950238341 Chr.1: 154577979-154577979 on Build GRCh38
rs202032765 Chr.1: 154578014-154578014 on Build GRCh38
rs899704374 Chr.1: 154577864-154577864 on Build GRCh38
rs550799390 Chr.1: 154578041-154578041 on Build GRCh38
rs560589414 Chr.1: 154577774-154577774 on Build GRCh38
rs76242649 Chr.1: 154577708-154577708 on Build GRCh38
rs201040578 Chr.1: 154577987-154577987 on Build GRCh38
rs978424847 Chr.1: 154577927-154577927 on Build GRCh38
rs202151842 Chr.1: 154578053-154578053 on Build GRCh38
rs202148264 Chr.1: 154577792-154577792 on Build GRCh38
rs899101815 Chr.1: 154577863-154577863 on Build GRCh38
rs142105069 Chr.1: 154577871-154577871 on Build GRCh38
rs201862599 Chr.1: 154577988-154577988 on Build GRCh38
rs199604285 Chr.1: 154577929-154577929 on Build GRCh38
rs200846386 Chr.1: 154578029-154578029 on Build GRCh38
rs201452114 Chr.1: 154577840-154577840 on Build GRCh38
rs1050230876 Chr.1: 154578081-154578081 on Build GRCh38
rs201424237 Chr.1: 154577876-154577876 on Build GRCh38
rs1005591149 Chr.1: 154577811-154577811 on Build GRCh38
rs201069070 Chr.1: 154577784-154577784 on Build GRCh38
rs917409903 Chr.1: 154577976-154577977 on Build GRCh38
rs200264450 Chr.1: 154578015-154578015 on Build GRCh38
rs1040752848 Chr.1: 154577855-154577855 on Build GRCh38
rs996503836 Chr.1: 154578079-154578080 on Build GRCh38
rs895874423 Chr.1: 154577755-154577755 on Build GRCh38
rs146345695 Chr.1: 154578043-154578044 on Build GRCh38
rs532168074 Chr.1: 154578036-154578036 on Build GRCh38
rs199543295 Chr.1: 154578127-154578127 on Build GRCh38
rs530260006 Chr.1: 154578077-154578077 on Build GRCh38
rs937509552 Chr.1: 154577735-154577735 on Build GRCh38
rs943819688 Chr.1: 154577845-154577845 on Build GRCh38

View Full Product Details