Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145886096-145886598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100507557
Entrez Gene ID
100507557
Gene Name
uncharacterized LOC100507557
Gene Aliases
-
UniGene
Hs.124537
Gene Chromosome Location
Chr.6: 145734869-145886585 on Build GRCh38
Transcripts
NR_038244.1 NR_038245.1 NR_038246.1 NM_001042683.2 NM_173082.3 XM_006715439.3 XM_006715441.3 XM_006715443.3 XM_011535719.2 XM_017010691.1 XM_017010692.1 XM_017010693.1 XM_017010694.1 XM_017010695.1 XM_017010696.1

Gene Symbol
SHPRH
Entrez Gene ID
257218
Gene Name
SNF2 histone linker PHD RING helicase
Gene Aliases
bA545I5.2
UniGene
Hs.723297
Gene Chromosome Location
Chr.6: 145857838-145964436 on Build GRCh38
Transcripts
NR_038244.1 NR_038245.1 NR_038246.1 NM_001042683.2 NM_173082.3 XM_006715439.3 XM_006715441.3 XM_006715443.3 XM_011535719.2 XM_017010691.1 XM_017010692.1 XM_017010693.1 XM_017010694.1 XM_017010695.1 XM_017010696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6777973 c.4968-1G>T p.? Chr.6: 145886441-145886441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903956684 Chr.6: 145886451-145886451 on Build GRCh38
rs1002260233 Chr.6: 145886496-145886496 on Build GRCh38
rs11421576 Chr.6: 145886366-145886367 on Build GRCh38
rs386408856 Chr.6: 145886367-145886368 on Build GRCh38
rs769057470 Chr.6: 145886244-145886244 on Build GRCh38
rs778657069 Chr.6: 145886440-145886440 on Build GRCh38
rs543975858 Chr.6: 145886155-145886155 on Build GRCh38
rs901751634 Chr.6: 145886209-145886209 on Build GRCh38
rs147357063 Chr.6: 145886549-145886549 on Build GRCh38
rs961012628 Chr.6: 145886527-145886527 on Build GRCh38
rs1062068 Chr.6: 145886329-145886329 on Build GRCh38
rs527296988 Chr.6: 145886410-145886410 on Build GRCh38
rs142690339 Chr.6: 145886530-145886530 on Build GRCh38
rs969860862 Chr.6: 145886401-145886401 on Build GRCh38
rs745679086 Chr.6: 145886462-145886462 on Build GRCh38
rs140428826 Chr.6: 145886176-145886176 on Build GRCh38
rs753780502 Chr.6: 145886426-145886426 on Build GRCh38
rs929168606 Chr.6: 145886218-145886218 on Build GRCh38
rs2256998 Chr.6: 145886521-145886521 on Build GRCh38
rs1002353500 Chr.6: 145886423-145886423 on Build GRCh38
rs113686932 Chr.6: 145886550-145886550 on Build GRCh38
rs779986457 Chr.6: 145886451-145886452 on Build GRCh38
rs375293591 Chr.6: 145886369-145886370 on Build GRCh38
rs758226661 Chr.6: 145886483-145886483 on Build GRCh38
rs1062067 Chr.6: 145886427-145886427 on Build GRCh38
rs1038691837 Chr.6: 145886330-145886330 on Build GRCh38
rs797006839 Chr.6: 145886370-145886371 on Build GRCh38
rs910677328 Chr.6: 145886161-145886161 on Build GRCh38
rs988357760 Chr.6: 145886460-145886460 on Build GRCh38
rs752876818 Chr.6: 145886386-145886386 on Build GRCh38
rs1043877540 Chr.6: 145886446-145886446 on Build GRCh38
rs887408660 Chr.6: 145886281-145886281 on Build GRCh38
rs200617992 Chr.6: 145886456-145886456 on Build GRCh38
rs532656034 Chr.6: 145886216-145886216 on Build GRCh38
rs35420276 Chr.6: 145886370-145886371 on Build GRCh38
rs869285509 Chr.6: 145886371-145886372 on Build GRCh38
rs183496929 Chr.6: 145886372-145886372 on Build GRCh38
rs992512016 Chr.6: 145886174-145886174 on Build GRCh38
rs1040214906 Chr.6: 145886183-145886183 on Build GRCh38
rs756227410 Chr.6: 145886391-145886391 on Build GRCh38
rs908970572 Chr.6: 145886307-145886307 on Build GRCh38
rs779888434 Chr.6: 145886484-145886484 on Build GRCh38
rs984579550 Chr.6: 145886226-145886226 on Build GRCh38
rs1044161779 Chr.6: 145886146-145886146 on Build GRCh38

View Full Product Details