Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158999725-159000219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH3
Entrez Gene ID
83861
Gene Name
radial spoke 3 homolog
Gene Aliases
CILD32, RSHL2, RSP3, dJ111C20.1
UniGene
Hs.154628
Gene Chromosome Location
Chr.6: 158947012-159000432 on Build GRCh38
Transcripts
NM_031924.4 XM_005267153.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804550 c.11A>C p.K4T Chr.6: 158999966-158999966 on Build GRCh38
4160334 c.207C>A p.P69P Chr.6: 158999770-158999770 on Build GRCh38
1754627 c.235G>C p.E79Q Chr.6: 158999742-158999742 on Build GRCh38
7186060 c.124C>A p.P42T Chr.6: 158999853-158999853 on Build GRCh38
3622405 c.137G>A p.G46E Chr.6: 158999840-158999840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777464382 Chr.6: 158999773-158999773 on Build GRCh38
rs975420141 Chr.6: 158999984-158999984 on Build GRCh38
rs772669258 Chr.6: 158999948-158999948 on Build GRCh38
rs542739531 Chr.6: 158999916-158999916 on Build GRCh38
rs1008666489 Chr.6: 159000138-159000138 on Build GRCh38
rs775284185 Chr.6: 158999967-158999967 on Build GRCh38
rs753412130 Chr.6: 158999769-158999769 on Build GRCh38
rs757160822 Chr.6: 158999915-158999915 on Build GRCh38
rs528976843 Chr.6: 159000018-159000018 on Build GRCh38
rs140715430 Chr.6: 158999934-158999934 on Build GRCh38
rs56278154 Chr.6: 159000151-159000151 on Build GRCh38
rs769171820 Chr.6: 159000051-159000051 on Build GRCh38
rs867910746 Chr.6: 158999918-158999918 on Build GRCh38
rs549358371 Chr.6: 158999799-158999799 on Build GRCh38
rs759294948 Chr.6: 158999900-158999900 on Build GRCh38
rs199715125 Chr.6: 158999748-158999748 on Build GRCh38
rs889809628 Chr.6: 159000124-159000124 on Build GRCh38
rs545216136 Chr.6: 158999965-158999965 on Build GRCh38
rs537394123 Chr.6: 159000118-159000118 on Build GRCh38
rs751551909 Chr.6: 158999909-158999909 on Build GRCh38
rs776041722 Chr.6: 158999763-158999763 on Build GRCh38
rs1019379817 Chr.6: 158999925-158999925 on Build GRCh38
rs1047331345 Chr.6: 159000013-159000013 on Build GRCh38
rs990850821 Chr.6: 158999981-158999981 on Build GRCh38
rs762178749 Chr.6: 158999818-158999818 on Build GRCh38
rs148593164 Chr.6: 158999834-158999834 on Build GRCh38
rs751193355 Chr.6: 158999809-158999810 on Build GRCh38
rs769897799 Chr.6: 158999800-158999800 on Build GRCh38
rs537001972 Chr.6: 158999889-158999889 on Build GRCh38
rs147256615 Chr.6: 158999933-158999933 on Build GRCh38
rs868234226 Chr.6: 158999828-158999828 on Build GRCh38
rs138878442 Chr.6: 158999751-158999751 on Build GRCh38
rs762486110 Chr.6: 158999971-158999971 on Build GRCh38
rs546435437 Chr.6: 158999871-158999871 on Build GRCh38
rs767979912 Chr.6: 158999823-158999823 on Build GRCh38
rs746082927 Chr.6: 158999795-158999795 on Build GRCh38
rs368216318 Chr.6: 158999764-158999764 on Build GRCh38
rs1032137415 Chr.6: 159000159-159000159 on Build GRCh38
rs898806447 Chr.6: 158999878-158999878 on Build GRCh38
rs1054831509 Chr.6: 158999788-158999788 on Build GRCh38
rs778288389 Chr.6: 158999954-158999954 on Build GRCh38
rs764036988 Chr.6: 158999842-158999842 on Build GRCh38
rs751442066 Chr.6: 158999854-158999854 on Build GRCh38
rs538126852 Chr.6: 159000062-159000062 on Build GRCh38
rs774581216 Chr.6: 158999814-158999814 on Build GRCh38
rs898573832 Chr.6: 159000064-159000064 on Build GRCh38
rs377436847 Chr.6: 158999863-158999863 on Build GRCh38
rs375403935 Chr.6: 158999976-158999976 on Build GRCh38
rs371339284 Chr.6: 158999746-158999746 on Build GRCh38
rs966631368 Chr.6: 159000143-159000143 on Build GRCh38
rs773663592 Chr.6: 158999898-158999898 on Build GRCh38
rs964002768 Chr.6: 158999980-158999980 on Build GRCh38
rs756900997 Chr.6: 158999976-158999983 on Build GRCh38
rs757114046 Chr.6: 158999857-158999857 on Build GRCh38
rs758989581 Chr.6: 159000015-159000015 on Build GRCh38
rs754969122 Chr.6: 158999745-158999745 on Build GRCh38
rs1003857567 Chr.6: 159000085-159000085 on Build GRCh38
rs747218607 Chr.6: 158999752-158999752 on Build GRCh38
rs748095203 Chr.6: 158999747-158999747 on Build GRCh38
rs571444929 Chr.6: 159000183-159000184 on Build GRCh38
rs202214576 Chr.6: 158999902-158999902 on Build GRCh38
rs41267757 Chr.6: 158999770-158999770 on Build GRCh38
rs369484408 Chr.6: 158999804-158999804 on Build GRCh38
rs1022356477 Chr.6: 159000155-159000155 on Build GRCh38
rs749778647 Chr.6: 158999806-158999806 on Build GRCh38
rs549135750 Chr.6: 159000040-159000040 on Build GRCh38
rs370030962 Chr.6: 159000029-159000029 on Build GRCh38
rs1004276798 Chr.6: 158999859-158999859 on Build GRCh38
rs565392186 Chr.6: 158999972-158999972 on Build GRCh38
rs759241960 Chr.6: 158999825-158999825 on Build GRCh38
rs779081089 Chr.6: 159000010-159000010 on Build GRCh38
rs763294982 Chr.6: 159000014-159000014 on Build GRCh38
rs756954187 Chr.6: 158999838-158999838 on Build GRCh38
rs771147763 Chr.6: 158999755-158999755 on Build GRCh38
rs987037289 Chr.6: 158999765-158999765 on Build GRCh38
rs750176051 Chr.6: 158999923-158999923 on Build GRCh38
rs916255546 Chr.6: 158999996-158999996 on Build GRCh38
rs907382630 Chr.6: 158999850-158999850 on Build GRCh38
rs45484795 Chr.6: 159000008-159000008 on Build GRCh38
rs557335543 Chr.6: 158999893-158999893 on Build GRCh38
rs760519029 Chr.6: 158999999-158999999 on Build GRCh38
rs370487181 Chr.6: 158999782-158999782 on Build GRCh38
rs879825037 Chr.6: 159000178-159000178 on Build GRCh38
rs769854699 Chr.6: 158999762-158999762 on Build GRCh38
rs754166423 Chr.6: 158999936-158999936 on Build GRCh38
rs200029382 Chr.6: 158999824-158999824 on Build GRCh38
rs757935663 Chr.6: 158999754-158999755 on Build GRCh38
rs565912272 Chr.6: 158999768-158999768 on Build GRCh38
rs748598085 Chr.6: 158999897-158999897 on Build GRCh38
rs143832203 Chr.6: 158999906-158999906 on Build GRCh38
rs778661797 Chr.6: 158999924-158999924 on Build GRCh38
rs759557879 Chr.6: 158999758-158999758 on Build GRCh38
rs748686781 Chr.6: 158999947-158999947 on Build GRCh38
rs377747951 Chr.6: 158999977-158999977 on Build GRCh38
rs766082269 Chr.6: 159000007-159000007 on Build GRCh38
rs376047207 Chr.6: 158999956-158999956 on Build GRCh38
rs551558237 Chr.6: 159000065-159000065 on Build GRCh38
rs762587091 Chr.6: 158999904-158999904 on Build GRCh38
rs776701419 Chr.6: 158999757-158999757 on Build GRCh38
rs761767781 Chr.6: 158999986-158999986 on Build GRCh38
rs990597895 Chr.6: 159000185-159000185 on Build GRCh38
rs752628369 Chr.6: 158999772-158999772 on Build GRCh38
rs371925841 Chr.6: 158999774-158999774 on Build GRCh38
rs749457687 Chr.6: 158999926-158999926 on Build GRCh38
rs201139812 Chr.6: 158999767-158999767 on Build GRCh38
rs201220807 Chr.6: 158999853-158999853 on Build GRCh38
rs371164829 Chr.6: 158999901-158999901 on Build GRCh38
rs144832123 Chr.6: 158999775-158999775 on Build GRCh38
rs750031667 Chr.6: 158999989-158999989 on Build GRCh38
rs571413579 Chr.6: 159000092-159000092 on Build GRCh38
rs769165749 Chr.6: 158999810-158999810 on Build GRCh38
rs780026125 Chr.6: 158999866-158999866 on Build GRCh38
rs866399970 Chr.6: 159000061-159000061 on Build GRCh38
rs916209081 Chr.6: 159000195-159000195 on Build GRCh38
rs185887960 Chr.6: 159000043-159000043 on Build GRCh38
rs1019474400 Chr.6: 159000142-159000142 on Build GRCh38
rs1028646396 Chr.6: 158999750-158999750 on Build GRCh38
rs747323201 Chr.6: 158999964-158999964 on Build GRCh38
rs372470923 Chr.6: 158999860-158999860 on Build GRCh38
rs954060091 Chr.6: 158999922-158999922 on Build GRCh38
rs1011886033 Chr.6: 158999883-158999883 on Build GRCh38
rs563341320 Chr.6: 159000016-159000016 on Build GRCh38
rs898921159 Chr.6: 159000045-159000045 on Build GRCh38
rs531805169 Chr.6: 158999983-158999983 on Build GRCh38
rs995608710 Chr.6: 159000079-159000079 on Build GRCh38
rs763687230 Chr.6: 158999753-158999754 on Build GRCh38

View Full Product Details