Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 165336031-165336471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDE10A
Entrez Gene ID
10846
Gene Name
phosphodiesterase 10A
Gene Aliases
ADSD2, HSPDE10A, IOLOD, LINC00473, PDE10A19
UniGene
Hs.348762
Gene Chromosome Location
Chr.6: 165327287-165988121 on Build GRCh38
Transcripts
NM_001130690.2 NM_006661.3 XM_006715321.3 XM_011535387.2 XM_011535388.2 XM_011535393.2 XM_017010194.1 XM_017010195.1 XM_017010196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4713021 c.2196C>T p.A732A Chr.6: 165336194-165336194 on Build GRCh38
7291168 c.2246A>G p.E749G Chr.6: 165336144-165336144 on Build GRCh38
6742338 c.2189C>A p.T730N Chr.6: 165336171-165336171 on Build GRCh38
7291169 c.2216A>G p.E739G Chr.6: 165336144-165336144 on Build GRCh38
7316218 c.2266A>T p.R756W Chr.6: 165336124-165336124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540559098 Chr.6: 165336193-165336193 on Build GRCh38
rs567788873 Chr.6: 165336395-165336395 on Build GRCh38
rs1005893495 Chr.6: 165336408-165336408 on Build GRCh38
rs773012316 Chr.6: 165336112-165336112 on Build GRCh38
rs549725764 Chr.6: 165336419-165336419 on Build GRCh38
rs958904091 Chr.6: 165336214-165336214 on Build GRCh38
rs370798686 Chr.6: 165336232-165336232 on Build GRCh38
rs545276418 Chr.6: 165336386-165336386 on Build GRCh38
rs578026067 Chr.6: 165336293-165336293 on Build GRCh38
rs146776051 Chr.6: 165336206-165336206 on Build GRCh38
rs929679443 Chr.6: 165336316-165336316 on Build GRCh38
rs749454326 Chr.6: 165336110-165336110 on Build GRCh38
rs752329136 Chr.6: 165336083-165336083 on Build GRCh38
rs764780388 Chr.6: 165336078-165336078 on Build GRCh38
rs945923792 Chr.6: 165336314-165336314 on Build GRCh38
rs73028216 Chr.6: 165336424-165336424 on Build GRCh38
rs986428205 Chr.6: 165336328-165336328 on Build GRCh38
rs918223871 Chr.6: 165336065-165336065 on Build GRCh38
rs749403215 Chr.6: 165336174-165336174 on Build GRCh38
rs756240026 Chr.6: 165336108-165336108 on Build GRCh38
rs773852367 Chr.6: 165336239-165336239 on Build GRCh38
rs917377023 Chr.6: 165336196-165336196 on Build GRCh38
rs35370607 Chr.6: 165336194-165336194 on Build GRCh38
rs547329058 Chr.6: 165336426-165336426 on Build GRCh38
rs764717397 Chr.6: 165336117-165336117 on Build GRCh38
rs373834600 Chr.6: 165336113-165336113 on Build GRCh38
rs375017027 Chr.6: 165336247-165336247 on Build GRCh38
rs768252127 Chr.6: 165336237-165336237 on Build GRCh38
rs781578747 Chr.6: 165336104-165336104 on Build GRCh38
rs377355629 Chr.6: 165336146-165336146 on Build GRCh38
rs756169867 Chr.6: 165336384-165336384 on Build GRCh38
rs781103938 Chr.6: 165336207-165336207 on Build GRCh38
rs776414331 Chr.6: 165336115-165336115 on Build GRCh38
rs774982484 Chr.6: 165336118-165336118 on Build GRCh38
rs774929597 Chr.6: 165336233-165336233 on Build GRCh38
rs942589151 Chr.6: 165336377-165336377 on Build GRCh38
rs764450783 Chr.6: 165336092-165336092 on Build GRCh38
rs750961282 Chr.6: 165336158-165336158 on Build GRCh38
rs368948802 Chr.6: 165336215-165336215 on Build GRCh38
rs759531574 Chr.6: 165336116-165336116 on Build GRCh38
rs780227304 Chr.6: 165336171-165336171 on Build GRCh38
rs200733566 Chr.6: 165336107-165336107 on Build GRCh38
rs779410560 Chr.6: 165336259-165336259 on Build GRCh38
rs980181590 Chr.6: 165336228-165336228 on Build GRCh38
rs901695883 Chr.6: 165336357-165336357 on Build GRCh38
rs762128501 Chr.6: 165336235-165336235 on Build GRCh38
rs1040277450 Chr.6: 165336338-165336338 on Build GRCh38
rs746923112 Chr.6: 165336243-165336243 on Build GRCh38
rs867236726 Chr.6: 165336163-165336163 on Build GRCh38
rs143718875 Chr.6: 165336155-165336155 on Build GRCh38
rs376748454 Chr.6: 165336091-165336091 on Build GRCh38
rs370397127 Chr.6: 165336160-165336160 on Build GRCh38
rs912482915 Chr.6: 165336345-165336345 on Build GRCh38
rs887491199 Chr.6: 165336397-165336397 on Build GRCh38
rs562660076 Chr.6: 165336289-165336289 on Build GRCh38
rs148845227 Chr.6: 165336176-165336176 on Build GRCh38
rs200503076 Chr.6: 165336234-165336234 on Build GRCh38
rs184966453 Chr.6: 165336418-165336418 on Build GRCh38
rs757732707 Chr.6: 165336102-165336102 on Build GRCh38
rs751909982 Chr.6: 165336147-165336147 on Build GRCh38
rs370502092 Chr.6: 165336101-165336101 on Build GRCh38
rs918329404 Chr.6: 165336270-165336278 on Build GRCh38
rs560696132 Chr.6: 165336393-165336393 on Build GRCh38
rs539276206 Chr.6: 165336151-165336151 on Build GRCh38

View Full Product Details