Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 16861995-16862532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGR3
Entrez Gene ID
155465
Gene Name
anterior gradient 3, protein disulphide isomerase family member
Gene Aliases
AG-3, AG3, BCMP11, HAG3, PDIA18, hAG-3
UniGene
Hs.100686
Gene Chromosome Location
Chr.7: 16854711-16881989 on Build GRCh38
Transcripts
NM_176813.4 XM_005249627.2 XM_011515152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4155684 c.262C>A p.Q88K Chr.7: 16862025-16862025 on Build GRCh38
1176784 c.234G>T p.K78N Chr.7: 16862053-16862053 on Build GRCh38
1182236 c.265G>T p.E89* Chr.7: 16862022-16862022 on Build GRCh38
5526423 c.272C>T p.A91V Chr.7: 16862015-16862015 on Build GRCh38
5526424 c.274C>T p.Q92* Chr.7: 16862013-16862013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187669149 Chr.7: 16862339-16862339 on Build GRCh38
rs1024203460 Chr.7: 16862420-16862420 on Build GRCh38
rs1023253626 Chr.7: 16862495-16862496 on Build GRCh38
rs140516173 Chr.7: 16862495-16862495 on Build GRCh38
rs74986534 Chr.7: 16862396-16862396 on Build GRCh38
rs374938037 Chr.7: 16862215-16862215 on Build GRCh38
rs1031000703 Chr.7: 16862447-16862447 on Build GRCh38
rs548409939 Chr.7: 16862346-16862346 on Build GRCh38
rs1050365398 Chr.7: 16862123-16862123 on Build GRCh38
rs758962226 Chr.7: 16862112-16862112 on Build GRCh38
rs1043677675 Chr.7: 16862023-16862023 on Build GRCh38
rs534343722 Chr.7: 16862468-16862468 on Build GRCh38
rs991362964 Chr.7: 16862220-16862220 on Build GRCh38
rs140779232 Chr.7: 16862061-16862061 on Build GRCh38
rs1043061926 Chr.7: 16862435-16862435 on Build GRCh38
rs374680356 Chr.7: 16862103-16862103 on Build GRCh38
rs1005909938 Chr.7: 16862164-16862164 on Build GRCh38
rs916853287 Chr.7: 16862225-16862225 on Build GRCh38
rs866061545 Chr.7: 16862325-16862325 on Build GRCh38
rs959819312 Chr.7: 16862478-16862478 on Build GRCh38
rs968658415 Chr.7: 16862325-16862325 on Build GRCh38
rs1007563394 Chr.7: 16862287-16862287 on Build GRCh38
rs752745210 Chr.7: 16862024-16862024 on Build GRCh38
rs1051759630 Chr.7: 16862405-16862405 on Build GRCh38
rs776385310 Chr.7: 16862111-16862111 on Build GRCh38
rs770684235 Chr.7: 16862110-16862110 on Build GRCh38
rs935399127 Chr.7: 16862081-16862081 on Build GRCh38
rs992604589 Chr.7: 16862148-16862148 on Build GRCh38
rs909537862 Chr.7: 16862506-16862506 on Build GRCh38
rs558250965 Chr.7: 16862411-16862411 on Build GRCh38
rs1040433740 Chr.7: 16862275-16862275 on Build GRCh38
rs923392440 Chr.7: 16862473-16862473 on Build GRCh38
rs942309729 Chr.7: 16862242-16862242 on Build GRCh38
rs961348878 Chr.7: 16862508-16862508 on Build GRCh38
rs909576542 Chr.7: 16862361-16862361 on Build GRCh38
rs758172888 Chr.7: 16862033-16862033 on Build GRCh38
rs1031608957 Chr.7: 16862470-16862470 on Build GRCh38
rs984300452 Chr.7: 16862505-16862505 on Build GRCh38
rs563599099 Chr.7: 16862193-16862193 on Build GRCh38
rs570858617 Chr.7: 16862290-16862290 on Build GRCh38
rs976021094 Chr.7: 16862461-16862461 on Build GRCh38
rs182314052 Chr.7: 16862494-16862494 on Build GRCh38
rs983796642 Chr.7: 16862355-16862355 on Build GRCh38
rs1002618575 Chr.7: 16862201-16862201 on Build GRCh38
rs959873388 Chr.7: 16862328-16862329 on Build GRCh38
rs554252804 Chr.7: 16862364-16862364 on Build GRCh38
rs961821215 Chr.7: 16862198-16862198 on Build GRCh38
rs1002502540 Chr.7: 16862347-16862347 on Build GRCh38
rs948354073 Chr.7: 16862349-16862349 on Build GRCh38
rs756871684 Chr.7: 16862068-16862068 on Build GRCh38
rs201972239 Chr.7: 16862042-16862042 on Build GRCh38
rs923928829 Chr.7: 16862264-16862264 on Build GRCh38
rs968297855 Chr.7: 16862440-16862440 on Build GRCh38
rs190807037 Chr.7: 16862445-16862445 on Build GRCh38
rs780349621 Chr.7: 16862063-16862063 on Build GRCh38
rs887655284 Chr.7: 16862154-16862154 on Build GRCh38
rs1014582835 Chr.7: 16862189-16862189 on Build GRCh38
rs915548999 Chr.7: 16862152-16862152 on Build GRCh38
rs935451499 Chr.7: 16862273-16862273 on Build GRCh38
rs946747463 Chr.7: 16862021-16862021 on Build GRCh38
rs73307290 Chr.7: 16862394-16862394 on Build GRCh38
rs777482636 Chr.7: 16862035-16862035 on Build GRCh38
rs898023171 Chr.7: 16862265-16862265 on Build GRCh38
rs60924576 Chr.7: 16862308-16862308 on Build GRCh38
rs968679442 Chr.7: 16862253-16862253 on Build GRCh38
rs552144946 Chr.7: 16862228-16862228 on Build GRCh38
rs979424050 Chr.7: 16862262-16862262 on Build GRCh38
rs373495381 Chr.7: 16862096-16862096 on Build GRCh38
rs568479603 Chr.7: 16862336-16862336 on Build GRCh38
rs923310498 Chr.7: 16862286-16862286 on Build GRCh38
rs956707200 Chr.7: 16862455-16862455 on Build GRCh38
rs773645353 Chr.7: 16862108-16862108 on Build GRCh38
rs531112903 Chr.7: 16862203-16862203 on Build GRCh38
rs969829872 Chr.7: 16862496-16862496 on Build GRCh38
rs74534736 Chr.7: 16862326-16862326 on Build GRCh38
rs931840802 Chr.7: 16862289-16862289 on Build GRCh38
rs932042180 Chr.7: 16862476-16862476 on Build GRCh38
rs942277703 Chr.7: 16862373-16862373 on Build GRCh38

View Full Product Details