Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23200016-23200513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUPL2
Entrez Gene ID
11097
Gene Name
nucleoporin like 2
Gene Aliases
CG1, NLP-1, NLP_1, hCG1
UniGene
Hs.408241
Gene Chromosome Location
Chr.7: 23181567-23201011 on Build GRCh38
Transcripts
NM_007342.2 XM_005249592.1 XM_005249593.3 XM_011515104.2 XM_017011714.1 XM_017011715.1 XM_017011716.1 XM_017011717.1 XM_017011718.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3880262 c.714C>T p.S238S Chr.7: 23200187-23200187 on Build GRCh38
121363 c.1032T>G p.G344G Chr.7: 23200505-23200505 on Build GRCh38
5772113 c.721G>A p.D241N Chr.7: 23200194-23200194 on Build GRCh38
3637511 c.955C>G p.L319V Chr.7: 23200428-23200428 on Build GRCh38
1088582 c.890C>T p.S297L Chr.7: 23200363-23200363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969724772 Chr.7: 23200392-23200392 on Build GRCh38
rs150180643 Chr.7: 23200236-23200236 on Build GRCh38
rs766463707 Chr.7: 23200249-23200249 on Build GRCh38
rs373233862 Chr.7: 23200331-23200331 on Build GRCh38
rs763992632 Chr.7: 23200405-23200405 on Build GRCh38
rs77508599 Chr.7: 23200205-23200205 on Build GRCh38
rs752973301 Chr.7: 23200364-23200364 on Build GRCh38
rs772427957 Chr.7: 23200234-23200234 on Build GRCh38
rs1023964230 Chr.7: 23200382-23200382 on Build GRCh38
rs760484857 Chr.7: 23200145-23200146 on Build GRCh38
rs746483408 Chr.7: 23200180-23200180 on Build GRCh38
rs777811000 Chr.7: 23200277-23200277 on Build GRCh38
rs751562765 Chr.7: 23200280-23200280 on Build GRCh38
rs760698606 Chr.7: 23200238-23200238 on Build GRCh38
rs757255600 Chr.7: 23200417-23200417 on Build GRCh38
rs758251492 Chr.7: 23200176-23200176 on Build GRCh38
rs1024016455 Chr.7: 23200090-23200090 on Build GRCh38
rs189374272 Chr.7: 23200104-23200104 on Build GRCh38
rs773653043 Chr.7: 23200350-23200350 on Build GRCh38
rs753518076 Chr.7: 23200449-23200449 on Build GRCh38
rs891401693 Chr.7: 23200418-23200418 on Build GRCh38
rs858236 Chr.7: 23200148-23200148 on Build GRCh38
rs755681615 Chr.7: 23200439-23200439 on Build GRCh38
rs774574630 Chr.7: 23200224-23200224 on Build GRCh38
rs747435974 Chr.7: 23200460-23200460 on Build GRCh38
rs375404356 Chr.7: 23200263-23200263 on Build GRCh38
rs80001203 Chr.7: 23200483-23200483 on Build GRCh38
rs138284235 Chr.7: 23200429-23200429 on Build GRCh38
rs777902850 Chr.7: 23200178-23200178 on Build GRCh38
rs756227242 Chr.7: 23200126-23200126 on Build GRCh38
rs762603996 Chr.7: 23200399-23200399 on Build GRCh38
rs766337366 Chr.7: 23200146-23200148 on Build GRCh38
rs778108978 Chr.7: 23200455-23200455 on Build GRCh38
rs755239237 Chr.7: 23200148-23200148 on Build GRCh38
rs752992654 Chr.7: 23200395-23200395 on Build GRCh38
rs772450262 Chr.7: 23200346-23200346 on Build GRCh38
rs780941512 Chr.7: 23200208-23200208 on Build GRCh38
rs572788556 Chr.7: 23200145-23200145 on Build GRCh38
rs796192488 Chr.7: 23200414-23200414 on Build GRCh38
rs779525178 Chr.7: 23200446-23200446 on Build GRCh38
rs771410154 Chr.7: 23200461-23200461 on Build GRCh38
rs768998260 Chr.7: 23200215-23200215 on Build GRCh38
rs765557668 Chr.7: 23200267-23200267 on Build GRCh38
rs969944423 Chr.7: 23200091-23200091 on Build GRCh38
rs769317648 Chr.7: 23200309-23200309 on Build GRCh38
rs140632442 Chr.7: 23200447-23200447 on Build GRCh38
rs1051046213 Chr.7: 23200154-23200154 on Build GRCh38
rs112626242 Chr.7: 23200147-23200147 on Build GRCh38
rs748045514 Chr.7: 23200170-23200170 on Build GRCh38
rs778579566 Chr.7: 23200157-23200157 on Build GRCh38
rs776565063 Chr.7: 23200364-23200364 on Build GRCh38
rs138813712 Chr.7: 23200373-23200373 on Build GRCh38
rs916304763 Chr.7: 23200304-23200304 on Build GRCh38
rs780851691 Chr.7: 23200297-23200297 on Build GRCh38
rs762288670 Chr.7: 23200226-23200226 on Build GRCh38
rs751473213 Chr.7: 23200412-23200412 on Build GRCh38
rs531702000 Chr.7: 23200139-23200139 on Build GRCh38
rs753995702 Chr.7: 23200250-23200250 on Build GRCh38
rs756945837 Chr.7: 23200380-23200380 on Build GRCh38
rs935548376 Chr.7: 23200389-23200389 on Build GRCh38
rs745450053 Chr.7: 23200299-23200299 on Build GRCh38
rs904070316 Chr.7: 23200040-23200040 on Build GRCh38
rs992485644 Chr.7: 23200065-23200065 on Build GRCh38
rs758249825 Chr.7: 23200276-23200276 on Build GRCh38
rs745892431 Chr.7: 23200472-23200472 on Build GRCh38
rs549312685 Chr.7: 23200122-23200122 on Build GRCh38
rs889810135 Chr.7: 23200179-23200179 on Build GRCh38
rs756767732 Chr.7: 23200291-23200291 on Build GRCh38
rs964116682 Chr.7: 23200141-23200141 on Build GRCh38
rs752409597 Chr.7: 23200272-23200272 on Build GRCh38
rs754747934 Chr.7: 23200151-23200151 on Build GRCh38
rs1031202329 Chr.7: 23200312-23200312 on Build GRCh38
rs762014071 Chr.7: 23200139-23200140 on Build GRCh38
rs201736904 Chr.7: 23200340-23200340 on Build GRCh38
rs547828229 Chr.7: 23200363-23200363 on Build GRCh38
rs745518385 Chr.7: 23200211-23200211 on Build GRCh38
rs749871373 Chr.7: 23200433-23200433 on Build GRCh38
rs374794638 Chr.7: 23200123-23200123 on Build GRCh38
rs759706407 Chr.7: 23200365-23200365 on Build GRCh38
rs779796981 Chr.7: 23200335-23200335 on Build GRCh38
rs200946375 Chr.7: 23200160-23200161 on Build GRCh38
rs369470475 Chr.7: 23200319-23200319 on Build GRCh38
rs531348706 Chr.7: 23200140-23200140 on Build GRCh38
rs777230992 Chr.7: 23200465-23200465 on Build GRCh38

View Full Product Details