Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157514458-157514955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCRL5
Entrez Gene ID
83416
Gene Name
Fc receptor like 5
Gene Aliases
BXMAS1, CD307, CD307e, FCRH5, IRTA2, PRO820
UniGene
Hs.415950
Gene Chromosome Location
Chr.1: 157513377-157552520 on Build GRCh38
Transcripts
NM_001195388.1 NM_031281.2

SNP Detail

SNP ID Chromosome Location
rs1035697465 Chr.1: 157514567-157514567 on Build GRCh38
rs969616506 Chr.1: 157514890-157514890 on Build GRCh38
rs142146846 Chr.1: 157514531-157514531 on Build GRCh38
rs1044887339 Chr.1: 157514762-157514762 on Build GRCh38
rs979752779 Chr.1: 157514616-157514616 on Build GRCh38
rs530256254 Chr.1: 157514709-157514709 on Build GRCh38
rs1014516926 Chr.1: 157514504-157514504 on Build GRCh38
rs781672601 Chr.1: 157514672-157514672 on Build GRCh38
rs570094344 Chr.1: 157514585-157514585 on Build GRCh38
rs1012041381 Chr.1: 157514595-157514595 on Build GRCh38
rs3900770 Chr.1: 157514908-157514908 on Build GRCh38
rs1036559889 Chr.1: 157514798-157514798 on Build GRCh38
rs754632425 Chr.1: 157514590-157514590 on Build GRCh38
rs571965033 Chr.1: 157514874-157514875 on Build GRCh38
rs533320063 Chr.1: 157514564-157514564 on Build GRCh38
rs74709991 Chr.1: 157514497-157514497 on Build GRCh38
rs528706489 Chr.1: 157514815-157514815 on Build GRCh38
rs1005843067 Chr.1: 157514503-157514503 on Build GRCh38
rs1018526701 Chr.1: 157514912-157514912 on Build GRCh38
rs3845586 Chr.1: 157514663-157514663 on Build GRCh38
rs184632504 Chr.1: 157514688-157514688 on Build GRCh38
rs554500430 Chr.1: 157514894-157514894 on Build GRCh38
rs955618163 Chr.1: 157514701-157514701 on Build GRCh38
rs542497005 Chr.1: 157514480-157514480 on Build GRCh38
rs890883508 Chr.1: 157514511-157514511 on Build GRCh38
rs1056434193 Chr.1: 157514751-157514751 on Build GRCh38
rs911318516 Chr.1: 157514554-157514554 on Build GRCh38
rs749504374 Chr.1: 157514905-157514905 on Build GRCh38
rs534230420 Chr.1: 157514724-157514724 on Build GRCh38
rs376985073 Chr.1: 157514546-157514546 on Build GRCh38
rs924032985 Chr.1: 157514745-157514745 on Build GRCh38
rs1043768871 Chr.1: 157514573-157514573 on Build GRCh38
rs549339492 Chr.1: 157514648-157514648 on Build GRCh38
rs3843307 Chr.1: 157514785-157514785 on Build GRCh38
rs1044835781 Chr.1: 157514621-157514621 on Build GRCh38
rs531031733 Chr.1: 157514637-157514637 on Build GRCh38
rs778164943 Chr.1: 157514634-157514634 on Build GRCh38
rs867500929 Chr.1: 157514549-157514549 on Build GRCh38
rs1027278320 Chr.1: 157514542-157514542 on Build GRCh38
rs1040291042 Chr.1: 157514518-157514518 on Build GRCh38
rs59495616 Chr.1: 157514670-157514670 on Build GRCh38
rs536320575 Chr.1: 157514793-157514793 on Build GRCh38
rs897564794 Chr.1: 157514848-157514848 on Build GRCh38
rs924919740 Chr.1: 157514493-157514493 on Build GRCh38
rs372671003 Chr.1: 157514591-157514591 on Build GRCh38
rs76700775 Chr.1: 157514722-157514722 on Build GRCh38
rs866636111 Chr.1: 157514823-157514823 on Build GRCh38
rs1023373966 Chr.1: 157514488-157514488 on Build GRCh38
rs947948255 Chr.1: 157514756-157514756 on Build GRCh38
rs1029959373 Chr.1: 157514902-157514902 on Build GRCh38
rs770053858 Chr.1: 157514816-157514816 on Build GRCh38
rs1030362636 Chr.1: 157514691-157514691 on Build GRCh38
rs969270104 Chr.1: 157514593-157514593 on Build GRCh38
rs917827634 Chr.1: 157514752-157514752 on Build GRCh38
rs781223535 Chr.1: 157514739-157514739 on Build GRCh38
rs58039254 Chr.1: 157514545-157514545 on Build GRCh38
rs935370816 Chr.1: 157514749-157514749 on Build GRCh38
rs116190888 Chr.1: 157514651-157514651 on Build GRCh38
rs996143929 Chr.1: 157514539-157514539 on Build GRCh38
rs887544206 Chr.1: 157514646-157514646 on Build GRCh38
rs899997497 Chr.1: 157514714-157514714 on Build GRCh38
rs1023086933 Chr.1: 157514845-157514845 on Build GRCh38
rs369131290 Chr.1: 157514586-157514586 on Build GRCh38
rs961662314 Chr.1: 157514512-157514512 on Build GRCh38
rs1023875074 Chr.1: 157514582-157514582 on Build GRCh38
rs58335866 Chr.1: 157514580-157514580 on Build GRCh38

View Full Product Details