Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30284216-30284709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105375218
Entrez Gene ID
105375218
Gene Name
uncharacterized LOC105375218
Gene Aliases
-
UniGene
Hs.566765
Gene Chromosome Location
Chr.7: 30247427-30286085 on Build GRCh38
Transcripts
NR_136267.1 NM_147128.3

Gene Symbol
ZNRF2
Entrez Gene ID
223082
Gene Name
zinc and ring finger 2, E3 ubiquitin protein ligase
Gene Aliases
RNF202
UniGene
Hs.263912
Gene Chromosome Location
Chr.7: 30284307-30367692 on Build GRCh38
Transcripts
NR_136267.1 NM_147128.3

SNP Detail

SNP ID Chromosome Location
rs1031776663 Chr.7: 30284450-30284450 on Build GRCh38
rs1032754551 Chr.7: 30284536-30284536 on Build GRCh38
rs983178451 Chr.7: 30284639-30284639 on Build GRCh38
rs571732451 Chr.7: 30284683-30284683 on Build GRCh38
rs898479080 Chr.7: 30284384-30284384 on Build GRCh38
rs895122981 Chr.7: 30284642-30284642 on Build GRCh38
rs372355877 Chr.7: 30284279-30284279 on Build GRCh38
rs765547915 Chr.7: 30284390-30284390 on Build GRCh38
rs1031022602 Chr.7: 30284676-30284676 on Build GRCh38
rs907214969 Chr.7: 30284591-30284591 on Build GRCh38
rs1014604478 Chr.7: 30284687-30284687 on Build GRCh38
rs374233886 Chr.7: 30284386-30284386 on Build GRCh38
rs891916318 Chr.7: 30284665-30284665 on Build GRCh38
rs990380026 Chr.7: 30284331-30284331 on Build GRCh38
rs575688079 Chr.7: 30284270-30284270 on Build GRCh38
rs1044363212 Chr.7: 30284565-30284565 on Build GRCh38
rs936965004 Chr.7: 30284688-30284688 on Build GRCh38
rs551171163 Chr.7: 30284410-30284410 on Build GRCh38
rs373972638 Chr.7: 30284592-30284592 on Build GRCh38
rs1003990359 Chr.7: 30284241-30284241 on Build GRCh38
rs991378267 Chr.7: 30284343-30284343 on Build GRCh38
rs1042495888 Chr.7: 30284627-30284627 on Build GRCh38
rs908466161 Chr.7: 30284603-30284603 on Build GRCh38
rs908437090 Chr.7: 30284427-30284427 on Build GRCh38
rs974383025 Chr.7: 30284661-30284661 on Build GRCh38
rs553651535 Chr.7: 30284538-30284538 on Build GRCh38
rs907249750 Chr.7: 30284424-30284424 on Build GRCh38
rs1038342230 Chr.7: 30284291-30284291 on Build GRCh38
rs966899954 Chr.7: 30284512-30284512 on Build GRCh38
rs938750510 Chr.7: 30284609-30284609 on Build GRCh38
rs1008425942 Chr.7: 30284297-30284297 on Build GRCh38
rs954552660 Chr.7: 30284636-30284636 on Build GRCh38
rs915741982 Chr.7: 30284345-30284345 on Build GRCh38
rs1043192656 Chr.7: 30284670-30284670 on Build GRCh38
rs973801779 Chr.7: 30284616-30284616 on Build GRCh38
rs1031807628 Chr.7: 30284304-30284304 on Build GRCh38
rs919625646 Chr.7: 30284666-30284666 on Build GRCh38
rs964731110 Chr.7: 30284483-30284483 on Build GRCh38
rs553500768 Chr.7: 30284653-30284653 on Build GRCh38
rs981244814 Chr.7: 30284596-30284596 on Build GRCh38
rs147515655 Chr.7: 30284293-30284293 on Build GRCh38
rs1021179893 Chr.7: 30284510-30284510 on Build GRCh38
rs879881583 Chr.7: 30284479-30284479 on Build GRCh38
rs964438000 Chr.7: 30284340-30284340 on Build GRCh38
rs545627962 Chr.7: 30284684-30284684 on Build GRCh38
rs1020939429 Chr.7: 30284300-30284300 on Build GRCh38
rs914240981 Chr.7: 30284256-30284256 on Build GRCh38
rs1038795526 Chr.7: 30284260-30284260 on Build GRCh38
rs1012957049 Chr.7: 30284399-30284399 on Build GRCh38
rs916534269 Chr.7: 30284524-30284524 on Build GRCh38
rs1001517241 Chr.7: 30284637-30284637 on Build GRCh38
rs954448638 Chr.7: 30284494-30284494 on Build GRCh38
rs914211367 Chr.7: 30284333-30284333 on Build GRCh38
rs533007688 Chr.7: 30284391-30284391 on Build GRCh38
rs1030421400 Chr.7: 30284488-30284488 on Build GRCh38
rs187801901 Chr.7: 30284620-30284620 on Build GRCh38
rs991300816 Chr.7: 30284541-30284541 on Build GRCh38
rs974881715 Chr.7: 30284347-30284347 on Build GRCh38
rs1011771543 Chr.7: 30284468-30284468 on Build GRCh38
rs1015430980 Chr.7: 30284657-30284657 on Build GRCh38
rs941895947 Chr.7: 30284382-30284382 on Build GRCh38
rs113486583 Chr.7: 30284466-30284466 on Build GRCh38
rs932326437 Chr.7: 30284671-30284671 on Build GRCh38
rs1019994699 Chr.7: 30284570-30284570 on Build GRCh38
rs979817830 Chr.7: 30284304-30284304 on Build GRCh38
rs915773175 Chr.7: 30284572-30284572 on Build GRCh38
rs1021856801 Chr.7: 30284472-30284472 on Build GRCh38
rs539151576 Chr.7: 30284689-30284689 on Build GRCh38
rs529109325 Chr.7: 30284350-30284350 on Build GRCh38
rs1049924505 Chr.7: 30284623-30284623 on Build GRCh38
rs150031568 Chr.7: 30284647-30284647 on Build GRCh38
rs1000876743 Chr.7: 30284517-30284517 on Build GRCh38
rs890851810 Chr.7: 30284624-30284624 on Build GRCh38
rs866389016 Chr.7: 30284612-30284612 on Build GRCh38
rs1011740501 Chr.7: 30284622-30284622 on Build GRCh38
rs961349757 Chr.7: 30284659-30284659 on Build GRCh38
rs997031194 Chr.7: 30284274-30284274 on Build GRCh38
rs981274396 Chr.7: 30284656-30284656 on Build GRCh38
rs530420086 Chr.7: 30284487-30284487 on Build GRCh38
rs563736742 Chr.7: 30284259-30284259 on Build GRCh38
rs947971826 Chr.7: 30284560-30284560 on Build GRCh38
rs1029977274 Chr.7: 30284629-30284629 on Build GRCh38
rs868092606 Chr.7: 30284254-30284254 on Build GRCh38
rs985061855 Chr.7: 30284672-30284672 on Build GRCh38
rs919575325 Chr.7: 30284618-30284618 on Build GRCh38
rs996996046 Chr.7: 30284481-30284481 on Build GRCh38
rs947483457 Chr.7: 30284668-30284668 on Build GRCh38
rs182360842 Chr.7: 30284358-30284358 on Build GRCh38
rs912315745 Chr.7: 30284674-30284674 on Build GRCh38
rs959518871 Chr.7: 30284537-30284537 on Build GRCh38
rs983219826 Chr.7: 30284497-30284497 on Build GRCh38
rs896073877 Chr.7: 30284278-30284278 on Build GRCh38
rs898245319 Chr.7: 30284475-30284475 on Build GRCh38
rs891281626 Chr.7: 30284499-30284499 on Build GRCh38
rs1008456966 Chr.7: 30284502-30284502 on Build GRCh38
rs999970221 Chr.7: 30284682-30284682 on Build GRCh38
rs902813008 Chr.7: 30284630-30284630 on Build GRCh38
rs996416461 Chr.7: 30284675-30284676 on Build GRCh38
rs966662198 Chr.7: 30284301-30284301 on Build GRCh38
rs1032374694 Chr.7: 30284640-30284640 on Build GRCh38
rs1004583734 Chr.7: 30284686-30284686 on Build GRCh38
rs534912238 Chr.7: 30284651-30284651 on Build GRCh38
rs1044865475 Chr.7: 30284408-30284408 on Build GRCh38
rs933192815 Chr.7: 30284267-30284267 on Build GRCh38
rs925400435 Chr.7: 30284306-30284306 on Build GRCh38
rs971238900 Chr.7: 30284595-30284595 on Build GRCh38
rs547522625 Chr.7: 30284351-30284351 on Build GRCh38
rs941864716 Chr.7: 30284508-30284508 on Build GRCh38
rs923086287 Chr.7: 30284262-30284262 on Build GRCh38
rs561952415 Chr.7: 30284308-30284308 on Build GRCh38
rs943788093 Chr.7: 30284678-30284678 on Build GRCh38
rs939678919 Chr.7: 30284441-30284441 on Build GRCh38
rs571744607 Chr.7: 30284576-30284577 on Build GRCh38
rs944222074 Chr.7: 30284626-30284626 on Build GRCh38

View Full Product Details