Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39623461-39623958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALA
Entrez Gene ID
5898
Gene Name
RALA Ras like proto-oncogene A
Gene Aliases
RAL
UniGene
Hs.6906
Gene Chromosome Location
Chr.7: 39622955-39708124 on Build GRCh38
Transcripts
NM_005402.3 XM_011515466.1 XM_011515467.1

SNP Detail

SNP ID Chromosome Location
rs890522366 Chr.7: 39623633-39623635 on Build GRCh38
rs991218452 Chr.7: 39623658-39623658 on Build GRCh38
rs982508467 Chr.7: 39623558-39623558 on Build GRCh38
rs916768451 Chr.7: 39623595-39623595 on Build GRCh38
rs990586218 Chr.7: 39623580-39623580 on Build GRCh38
rs1043936300 Chr.7: 39623710-39623710 on Build GRCh38
rs1013052414 Chr.7: 39623605-39623605 on Build GRCh38
rs1043708274 Chr.7: 39623482-39623482 on Build GRCh38
rs999929407 Chr.7: 39623608-39623608 on Build GRCh38
rs1056617237 Chr.7: 39623594-39623594 on Build GRCh38
rs568830599 Chr.7: 39623512-39623512 on Build GRCh38
rs935612123 Chr.7: 39623762-39623762 on Build GRCh38
rs867759039 Chr.7: 39623559-39623559 on Build GRCh38
rs919883576 Chr.7: 39623544-39623544 on Build GRCh38
rs1011100724 Chr.7: 39623875-39623875 on Build GRCh38
rs541683658 Chr.7: 39623899-39623899 on Build GRCh38
rs192369004 Chr.7: 39623489-39623489 on Build GRCh38
rs1000196028 Chr.7: 39623913-39623913 on Build GRCh38
rs1008992438 Chr.7: 39623647-39623655 on Build GRCh38
rs974381090 Chr.7: 39623534-39623534 on Build GRCh38
rs960411951 Chr.7: 39623744-39623744 on Build GRCh38
rs982821171 Chr.7: 39623650-39623650 on Build GRCh38
rs978229572 Chr.7: 39623552-39623552 on Build GRCh38
rs536543697 Chr.7: 39623824-39623824 on Build GRCh38
rs900656661 Chr.7: 39623867-39623867 on Build GRCh38
rs989733831 Chr.7: 39623532-39623532 on Build GRCh38
rs903833060 Chr.7: 39623483-39623483 on Build GRCh38
rs866077945 Chr.7: 39623492-39623492 on Build GRCh38
rs1052395667 Chr.7: 39623612-39623612 on Build GRCh38
rs796540644 Chr.7: 39623870-39623870 on Build GRCh38
rs995387465 Chr.7: 39623637-39623637 on Build GRCh38
rs939114696 Chr.7: 39623574-39623574 on Build GRCh38
rs572887912 Chr.7: 39623855-39623855 on Build GRCh38
rs1022571812 Chr.7: 39623684-39623685 on Build GRCh38
rs1052365311 Chr.7: 39623853-39623853 on Build GRCh38
rs1007983883 Chr.7: 39623515-39623515 on Build GRCh38
rs916414859 Chr.7: 39623758-39623758 on Build GRCh38
rs1027314816 Chr.7: 39623638-39623638 on Build GRCh38
rs928330875 Chr.7: 39623589-39623589 on Build GRCh38
rs1018339328 Chr.7: 39623620-39623620 on Build GRCh38
rs1017799336 Chr.7: 39623496-39623496 on Build GRCh38
rs915691192 Chr.7: 39623668-39623668 on Build GRCh38
rs929902391 Chr.7: 39623795-39623795 on Build GRCh38
rs990534382 Chr.7: 39623748-39623748 on Build GRCh38
rs951113391 Chr.7: 39623545-39623545 on Build GRCh38
rs1008286790 Chr.7: 39623619-39623619 on Build GRCh38
rs552975934 Chr.7: 39623842-39623842 on Build GRCh38
rs918535619 Chr.7: 39623779-39623779 on Build GRCh38
rs1026828071 Chr.7: 39623937-39623937 on Build GRCh38
rs964923866 Chr.7: 39623520-39623520 on Build GRCh38
rs867603417 Chr.7: 39623864-39623864 on Build GRCh38
rs951332599 Chr.7: 39623647-39623647 on Build GRCh38
rs902733545 Chr.7: 39623905-39623905 on Build GRCh38
rs757575311 Chr.7: 39623628-39623628 on Build GRCh38
rs938419741 Chr.7: 39623592-39623592 on Build GRCh38
rs1018079490 Chr.7: 39623516-39623516 on Build GRCh38
rs948269226 Chr.7: 39623599-39623599 on Build GRCh38
rs550852719 Chr.7: 39623601-39623601 on Build GRCh38
rs1026433739 Chr.7: 39623486-39623486 on Build GRCh38
rs955493465 Chr.7: 39623509-39623509 on Build GRCh38
rs993987040 Chr.7: 39623485-39623485 on Build GRCh38
rs1011154765 Chr.7: 39623658-39623659 on Build GRCh38
rs974384450 Chr.7: 39623767-39623767 on Build GRCh38
rs959752660 Chr.7: 39623652-39623652 on Build GRCh38
rs928298183 Chr.7: 39623651-39623651 on Build GRCh38
rs925485368 Chr.7: 39623721-39623721 on Build GRCh38
rs948210566 Chr.7: 39623703-39623703 on Build GRCh38
rs567655795 Chr.7: 39623602-39623602 on Build GRCh38
rs891183706 Chr.7: 39623613-39623613 on Build GRCh38
rs963906494 Chr.7: 39623519-39623519 on Build GRCh38
rs538623575 Chr.7: 39623858-39623858 on Build GRCh38
rs963833223 Chr.7: 39623629-39623629 on Build GRCh38
rs899953836 Chr.7: 39623903-39623903 on Build GRCh38
rs1022729662 Chr.7: 39623860-39623860 on Build GRCh38
rs537654996 Chr.7: 39623557-39623557 on Build GRCh38
rs752305388 Chr.7: 39623625-39623625 on Build GRCh38
rs995709081 Chr.7: 39623925-39623925 on Build GRCh38
rs966434756 Chr.7: 39623551-39623551 on Build GRCh38

View Full Product Details