Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44802079-44802585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIA
Entrez Gene ID
5478
Gene Name
peptidylprolyl isomerase A
Gene Aliases
CYPA, CYPH, HEL-S-69p
UniGene
Hs.356331
Gene Chromosome Location
Chr.7: 44796636-44803123 on Build GRCh38
Transcripts
NM_001300981.1 NM_021130.4

SNP Detail

SNP ID Chromosome Location
rs973170802 Chr.7: 44802405-44802406 on Build GRCh38
rs17860093 Chr.7: 44802321-44802321 on Build GRCh38
rs35100141 Chr.7: 44802174-44802175 on Build GRCh38
rs1027440800 Chr.7: 44802541-44802541 on Build GRCh38
rs896699724 Chr.7: 44802519-44802519 on Build GRCh38
rs180875884 Chr.7: 44802391-44802391 on Build GRCh38
rs961791636 Chr.7: 44802224-44802224 on Build GRCh38
rs905224399 Chr.7: 44802347-44802347 on Build GRCh38
rs190544520 Chr.7: 44802234-44802234 on Build GRCh38
rs186066318 Chr.7: 44802491-44802491 on Build GRCh38
rs1041703910 Chr.7: 44802472-44802472 on Build GRCh38
rs532605623 Chr.7: 44802210-44802210 on Build GRCh38
rs904657544 Chr.7: 44802314-44802314 on Build GRCh38
rs904500541 Chr.7: 44802483-44802483 on Build GRCh38
rs534741910 Chr.7: 44802226-44802226 on Build GRCh38
rs57534886 Chr.7: 44802192-44802192 on Build GRCh38
rs754063499 Chr.7: 44802507-44802507 on Build GRCh38
rs892512591 Chr.7: 44802307-44802307 on Build GRCh38
rs992978462 Chr.7: 44802190-44802190 on Build GRCh38
rs946784589 Chr.7: 44802488-44802488 on Build GRCh38
rs918372446 Chr.7: 44802218-44802218 on Build GRCh38
rs769049572 Chr.7: 44802420-44802420 on Build GRCh38
rs965665942 Chr.7: 44802379-44802379 on Build GRCh38
rs767884699 Chr.7: 44802116-44802116 on Build GRCh38
rs558361547 Chr.7: 44802293-44802293 on Build GRCh38
rs1057480991 Chr.7: 44802439-44802439 on Build GRCh38
rs953851611 Chr.7: 44802345-44802345 on Build GRCh38
rs539726282 Chr.7: 44802438-44802438 on Build GRCh38
rs547925179 Chr.7: 44802213-44802213 on Build GRCh38
rs568616291 Chr.7: 44802257-44802257 on Build GRCh38
rs181557589 Chr.7: 44802109-44802109 on Build GRCh38
rs773905068 Chr.7: 44802471-44802471 on Build GRCh38
rs920273857 Chr.7: 44802231-44802231 on Build GRCh38
rs112278761 Chr.7: 44802249-44802249 on Build GRCh38
rs1004887516 Chr.7: 44802526-44802526 on Build GRCh38
rs751464359 Chr.7: 44802191-44802192 on Build GRCh38
rs979270449 Chr.7: 44802139-44802139 on Build GRCh38
rs972797081 Chr.7: 44802217-44802217 on Build GRCh38
rs565236966 Chr.7: 44802510-44802510 on Build GRCh38
rs985361164 Chr.7: 44802200-44802200 on Build GRCh38
rs913755794 Chr.7: 44802482-44802482 on Build GRCh38
rs1048595500 Chr.7: 44802242-44802242 on Build GRCh38
rs905995511 Chr.7: 44802255-44802255 on Build GRCh38
rs898690749 Chr.7: 44802535-44802535 on Build GRCh38
rs1000249398 Chr.7: 44802504-44802504 on Build GRCh38
rs548241826 Chr.7: 44802239-44802239 on Build GRCh38
rs17860095 Chr.7: 44802440-44802440 on Build GRCh38
rs1007238452 Chr.7: 44802356-44802356 on Build GRCh38
rs28403980 Chr.7: 44802176-44802176 on Build GRCh38
rs554578118 Chr.7: 44802455-44802455 on Build GRCh38
rs10627827 Chr.7: 44802173-44802174 on Build GRCh38
rs567298870 Chr.7: 44802190-44802192 on Build GRCh38
rs889186398 Chr.7: 44802514-44802514 on Build GRCh38
rs931864716 Chr.7: 44802233-44802233 on Build GRCh38
rs17860094 Chr.7: 44802325-44802325 on Build GRCh38
rs149127139 Chr.7: 44802431-44802431 on Build GRCh38
rs983841480 Chr.7: 44802424-44802424 on Build GRCh38
rs1019863030 Chr.7: 44802538-44802538 on Build GRCh38
rs780290604 Chr.7: 44802366-44802366 on Build GRCh38
rs190180753 Chr.7: 44802529-44802529 on Build GRCh38
rs565514926 Chr.7: 44802201-44802201 on Build GRCh38
rs1002396865 Chr.7: 44802364-44802364 on Build GRCh38
rs1004342499 Chr.7: 44802388-44802388 on Build GRCh38
rs1016183688 Chr.7: 44802390-44802390 on Build GRCh38
rs7784201 Chr.7: 44802289-44802289 on Build GRCh38
rs944591276 Chr.7: 44802280-44802280 on Build GRCh38
rs760202453 Chr.7: 44802494-44802494 on Build GRCh38
rs896643259 Chr.7: 44802384-44802384 on Build GRCh38
rs60471039 Chr.7: 44802175-44802176 on Build GRCh38
rs531716578 Chr.7: 44802171-44802171 on Build GRCh38
rs749539510 Chr.7: 44802392-44802392 on Build GRCh38
rs560929101 Chr.7: 44802147-44802147 on Build GRCh38
rs989631533 Chr.7: 44802177-44802177 on Build GRCh38
rs943741067 Chr.7: 44802216-44802216 on Build GRCh38
rs1042104903 Chr.7: 44802294-44802294 on Build GRCh38
rs879288197 Chr.7: 44802235-44802235 on Build GRCh38
rs570990249 Chr.7: 44802323-44802323 on Build GRCh38
rs553096208 Chr.7: 44802368-44802368 on Build GRCh38
rs569649936 Chr.7: 44802275-44802275 on Build GRCh38
rs953968318 Chr.7: 44802159-44802159 on Build GRCh38
rs117681717 Chr.7: 44802191-44802191 on Build GRCh38

View Full Product Details