Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 72744082-72744594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYW1B
Entrez Gene ID
441250
Gene Name
tRNA-yW synthesizing protein 1 homolog B
Gene Aliases
LINC00069, NCRNA00069, RSAFD2
UniGene
Hs.488614
Gene Chromosome Location
Chr.7: 72574507-72828234 on Build GRCh38
Transcripts
NM_001145440.2 XM_011516230.2 XM_011516231.2 XM_011516232.2 XM_011516233.2 XM_017012221.1

SNP Detail

SNP ID Chromosome Location
rs782748869 Chr.7: 72744479-72744479 on Build GRCh38
rs575651173 Chr.7: 72744451-72744451 on Build GRCh38
rs567640261 Chr.7: 72744361-72744361 on Build GRCh38
rs782604845 Chr.7: 72744523-72744523 on Build GRCh38
rs538266643 Chr.7: 72744416-72744416 on Build GRCh38
rs369723478 Chr.7: 72744453-72744453 on Build GRCh38
rs207468086 Chr.7: 72744396-72744396 on Build GRCh38
rs782311963 Chr.7: 72744537-72744537 on Build GRCh38
rs2687015 Chr.7: 72744485-72744485 on Build GRCh38
rs184803775 Chr.7: 72744428-72744428 on Build GRCh38
rs781845199 Chr.7: 72744503-72744503 on Build GRCh38
rs782767508 Chr.7: 72744490-72744490 on Build GRCh38
rs782460068 Chr.7: 72744513-72744513 on Build GRCh38
rs192971967 Chr.7: 72744196-72744196 on Build GRCh38
rs782057909 Chr.7: 72744156-72744156 on Build GRCh38
rs782625699 Chr.7: 72744506-72744506 on Build GRCh38
rs782138897 Chr.7: 72744467-72744467 on Build GRCh38
rs781987940 Chr.7: 72744563-72744563 on Build GRCh38
rs558055883 Chr.7: 72744522-72744522 on Build GRCh38
rs2844104 Chr.7: 72744400-72744400 on Build GRCh38
rs782539370 Chr.7: 72744523-72744523 on Build GRCh38
rs782650215 Chr.7: 72744514-72744514 on Build GRCh38
rs781889635 Chr.7: 72744446-72744446 on Build GRCh38
rs555847834 Chr.7: 72744272-72744272 on Build GRCh38
rs782435864 Chr.7: 72744557-72744557 on Build GRCh38
rs782708042 Chr.7: 72744486-72744486 on Build GRCh38
rs782144257 Chr.7: 72744542-72744542 on Build GRCh38
rs781892655 Chr.7: 72744487-72744487 on Build GRCh38
rs546129513 Chr.7: 72744460-72744460 on Build GRCh38
rs566729179 Chr.7: 72744108-72744108 on Build GRCh38
rs796164773 Chr.7: 72744552-72744552 on Build GRCh38
rs781446415 Chr.7: 72744115-72744116 on Build GRCh38
rs782717676 Chr.7: 72744185-72744185 on Build GRCh38
rs2844102 Chr.7: 72744183-72744183 on Build GRCh38
rs781789516 Chr.7: 72744509-72744509 on Build GRCh38
rs782237838 Chr.7: 72744533-72744533 on Build GRCh38
rs782461069 Chr.7: 72744500-72744500 on Build GRCh38
rs543456380 Chr.7: 72744331-72744331 on Build GRCh38
rs781872590 Chr.7: 72744491-72744491 on Build GRCh38
rs782291559 Chr.7: 72744518-72744518 on Build GRCh38
rs782208895 Chr.7: 72744450-72744450 on Build GRCh38
rs782191530 Chr.7: 72744455-72744455 on Build GRCh38
rs782566385 Chr.7: 72744483-72744483 on Build GRCh38
rs748202129 Chr.7: 72744405-72744405 on Build GRCh38
rs781946271 Chr.7: 72744541-72744541 on Build GRCh38

View Full Product Details