Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73303593-73304085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NSUN5
Entrez Gene ID
55695
Gene Name
NOP2/Sun RNA methyltransferase family member 5
Gene Aliases
NOL1, NOL1R, NSUN5A, WBSCR20, WBSCR20A, p120, p120(NOL1)
UniGene
Hs.510927
Gene Chromosome Location
Chr.7: 73302516-73308867 on Build GRCh38
Transcripts
NM_001168347.2 NM_001168348.2 NM_018044.4 NM_148956.3 XM_005277604.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467195 c.1264A>C p.I422L Chr.7: 73303622-73303622 on Build GRCh38
3086849 c.1178G>A p.R393Q Chr.7: 73303708-73303708 on Build GRCh38
4984343 c.989G>A p.R330H Chr.7: 73303982-73303982 on Build GRCh38
3412263 c.1062C>T p.L354L Chr.7: 73303909-73303909 on Build GRCh38
3640290 c.1011C>T p.F337F Chr.7: 73303960-73303960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368919057 Chr.7: 73303802-73303802 on Build GRCh38
rs782076709 Chr.7: 73303784-73303784 on Build GRCh38
rs4030984 Chr.7: 73303780-73303780 on Build GRCh38
rs782723244 Chr.7: 73303892-73303892 on Build GRCh38
rs752850378 Chr.7: 73303837-73303837 on Build GRCh38
rs2353056 Chr.7: 73303643-73303643 on Build GRCh38
rs782587758 Chr.7: 73303853-73303853 on Build GRCh38
rs540752192 Chr.7: 73303768-73303768 on Build GRCh38
rs781860938 Chr.7: 73303982-73303982 on Build GRCh38
rs148120256 Chr.7: 73303862-73303862 on Build GRCh38
rs782366215 Chr.7: 73303736-73303736 on Build GRCh38
rs558140471 Chr.7: 73303753-73303753 on Build GRCh38
rs373541379 Chr.7: 73303860-73303860 on Build GRCh38
rs576497877 Chr.7: 73303761-73303761 on Build GRCh38
rs200246849 Chr.7: 73303690-73303690 on Build GRCh38
rs1044909 Chr.7: 73303665-73303665 on Build GRCh38
rs782138269 Chr.7: 73303747-73303747 on Build GRCh38
rs372362381 Chr.7: 73303696-73303696 on Build GRCh38
rs377011651 Chr.7: 73303911-73303911 on Build GRCh38
rs782327762 Chr.7: 73303937-73303937 on Build GRCh38
rs537004606 Chr.7: 73303740-73303740 on Build GRCh38
rs782122826 Chr.7: 73303815-73303815 on Build GRCh38
rs782586601 Chr.7: 73303776-73303776 on Build GRCh38
rs371339599 Chr.7: 73303758-73303758 on Build GRCh38
rs150043225 Chr.7: 73303644-73303644 on Build GRCh38
rs767712977 Chr.7: 73303794-73303794 on Build GRCh38
rs2883079 Chr.7: 73303647-73303647 on Build GRCh38
rs782015832 Chr.7: 73303614-73303615 on Build GRCh38
rs142509909 Chr.7: 73303987-73303987 on Build GRCh38
rs781857962 Chr.7: 73304051-73304051 on Build GRCh38
rs781806696 Chr.7: 73303833-73303833 on Build GRCh38
rs112696722 Chr.7: 73303852-73303852 on Build GRCh38
rs782288781 Chr.7: 73303635-73303635 on Build GRCh38
rs62000387 Chr.7: 73303834-73303834 on Build GRCh38
rs11544045 Chr.7: 73303912-73303912 on Build GRCh38
rs782708710 Chr.7: 73303822-73303822 on Build GRCh38
rs144929830 Chr.7: 73303989-73303989 on Build GRCh38
rs782024774 Chr.7: 73303803-73303803 on Build GRCh38
rs377460761 Chr.7: 73304008-73304008 on Build GRCh38
rs367945643 Chr.7: 73304057-73304057 on Build GRCh38
rs369982378 Chr.7: 73303730-73303730 on Build GRCh38
rs781915455 Chr.7: 73303965-73303965 on Build GRCh38
rs782688881 Chr.7: 73304011-73304011 on Build GRCh38
rs781834379 Chr.7: 73303684-73303684 on Build GRCh38
rs782647287 Chr.7: 73303714-73303714 on Build GRCh38
rs782749086 Chr.7: 73304050-73304050 on Build GRCh38
rs782134220 Chr.7: 73303988-73303988 on Build GRCh38
rs782423070 Chr.7: 73303867-73303867 on Build GRCh38
rs371885777 Chr.7: 73303683-73303683 on Build GRCh38
rs781953960 Chr.7: 73303731-73303731 on Build GRCh38
rs781884776 Chr.7: 73303691-73303691 on Build GRCh38
rs781810144 Chr.7: 73304041-73304041 on Build GRCh38
rs566215915 Chr.7: 73303925-73303925 on Build GRCh38
rs374741797 Chr.7: 73303625-73303625 on Build GRCh38
rs782687447 Chr.7: 73303931-73303931 on Build GRCh38
rs782753611 Chr.7: 73303757-73303757 on Build GRCh38
rs782124345 Chr.7: 73304022-73304022 on Build GRCh38
rs1044904 Chr.7: 73303658-73303658 on Build GRCh38
rs782068075 Chr.7: 73303682-73303682 on Build GRCh38
rs782671113 Chr.7: 73304056-73304056 on Build GRCh38
rs4030996 Chr.7: 73303861-73303861 on Build GRCh38
rs781911649 Chr.7: 73303995-73303995 on Build GRCh38
rs782774767 Chr.7: 73303908-73303908 on Build GRCh38
rs782661866 Chr.7: 73303843-73303843 on Build GRCh38
rs782264697 Chr.7: 73303926-73303926 on Build GRCh38
rs559054153 Chr.7: 73303769-73303769 on Build GRCh38
rs201272154 Chr.7: 73303813-73303813 on Build GRCh38
rs781921404 Chr.7: 73303670-73303670 on Build GRCh38
rs138888800 Chr.7: 73304048-73304048 on Build GRCh38
rs782437790 Chr.7: 73303999-73303999 on Build GRCh38
rs782744197 Chr.7: 73303835-73303835 on Build GRCh38
rs781884357 Chr.7: 73303909-73303909 on Build GRCh38
rs782007407 Chr.7: 73303657-73303657 on Build GRCh38
rs782038900 Chr.7: 73304038-73304038 on Build GRCh38
rs782035354 Chr.7: 73303795-73303795 on Build GRCh38
rs1044901 Chr.7: 73303650-73303650 on Build GRCh38
rs147944881 Chr.7: 73303721-73303721 on Build GRCh38
rs4030985 Chr.7: 73303788-73303788 on Build GRCh38
rs141000540 Chr.7: 73303884-73303884 on Build GRCh38
rs782152836 Chr.7: 73303666-73303666 on Build GRCh38
rs147725314 Chr.7: 73303669-73303669 on Build GRCh38
rs201636338 Chr.7: 73303971-73303971 on Build GRCh38
rs374889055 Chr.7: 73303953-73303953 on Build GRCh38
rs200398918 Chr.7: 73303913-73303913 on Build GRCh38
rs370283321 Chr.7: 73304016-73304016 on Build GRCh38
rs570093741 Chr.7: 73304007-73304007 on Build GRCh38
rs782484744 Chr.7: 73303708-73303708 on Build GRCh38
rs781948180 Chr.7: 73304019-73304019 on Build GRCh38
rs367919442 Chr.7: 73304005-73304005 on Build GRCh38
rs782691822 Chr.7: 73304035-73304035 on Build GRCh38
rs191161664 Chr.7: 73303817-73303817 on Build GRCh38
rs781996576 Chr.7: 73303950-73303950 on Build GRCh38
rs547895380 Chr.7: 73303898-73303898 on Build GRCh38
rs781800534 Chr.7: 73303914-73303914 on Build GRCh38
rs374061204 Chr.7: 73304029-73304029 on Build GRCh38
rs11544043 Chr.7: 73303936-73303936 on Build GRCh38
rs782461246 Chr.7: 73303616-73303616 on Build GRCh38
rs782503162 Chr.7: 73303917-73303917 on Build GRCh38
rs372980729 Chr.7: 73303812-73303812 on Build GRCh38
rs201057912 Chr.7: 73303615-73303615 on Build GRCh38
rs149254341 Chr.7: 73304018-73304018 on Build GRCh38
rs570286769 Chr.7: 73303966-73303966 on Build GRCh38
rs138754274 Chr.7: 73304010-73304010 on Build GRCh38
rs1044907 Chr.7: 73303659-73303659 on Build GRCh38
rs375947782 Chr.7: 73303729-73303729 on Build GRCh38
rs781968973 Chr.7: 73303897-73303897 on Build GRCh38
rs574559331 Chr.7: 73303804-73303804 on Build GRCh38
rs782590697 Chr.7: 73304000-73304000 on Build GRCh38
rs782378455 Chr.7: 73303947-73303947 on Build GRCh38
rs370620770 Chr.7: 73303878-73303878 on Build GRCh38
rs558103608 Chr.7: 73304003-73304003 on Build GRCh38
rs782684287 Chr.7: 73303723-73303723 on Build GRCh38
rs782512255 Chr.7: 73304055-73304055 on Build GRCh38
rs4030983 Chr.7: 73303754-73303754 on Build GRCh38
rs782784195 Chr.7: 73303816-73303816 on Build GRCh38
rs367998478 Chr.7: 73303631-73303631 on Build GRCh38
rs782284985 Chr.7: 73303866-73303866 on Build GRCh38
rs782095631 Chr.7: 73303734-73303734 on Build GRCh38
rs369705145 Chr.7: 73303821-73303821 on Build GRCh38
rs782542839 Chr.7: 73303928-73303928 on Build GRCh38
rs782148643 Chr.7: 73303952-73303952 on Build GRCh38
rs782359467 Chr.7: 73303787-73303787 on Build GRCh38
rs782715083 Chr.7: 73303976-73303976 on Build GRCh38
rs782224363 Chr.7: 73303943-73303943 on Build GRCh38
rs4030982 Chr.7: 73303718-73303718 on Build GRCh38
rs782284429 Chr.7: 73303846-73303846 on Build GRCh38
rs3950374 Chr.7: 73303859-73303859 on Build GRCh38
rs565300005 Chr.7: 73303838-73303838 on Build GRCh38
rs782701866 Chr.7: 73303788-73303789 on Build GRCh38
rs782513760 Chr.7: 73303630-73303630 on Build GRCh38
rs782167084 Chr.7: 73304030-73304030 on Build GRCh38
rs781957460 Chr.7: 73303777-73303777 on Build GRCh38
rs368500521 Chr.7: 73303695-73303695 on Build GRCh38
rs145622470 Chr.7: 73303689-73303689 on Build GRCh38
rs782621751 Chr.7: 73303623-73303623 on Build GRCh38
rs782498693 Chr.7: 73304006-73304006 on Build GRCh38

View Full Product Details