Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99909178-99909695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM4
Entrez Gene ID
89122
Gene Name
tripartite motif containing 4
Gene Aliases
RNF87
UniGene
Hs.50749
Gene Chromosome Location
Chr.7: 99890407-99919600 on Build GRCh38
Transcripts
NM_033017.3 NM_033091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6515307 c.424G>T p.V142L Chr.7: 99909630-99909630 on Build GRCh38
4736086 c.416G>A p.R139H Chr.7: 99909638-99909638 on Build GRCh38
6297826 c.559C>A p.Q187K Chr.7: 99909573-99909573 on Build GRCh38
4411111 c.427G>C p.A143P Chr.7: 99909627-99909627 on Build GRCh38
7297105 c.475G>A p.A159T Chr.7: 99909579-99909579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73405434 Chr.7: 99909358-99909358 on Build GRCh38
rs748364312 Chr.7: 99909561-99909562 on Build GRCh38
rs376793435 Chr.7: 99909577-99909577 on Build GRCh38
rs769138861 Chr.7: 99909542-99909542 on Build GRCh38
rs781651283 Chr.7: 99909658-99909658 on Build GRCh38
rs1019325171 Chr.7: 99909594-99909594 on Build GRCh38
rs747105293 Chr.7: 99909533-99909533 on Build GRCh38
rs889092244 Chr.7: 99909531-99909531 on Build GRCh38
rs143668456 Chr.7: 99909600-99909600 on Build GRCh38
rs971330158 Chr.7: 99909300-99909300 on Build GRCh38
rs575082225 Chr.7: 99909462-99909462 on Build GRCh38
rs772216262 Chr.7: 99909524-99909524 on Build GRCh38
rs995109636 Chr.7: 99909232-99909232 on Build GRCh38
rs997961172 Chr.7: 99909242-99909242 on Build GRCh38
rs755841618 Chr.7: 99909631-99909631 on Build GRCh38
rs1008429977 Chr.7: 99909299-99909299 on Build GRCh38
rs565062091 Chr.7: 99909641-99909641 on Build GRCh38
rs972542787 Chr.7: 99909287-99909287 on Build GRCh38
rs748642361 Chr.7: 99909664-99909664 on Build GRCh38
rs188783002 Chr.7: 99909607-99909607 on Build GRCh38
rs777192846 Chr.7: 99909564-99909564 on Build GRCh38
rs774791061 Chr.7: 99909568-99909568 on Build GRCh38
rs762447808 Chr.7: 99909272-99909272 on Build GRCh38
rs768768697 Chr.7: 99909654-99909654 on Build GRCh38
rs902103401 Chr.7: 99909212-99909212 on Build GRCh38
rs926957044 Chr.7: 99909338-99909338 on Build GRCh38
rs946259333 Chr.7: 99909423-99909423 on Build GRCh38
rs558703541 Chr.7: 99909248-99909248 on Build GRCh38
rs956192263 Chr.7: 99909323-99909323 on Build GRCh38
rs183936802 Chr.7: 99909438-99909438 on Build GRCh38
rs759934856 Chr.7: 99909592-99909592 on Build GRCh38
rs777573423 Chr.7: 99909638-99909638 on Build GRCh38
rs1041978187 Chr.7: 99909210-99909210 on Build GRCh38
rs762624048 Chr.7: 99909579-99909579 on Build GRCh38
rs112979582 Chr.7: 99909262-99909262 on Build GRCh38
rs914306603 Chr.7: 99909402-99909402 on Build GRCh38
rs770246153 Chr.7: 99909574-99909574 on Build GRCh38
rs201741037 Chr.7: 99909628-99909628 on Build GRCh38
rs769390069 Chr.7: 99909282-99909282 on Build GRCh38
rs989475006 Chr.7: 99909333-99909333 on Build GRCh38
rs769067170 Chr.7: 99909461-99909461 on Build GRCh38
rs541316978 Chr.7: 99909360-99909360 on Build GRCh38
rs1025445082 Chr.7: 99909662-99909662 on Build GRCh38
rs778971772 Chr.7: 99909519-99909519 on Build GRCh38
rs541832124 Chr.7: 99909483-99909483 on Build GRCh38
rs771053588 Chr.7: 99909448-99909448 on Build GRCh38
rs747088496 Chr.7: 99909650-99909650 on Build GRCh38
rs1020198627 Chr.7: 99909308-99909308 on Build GRCh38
rs1028958600 Chr.7: 99909255-99909255 on Build GRCh38
rs773556525 Chr.7: 99909669-99909669 on Build GRCh38
rs902110057 Chr.7: 99909460-99909460 on Build GRCh38
rs747159641 Chr.7: 99909354-99909354 on Build GRCh38
rs968731245 Chr.7: 99909355-99909355 on Build GRCh38
rs898397822 Chr.7: 99909538-99909538 on Build GRCh38
rs775162479 Chr.7: 99909288-99909288 on Build GRCh38
rs1021855442 Chr.7: 99909339-99909339 on Build GRCh38
rs889201198 Chr.7: 99909257-99909257 on Build GRCh38
rs780043118 Chr.7: 99909527-99909527 on Build GRCh38
rs746150245 Chr.7: 99909523-99909523 on Build GRCh38
rs971739630 Chr.7: 99909665-99909665 on Build GRCh38
rs981518121 Chr.7: 99909325-99909325 on Build GRCh38
rs1050873195 Chr.7: 99909515-99909515 on Build GRCh38
rs962564788 Chr.7: 99909286-99909286 on Build GRCh38
rs1016788704 Chr.7: 99909272-99909272 on Build GRCh38
rs201165325 Chr.7: 99909580-99909580 on Build GRCh38
rs373018289 Chr.7: 99909630-99909630 on Build GRCh38
rs1006280208 Chr.7: 99909268-99909268 on Build GRCh38
rs376234532 Chr.7: 99909639-99909639 on Build GRCh38

View Full Product Details