Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100463879-100464378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf61
Entrez Gene ID
402573
Gene Name
chromosome 7 open reading frame 61
Gene Aliases
-
UniGene
Hs.632306
Gene Chromosome Location
Chr.7: 100456615-100464271 on Build GRCh38
Transcripts
NM_001004323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1453430 c.51G>C p.R17S Chr.7: 100463975-100463975 on Build GRCh38
5987072 c.37C>T p.R13W Chr.7: 100463989-100463989 on Build GRCh38
389777 c.29G>C p.W10S Chr.7: 100463997-100463997 on Build GRCh38
3831282 c.60C>T p.S20S Chr.7: 100463966-100463966 on Build GRCh38
3949569 c.62G>T p.W21L Chr.7: 100463964-100463964 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749751067 Chr.7: 100464021-100464021 on Build GRCh38
rs946826987 Chr.7: 100464085-100464085 on Build GRCh38
rs747158181 Chr.7: 100464043-100464044 on Build GRCh38
rs1053643755 Chr.7: 100464326-100464326 on Build GRCh38
rs371514811 Chr.7: 100463934-100463934 on Build GRCh38
rs1057188711 Chr.7: 100464099-100464099 on Build GRCh38
rs779108406 Chr.7: 100464195-100464195 on Build GRCh38
rs556572780 Chr.7: 100464294-100464294 on Build GRCh38
rs763278654 Chr.7: 100463936-100463936 on Build GRCh38
rs908867029 Chr.7: 100464314-100464314 on Build GRCh38
rs535071559 Chr.7: 100464208-100464208 on Build GRCh38
rs376639804 Chr.7: 100463939-100463939 on Build GRCh38
rs369577590 Chr.7: 100464005-100464005 on Build GRCh38
rs559167894 Chr.7: 100464017-100464017 on Build GRCh38
rs768556949 Chr.7: 100463929-100463929 on Build GRCh38
rs990179383 Chr.7: 100464200-100464200 on Build GRCh38
rs112473927 Chr.7: 100464100-100464100 on Build GRCh38
rs745412227 Chr.7: 100463899-100463899 on Build GRCh38
rs908767747 Chr.7: 100464138-100464138 on Build GRCh38
rs1044481830 Chr.7: 100464252-100464252 on Build GRCh38
rs777913675 Chr.7: 100463989-100463990 on Build GRCh38
rs775412546 Chr.7: 100463913-100463913 on Build GRCh38
rs778552272 Chr.7: 100464000-100464000 on Build GRCh38
rs868463264 Chr.7: 100464171-100464171 on Build GRCh38
rs530862954 Chr.7: 100463928-100463928 on Build GRCh38
rs745459414 Chr.7: 100464001-100464001 on Build GRCh38
rs865897108 Chr.7: 100464172-100464172 on Build GRCh38
rs920969861 Chr.7: 100464077-100464077 on Build GRCh38
rs892345137 Chr.7: 100464327-100464327 on Build GRCh38
rs776594258 Chr.7: 100464032-100464032 on Build GRCh38
rs1032201455 Chr.7: 100463979-100463979 on Build GRCh38
rs373703442 Chr.7: 100464076-100464076 on Build GRCh38
rs940192760 Chr.7: 100464186-100464186 on Build GRCh38
rs1030126460 Chr.7: 100464110-100464110 on Build GRCh38
rs767633973 Chr.7: 100463981-100463981 on Build GRCh38
rs948849675 Chr.7: 100464232-100464232 on Build GRCh38
rs200606904 Chr.7: 100463989-100463989 on Build GRCh38
rs764569240 Chr.7: 100463988-100463988 on Build GRCh38
rs879202625 Chr.7: 100464092-100464092 on Build GRCh38
rs377560034 Chr.7: 100463995-100463995 on Build GRCh38
rs574527620 Chr.7: 100464096-100464096 on Build GRCh38
rs984402564 Chr.7: 100464117-100464117 on Build GRCh38
rs956566535 Chr.7: 100463986-100463986 on Build GRCh38
rs747019846 Chr.7: 100464022-100464022 on Build GRCh38
rs999887012 Chr.7: 100464141-100464141 on Build GRCh38
rs878930291 Chr.7: 100464088-100464088 on Build GRCh38
rs369568702 Chr.7: 100463907-100463907 on Build GRCh38
rs895454120 Chr.7: 100464104-100464104 on Build GRCh38
rs768451070 Chr.7: 100464027-100464027 on Build GRCh38
rs748325933 Chr.7: 100464278-100464278 on Build GRCh38
rs1000139196 Chr.7: 100464315-100464315 on Build GRCh38
rs747981545 Chr.7: 100464038-100464038 on Build GRCh38
rs1048474580 Chr.7: 100464309-100464309 on Build GRCh38
rs200926910 Chr.7: 100463982-100463982 on Build GRCh38
rs941665357 Chr.7: 100464316-100464316 on Build GRCh38
rs780540290 Chr.7: 100463953-100463953 on Build GRCh38
rs774663214 Chr.7: 100463945-100463945 on Build GRCh38
rs553896089 Chr.7: 100464271-100464271 on Build GRCh38
rs759699614 Chr.7: 100463971-100463971 on Build GRCh38
rs952340937 Chr.7: 100464111-100464111 on Build GRCh38

View Full Product Details