Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100488468-100488956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYAP1
Entrez Gene ID
222950
Gene Name
neuronal tyrosine phosphorylated phosphoinositide-3-kinase adaptor 1
Gene Aliases
C7orf51
UniGene
Hs.320086
Gene Chromosome Location
Chr.7: 100483872-100494802 on Build GRCh38
Transcripts
NM_173564.3 XM_006715907.3 XM_017011868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5606828 c.792G>A p.M264I Chr.7: 100488513-100488513 on Build GRCh38
6732012 c.1227G>A p.P409P Chr.7: 100488948-100488948 on Build GRCh38
3922283 c.1092G>A p.K364K Chr.7: 100488813-100488813 on Build GRCh38
6175564 c.1069C>T p.L357L Chr.7: 100488790-100488790 on Build GRCh38
6732010 c.1060G>A p.V354I Chr.7: 100488781-100488781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141511133 Chr.7: 100488725-100488725 on Build GRCh38
rs755407329 Chr.7: 100488544-100488544 on Build GRCh38
rs142406376 Chr.7: 100488657-100488657 on Build GRCh38
rs746504435 Chr.7: 100488829-100488829 on Build GRCh38
rs140403396 Chr.7: 100488636-100488636 on Build GRCh38
rs770606536 Chr.7: 100488609-100488609 on Build GRCh38
rs759436289 Chr.7: 100488524-100488524 on Build GRCh38
rs747720979 Chr.7: 100488490-100488490 on Build GRCh38
rs778253643 Chr.7: 100488659-100488659 on Build GRCh38
rs751599850 Chr.7: 100488648-100488648 on Build GRCh38
rs372128959 Chr.7: 100488842-100488842 on Build GRCh38
rs748927970 Chr.7: 100488608-100488608 on Build GRCh38
rs896624520 Chr.7: 100488578-100488578 on Build GRCh38
rs117099690 Chr.7: 100488867-100488867 on Build GRCh38
rs775435240 Chr.7: 100488622-100488622 on Build GRCh38
rs751615498 Chr.7: 100488585-100488585 on Build GRCh38
rs748872578 Chr.7: 100488554-100488554 on Build GRCh38
rs202055895 Chr.7: 100488801-100488801 on Build GRCh38
rs775171542 Chr.7: 100488567-100488567 on Build GRCh38
rs755900669 Chr.7: 100488926-100488926 on Build GRCh38
rs757663289 Chr.7: 100488540-100488540 on Build GRCh38
rs574707021 Chr.7: 100488584-100488584 on Build GRCh38
rs760408557 Chr.7: 100488572-100488572 on Build GRCh38
rs540259144 Chr.7: 100488764-100488764 on Build GRCh38
rs201255293 Chr.7: 100488591-100488591 on Build GRCh38
rs979799343 Chr.7: 100488903-100488903 on Build GRCh38
rs760533187 Chr.7: 100488625-100488625 on Build GRCh38
rs774397288 Chr.7: 100488562-100488562 on Build GRCh38
rs762146711 Chr.7: 100488871-100488871 on Build GRCh38
rs761534576 Chr.7: 100488736-100488736 on Build GRCh38
rs905275171 Chr.7: 100488596-100488596 on Build GRCh38
rs771921544 Chr.7: 100488617-100488617 on Build GRCh38
rs767610039 Chr.7: 100488763-100488763 on Build GRCh38
rs759592245 Chr.7: 100488760-100488760 on Build GRCh38
rs774627495 Chr.7: 100488755-100488755 on Build GRCh38
rs373777983 Chr.7: 100488521-100488521 on Build GRCh38
rs777653190 Chr.7: 100488929-100488929 on Build GRCh38
rs774424184 Chr.7: 100488522-100488522 on Build GRCh38
rs145967305 Chr.7: 100488662-100488662 on Build GRCh38
rs750085796 Chr.7: 100488875-100488875 on Build GRCh38
rs141533724 Chr.7: 100488528-100488528 on Build GRCh38
rs781163986 Chr.7: 100488924-100488924 on Build GRCh38
rs765956216 Chr.7: 100488886-100488886 on Build GRCh38
rs777277373 Chr.7: 100488868-100488868 on Build GRCh38
rs1036551932 Chr.7: 100488534-100488534 on Build GRCh38
rs940581449 Chr.7: 100488716-100488716 on Build GRCh38
rs552968300 Chr.7: 100488563-100488563 on Build GRCh38
rs879308330 Chr.7: 100488928-100488928 on Build GRCh38
rs767974873 Chr.7: 100488651-100488651 on Build GRCh38
rs986963113 Chr.7: 100488602-100488602 on Build GRCh38
rs368554898 Chr.7: 100488601-100488601 on Build GRCh38
rs558145623 Chr.7: 100488512-100488512 on Build GRCh38
rs748095840 Chr.7: 100488925-100488925 on Build GRCh38
rs763190781 Chr.7: 100488644-100488644 on Build GRCh38
rs148267102 Chr.7: 100488546-100488546 on Build GRCh38
rs754476304 Chr.7: 100488921-100488921 on Build GRCh38
rs770549465 Chr.7: 100488555-100488555 on Build GRCh38
rs758533298 Chr.7: 100488582-100488582 on Build GRCh38
rs746929783 Chr.7: 100488722-100488722 on Build GRCh38
rs138038785 Chr.7: 100488649-100488649 on Build GRCh38
rs769617753 Chr.7: 100488852-100488852 on Build GRCh38
rs781679581 Chr.7: 100488594-100488594 on Build GRCh38
rs199706701 Chr.7: 100488580-100488580 on Build GRCh38
rs984511541 Chr.7: 100488920-100488920 on Build GRCh38
rs750079254 Chr.7: 100488821-100488821 on Build GRCh38
rs959739987 Chr.7: 100488932-100488932 on Build GRCh38
rs745378079 Chr.7: 100488679-100488679 on Build GRCh38
rs976301402 Chr.7: 100488650-100488650 on Build GRCh38
rs748002884 Chr.7: 100488850-100488850 on Build GRCh38
rs746070291 Chr.7: 100488854-100488854 on Build GRCh38
rs1015972777 Chr.7: 100488558-100488558 on Build GRCh38
rs758410182 Chr.7: 100488535-100488535 on Build GRCh38
rs780607719 Chr.7: 100488536-100488536 on Build GRCh38
rs371835236 Chr.7: 100488527-100488527 on Build GRCh38
rs148577061 Chr.7: 100488775-100488775 on Build GRCh38
rs778570072 Chr.7: 100488670-100488670 on Build GRCh38
rs562210404 Chr.7: 100488799-100488799 on Build GRCh38
rs775736706 Chr.7: 100488860-100488860 on Build GRCh38
rs779195344 Chr.7: 100488565-100488565 on Build GRCh38
rs765454360 Chr.7: 100488573-100488573 on Build GRCh38
rs771611386 Chr.7: 100488696-100488696 on Build GRCh38
rs779643944 Chr.7: 100488823-100488823 on Build GRCh38
rs746471660 Chr.7: 100488826-100488826 on Build GRCh38
rs777625057 Chr.7: 100488604-100488604 on Build GRCh38
rs144455324 Chr.7: 100488737-100488737 on Build GRCh38
rs543235608 Chr.7: 100488613-100488613 on Build GRCh38
rs781437614 Chr.7: 100488545-100488545 on Build GRCh38
rs372242225 Chr.7: 100488632-100488632 on Build GRCh38
rs750519084 Chr.7: 100488529-100488529 on Build GRCh38
rs376041666 Chr.7: 100488858-100488858 on Build GRCh38
rs200499928 Chr.7: 100488859-100488859 on Build GRCh38
rs767195579 Chr.7: 100488525-100488525 on Build GRCh38
rs748988680 Chr.7: 100488931-100488931 on Build GRCh38
rs769526948 Chr.7: 100488498-100488498 on Build GRCh38
rs745333637 Chr.7: 100488616-100488616 on Build GRCh38
rs772867483 Chr.7: 100488510-100488510 on Build GRCh38
rs374169379 Chr.7: 100488658-100488658 on Build GRCh38
rs747449301 Chr.7: 100488542-100488542 on Build GRCh38
rs778501741 Chr.7: 100488805-100488805 on Build GRCh38
rs142980120 Chr.7: 100488777-100488777 on Build GRCh38
rs202155758 Chr.7: 100488702-100488702 on Build GRCh38
rs150655253 Chr.7: 100488822-100488822 on Build GRCh38
rs760779356 Chr.7: 100488866-100488866 on Build GRCh38
rs761297215 Chr.7: 100488774-100488774 on Build GRCh38
rs1000868194 Chr.7: 100488785-100488785 on Build GRCh38
rs751069018 Chr.7: 100488915-100488915 on Build GRCh38
rs145226359 Chr.7: 100488663-100488663 on Build GRCh38
rs748465487 Chr.7: 100488600-100488600 on Build GRCh38
rs754919526 Chr.7: 100488831-100488831 on Build GRCh38
rs868289869 Chr.7: 100488820-100488820 on Build GRCh38
rs769584217 Chr.7: 100488547-100488547 on Build GRCh38
rs150340305 Chr.7: 100488645-100488645 on Build GRCh38
rs376546614 Chr.7: 100488878-100488878 on Build GRCh38
rs772931629 Chr.7: 100488548-100488548 on Build GRCh38
rs139880279 Chr.7: 100488853-100488853 on Build GRCh38
rs776430302 Chr.7: 100488732-100488732 on Build GRCh38

View Full Product Details