Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100678955-100679456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB2
Entrez Gene ID
2783
Gene Name
G protein subunit beta 2
Gene Aliases
-
UniGene
Hs.185172
Gene Chromosome Location
Chr.7: 100673740-100679169 on Build GRCh38
Transcripts
NM_005273.3

SNP Detail

SNP ID Chromosome Location
rs943867307 Chr.7: 100679417-100679417 on Build GRCh38
rs953957331 Chr.7: 100679251-100679251 on Build GRCh38
rs546914437 Chr.7: 100679087-100679087 on Build GRCh38
rs931082408 Chr.7: 100679389-100679389 on Build GRCh38
rs1052679 Chr.7: 100678995-100678995 on Build GRCh38
rs929886202 Chr.7: 100679344-100679344 on Build GRCh38
rs1003620621 Chr.7: 100679212-100679212 on Build GRCh38
rs913968200 Chr.7: 100679270-100679270 on Build GRCh38
rs563378471 Chr.7: 100679308-100679308 on Build GRCh38
rs1052713 Chr.7: 100679022-100679022 on Build GRCh38
rs1049557460 Chr.7: 100679334-100679334 on Build GRCh38
rs541196930 Chr.7: 100679223-100679223 on Build GRCh38
rs895241921 Chr.7: 100679373-100679373 on Build GRCh38
rs1056539769 Chr.7: 100679181-100679181 on Build GRCh38
rs1006771601 Chr.7: 100679016-100679016 on Build GRCh38
rs910996894 Chr.7: 100679309-100679309 on Build GRCh38
rs535520974 Chr.7: 100679118-100679118 on Build GRCh38
rs774276537 Chr.7: 100679108-100679112 on Build GRCh38
rs1035142494 Chr.7: 100679232-100679232 on Build GRCh38
rs1028236554 Chr.7: 100679238-100679238 on Build GRCh38
rs946414157 Chr.7: 100679000-100679000 on Build GRCh38
rs555727143 Chr.7: 100679428-100679428 on Build GRCh38
rs117099626 Chr.7: 100679339-100679339 on Build GRCh38
rs915519703 Chr.7: 100679063-100679063 on Build GRCh38
rs751656001 Chr.7: 100679126-100679126 on Build GRCh38
rs1047006688 Chr.7: 100679353-100679353 on Build GRCh38
rs995270428 Chr.7: 100679047-100679047 on Build GRCh38
rs1022514121 Chr.7: 100679264-100679264 on Build GRCh38
rs988659414 Chr.7: 100679270-100679275 on Build GRCh38
rs754175213 Chr.7: 100679375-100679375 on Build GRCh38
rs1033868198 Chr.7: 100679369-100679369 on Build GRCh38
rs918159215 Chr.7: 100679377-100679378 on Build GRCh38
rs1001849793 Chr.7: 100679068-100679074 on Build GRCh38
rs541909218 Chr.7: 100679301-100679301 on Build GRCh38
rs939722105 Chr.7: 100679378-100679378 on Build GRCh38
rs990451110 Chr.7: 100678988-100678988 on Build GRCh38
rs1053040990 Chr.7: 100679059-100679059 on Build GRCh38
rs959560733 Chr.7: 100679061-100679061 on Build GRCh38
rs558803838 Chr.7: 100679199-100679199 on Build GRCh38
rs557041052 Chr.7: 100679072-100679072 on Build GRCh38
rs547286204 Chr.7: 100679382-100679382 on Build GRCh38
rs535211810 Chr.7: 100679079-100679079 on Build GRCh38
rs11520986 Chr.7: 100679371-100679371 on Build GRCh38
rs968721891 Chr.7: 100679281-100679281 on Build GRCh38
rs966528148 Chr.7: 100679117-100679117 on Build GRCh38
rs184460999 Chr.7: 100679365-100679365 on Build GRCh38
rs548329910 Chr.7: 100679055-100679055 on Build GRCh38
rs931956154 Chr.7: 100679035-100679035 on Build GRCh38
rs559256726 Chr.7: 100679272-100679272 on Build GRCh38
rs986208962 Chr.7: 100679050-100679050 on Build GRCh38
rs1056548623 Chr.7: 100679372-100679372 on Build GRCh38
rs942523560 Chr.7: 100679314-100679314 on Build GRCh38
rs994820575 Chr.7: 100679165-100679165 on Build GRCh38
rs1803663 Chr.7: 100679034-100679034 on Build GRCh38
rs763404841 Chr.7: 100679088-100679088 on Build GRCh38
rs995397764 Chr.7: 100679100-100679100 on Build GRCh38
rs575194149 Chr.7: 100679174-100679174 on Build GRCh38
rs914902723 Chr.7: 100678998-100678998 on Build GRCh38
rs552858295 Chr.7: 100679068-100679068 on Build GRCh38
rs942579349 Chr.7: 100679147-100679147 on Build GRCh38
rs1804552 Chr.7: 100679075-100679075 on Build GRCh38
rs191644831 Chr.7: 100679204-100679204 on Build GRCh38
rs968296726 Chr.7: 100679383-100679383 on Build GRCh38
rs1035193348 Chr.7: 100679060-100679060 on Build GRCh38
rs189780826 Chr.7: 100679003-100679003 on Build GRCh38
rs1019513246 Chr.7: 100679348-100679348 on Build GRCh38
rs950876939 Chr.7: 100679057-100679057 on Build GRCh38
rs547765336 Chr.7: 100679041-100679041 on Build GRCh38
rs1050853133 Chr.7: 100679108-100679108 on Build GRCh38
rs933809531 Chr.7: 100679005-100679005 on Build GRCh38
rs898436358 Chr.7: 100679160-100679160 on Build GRCh38
rs4727459 Chr.7: 100679198-100679198 on Build GRCh38
rs568659479 Chr.7: 100679089-100679089 on Build GRCh38
rs530517890 Chr.7: 100679338-100679338 on Build GRCh38
rs1021082189 Chr.7: 100679359-100679359 on Build GRCh38
rs540907185 Chr.7: 100679010-100679010 on Build GRCh38
rs904886148 Chr.7: 100679065-100679072 on Build GRCh38
rs565033701 Chr.7: 100679001-100679001 on Build GRCh38
rs1012447927 Chr.7: 100679260-100679260 on Build GRCh38
rs901864113 Chr.7: 100679350-100679350 on Build GRCh38
rs563243432 Chr.7: 100679058-100679058 on Build GRCh38
rs571104378 Chr.7: 100679073-100679073 on Build GRCh38
rs968245235 Chr.7: 100679271-100679276 on Build GRCh38
rs566040333 Chr.7: 100679062-100679062 on Build GRCh38
rs953403367 Chr.7: 100679101-100679101 on Build GRCh38
rs557044221 Chr.7: 100679134-100679134 on Build GRCh38
rs758421314 Chr.7: 100679399-100679399 on Build GRCh38
rs568274188 Chr.7: 100679416-100679416 on Build GRCh38
rs977744813 Chr.7: 100679283-100679283 on Build GRCh38
rs573029189 Chr.7: 100679431-100679431 on Build GRCh38
rs1010532462 Chr.7: 100679432-100679432 on Build GRCh38
rs968130474 Chr.7: 100679363-100679363 on Build GRCh38
rs559381567 Chr.7: 100679032-100679032 on Build GRCh38
rs1003934982 Chr.7: 100679083-100679083 on Build GRCh38
rs959509913 Chr.7: 100679234-100679234 on Build GRCh38
rs752432702 Chr.7: 100678995-100678997 on Build GRCh38
rs962496155 Chr.7: 100679098-100679098 on Build GRCh38
rs947826845 Chr.7: 100679265-100679281 on Build GRCh38
rs1038232373 Chr.7: 100679154-100679154 on Build GRCh38
rs955077003 Chr.7: 100679295-100679295 on Build GRCh38
rs1016842410 Chr.7: 100679021-100679021 on Build GRCh38
rs545304637 Chr.7: 100679352-100679352 on Build GRCh38
rs978172296 Chr.7: 100679084-100679084 on Build GRCh38
rs979586445 Chr.7: 100679275-100679275 on Build GRCh38
rs558256133 Chr.7: 100679292-100679292 on Build GRCh38
rs914069153 Chr.7: 100679127-100679127 on Build GRCh38
rs926858605 Chr.7: 100679153-100679153 on Build GRCh38
rs938249639 Chr.7: 100679282-100679282 on Build GRCh38
rs889610400 Chr.7: 100679137-100679137 on Build GRCh38
rs759424579 Chr.7: 100679197-100679199 on Build GRCh38
rs889662549 Chr.7: 100679013-100679013 on Build GRCh38
rs1043971371 Chr.7: 100679064-100679064 on Build GRCh38
rs1044024110 Chr.7: 100679152-100679152 on Build GRCh38

View Full Product Details