Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111123029-111123551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMMP2L
Entrez Gene ID
83943
Gene Name
inner mitochondrial membrane peptidase subunit 2
Gene Aliases
IMMP2L-IT1, IMP2, IMP2-LIKE
UniGene
Hs.655722
Gene Chromosome Location
Chr.7: 110662645-111562531 on Build GRCh38
Transcripts
NM_001244606.1 NM_032549.3 XM_005250630.3 XM_011516605.2 XM_011516608.2 XM_011516609.2 XM_011516613.2 XM_017012699.1 XM_017012700.1 XM_017012701.1 XM_017012702.1 XM_017012703.1 XM_017012704.1 XM_017012705.1 NM_001099658.1 NM_001099660.1 NM_018334.4

Gene Symbol
LRRN3
Entrez Gene ID
54674
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
FIGLER5, NLRR-3, NLRR3
UniGene
Hs.3781
Gene Chromosome Location
Chr.7: 111091006-111125454 on Build GRCh38
Transcripts
NM_001244606.1 NM_032549.3 XM_005250630.3 XM_011516605.2 XM_011516608.2 XM_011516609.2 XM_011516613.2 XM_017012699.1 XM_017012700.1 XM_017012701.1 XM_017012702.1 XM_017012703.1 XM_017012704.1 XM_017012705.1 NM_001099658.1 NM_001099660.1 NM_018334.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4487163 c.314C>T p.S105F Chr.7: 111123086-111123086 on Build GRCh38
5440713 c.658G>A p.A220T Chr.7: 111123430-111123430 on Build GRCh38
6340134 c.417G>T p.L139L Chr.7: 111123189-111123189 on Build GRCh38
201078 c.690C>T p.N230N Chr.7: 111123462-111123462 on Build GRCh38
1213995 c.440T>A p.I147N Chr.7: 111123212-111123212 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368805422 Chr.7: 111123248-111123248 on Build GRCh38
rs776346546 Chr.7: 111123280-111123280 on Build GRCh38
rs770573988 Chr.7: 111123440-111123440 on Build GRCh38
rs868362652 Chr.7: 111123182-111123182 on Build GRCh38
rs970043745 Chr.7: 111123274-111123274 on Build GRCh38
rs35229264 Chr.7: 111123447-111123447 on Build GRCh38
rs758987493 Chr.7: 111123220-111123220 on Build GRCh38
rs760301401 Chr.7: 111123386-111123386 on Build GRCh38
rs939340462 Chr.7: 111123121-111123121 on Build GRCh38
rs536637511 Chr.7: 111123345-111123345 on Build GRCh38
rs567710418 Chr.7: 111123210-111123210 on Build GRCh38
rs1035980047 Chr.7: 111123356-111123356 on Build GRCh38
rs1016030551 Chr.7: 111123216-111123216 on Build GRCh38
rs762758462 Chr.7: 111123453-111123453 on Build GRCh38
rs770209205 Chr.7: 111123221-111123221 on Build GRCh38
rs759038442 Chr.7: 111123516-111123516 on Build GRCh38
rs17853393 Chr.7: 111123058-111123058 on Build GRCh38
rs267601235 Chr.7: 111123423-111123423 on Build GRCh38
rs200291058 Chr.7: 111123416-111123416 on Build GRCh38
rs761798162 Chr.7: 111123463-111123463 on Build GRCh38
rs766534500 Chr.7: 111123506-111123506 on Build GRCh38
rs214865 Chr.7: 111123507-111123507 on Build GRCh38
rs111242959 Chr.7: 111123206-111123206 on Build GRCh38
rs759192602 Chr.7: 111123311-111123311 on Build GRCh38
rs759403715 Chr.7: 111123099-111123099 on Build GRCh38
rs762992794 Chr.7: 111123487-111123488 on Build GRCh38
rs371479262 Chr.7: 111123505-111123505 on Build GRCh38
rs757334230 Chr.7: 111123359-111123359 on Build GRCh38
rs766188616 Chr.7: 111123060-111123060 on Build GRCh38
rs780343013 Chr.7: 111123233-111123233 on Build GRCh38
rs765065226 Chr.7: 111123318-111123318 on Build GRCh38
rs371746004 Chr.7: 111123064-111123064 on Build GRCh38
rs534069831 Chr.7: 111123114-111123114 on Build GRCh38
rs4601242 Chr.7: 111123242-111123242 on Build GRCh38
rs752690483 Chr.7: 111123142-111123142 on Build GRCh38
rs1039141410 Chr.7: 111123086-111123086 on Build GRCh38
rs145004274 Chr.7: 111123454-111123454 on Build GRCh38
rs997554194 Chr.7: 111123362-111123362 on Build GRCh38
rs900139751 Chr.7: 111123141-111123141 on Build GRCh38
rs1004357844 Chr.7: 111123424-111123424 on Build GRCh38
rs781214401 Chr.7: 111123369-111123369 on Build GRCh38
rs746153863 Chr.7: 111123211-111123211 on Build GRCh38
rs1039189238 Chr.7: 111123309-111123309 on Build GRCh38
rs780719826 Chr.7: 111123528-111123528 on Build GRCh38
rs769370648 Chr.7: 111123451-111123451 on Build GRCh38
rs767632314 Chr.7: 111123490-111123490 on Build GRCh38
rs748649668 Chr.7: 111123514-111123514 on Build GRCh38
rs773818533 Chr.7: 111123325-111123325 on Build GRCh38
rs139382575 Chr.7: 111123150-111123150 on Build GRCh38
rs749799526 Chr.7: 111123234-111123234 on Build GRCh38
rs1029878494 Chr.7: 111123377-111123377 on Build GRCh38
rs563286461 Chr.7: 111123407-111123407 on Build GRCh38
rs770663712 Chr.7: 111123273-111123273 on Build GRCh38
rs769240546 Chr.7: 111123409-111123409 on Build GRCh38
rs760392143 Chr.7: 111123504-111123504 on Build GRCh38
rs951604736 Chr.7: 111123415-111123415 on Build GRCh38
rs146578143 Chr.7: 111123353-111123353 on Build GRCh38
rs756359844 Chr.7: 111123374-111123374 on Build GRCh38
rs201219640 Chr.7: 111123508-111123508 on Build GRCh38
rs151294396 Chr.7: 111123057-111123057 on Build GRCh38
rs901551236 Chr.7: 111123348-111123348 on Build GRCh38
rs150007120 Chr.7: 111123183-111123183 on Build GRCh38
rs779501366 Chr.7: 111123510-111123510 on Build GRCh38
rs982110065 Chr.7: 111123298-111123298 on Build GRCh38
rs898612171 Chr.7: 111123219-111123219 on Build GRCh38
rs1051649493 Chr.7: 111123160-111123160 on Build GRCh38
rs758322925 Chr.7: 111123145-111123145 on Build GRCh38
rs146797927 Chr.7: 111123462-111123462 on Build GRCh38
rs757448665 Chr.7: 111123184-111123184 on Build GRCh38
rs772830687 Chr.7: 111123259-111123259 on Build GRCh38
rs760287037 Chr.7: 111123264-111123264 on Build GRCh38
rs749666399 Chr.7: 111123400-111123400 on Build GRCh38
rs376028588 Chr.7: 111123134-111123134 on Build GRCh38
rs750645851 Chr.7: 111123372-111123372 on Build GRCh38
rs780437672 Chr.7: 111123387-111123387 on Build GRCh38
rs921598665 Chr.7: 111123458-111123458 on Build GRCh38
rs762844497 Chr.7: 111123337-111123337 on Build GRCh38

View Full Product Details