Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120741441-120741985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCND2
Entrez Gene ID
3751
Gene Name
potassium voltage-gated channel subfamily D member 2
Gene Aliases
KV4.2, RK5
UniGene
Hs.654739
Gene Chromosome Location
Chr.7: 120273635-120750333 on Build GRCh38
Transcripts
NM_012281.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3257571 c.1366C>T p.Q456* Chr.7: 120741621-120741621 on Build GRCh38
368992 c.1279-1G>A p.? Chr.7: 120741533-120741533 on Build GRCh38
1084538 c.1301G>C p.R434P Chr.7: 120741556-120741556 on Build GRCh38
1312602 c.1329T>C p.A443A Chr.7: 120741584-120741584 on Build GRCh38
6323938 c.1313G>A p.S438N Chr.7: 120741568-120741568 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760481401 Chr.7: 120741529-120741529 on Build GRCh38
rs1050556143 Chr.7: 120741862-120741862 on Build GRCh38
rs1020247388 Chr.7: 120741625-120741625 on Build GRCh38
rs887575455 Chr.7: 120741716-120741716 on Build GRCh38
rs940916567 Chr.7: 120741825-120741825 on Build GRCh38
rs530629463 Chr.7: 120741530-120741530 on Build GRCh38
rs773978014 Chr.7: 120741503-120741503 on Build GRCh38
rs532958441 Chr.7: 120741793-120741793 on Build GRCh38
rs1042085531 Chr.7: 120741916-120741916 on Build GRCh38
rs144966242 Chr.7: 120741593-120741593 on Build GRCh38
rs568299556 Chr.7: 120741799-120741799 on Build GRCh38
rs775873681 Chr.7: 120741665-120741665 on Build GRCh38
rs191644528 Chr.7: 120741856-120741856 on Build GRCh38
rs929180490 Chr.7: 120741695-120741695 on Build GRCh38
rs141416667 Chr.7: 120741906-120741907 on Build GRCh38
rs7795370 Chr.7: 120741697-120741697 on Build GRCh38
rs184838392 Chr.7: 120741902-120741902 on Build GRCh38
rs181845314 Chr.7: 120741951-120741951 on Build GRCh38
rs770718038 Chr.7: 120741655-120741655 on Build GRCh38
rs777393457 Chr.7: 120741643-120741643 on Build GRCh38
rs753301968 Chr.7: 120741910-120741910 on Build GRCh38
rs923424738 Chr.7: 120741831-120741831 on Build GRCh38
rs758879867 Chr.7: 120741569-120741569 on Build GRCh38
rs776511448 Chr.7: 120741537-120741537 on Build GRCh38
rs932191568 Chr.7: 120741854-120741854 on Build GRCh38
rs957503097 Chr.7: 120741869-120741869 on Build GRCh38
rs919716916 Chr.7: 120741748-120741748 on Build GRCh38
rs1011490341 Chr.7: 120741596-120741596 on Build GRCh38
rs776700377 Chr.7: 120741659-120741659 on Build GRCh38
rs191400154 Chr.7: 120741514-120741514 on Build GRCh38
rs763575205 Chr.7: 120741724-120741724 on Build GRCh38
rs908216061 Chr.7: 120741824-120741824 on Build GRCh38
rs770202109 Chr.7: 120741518-120741519 on Build GRCh38
rs55755276 Chr.7: 120741669-120741669 on Build GRCh38
rs577148321 Chr.7: 120741467-120741467 on Build GRCh38
rs376029594 Chr.7: 120741555-120741555 on Build GRCh38
rs530060196 Chr.7: 120741601-120741601 on Build GRCh38
rs534250238 Chr.7: 120741809-120741809 on Build GRCh38
rs748003036 Chr.7: 120741508-120741508 on Build GRCh38
rs752967153 Chr.7: 120741562-120741562 on Build GRCh38
rs998297518 Chr.7: 120741507-120741507 on Build GRCh38
rs765617019 Chr.7: 120741556-120741556 on Build GRCh38
rs991231690 Chr.7: 120741735-120741735 on Build GRCh38
rs751810388 Chr.7: 120741671-120741671 on Build GRCh38
rs370527455 Chr.7: 120741644-120741644 on Build GRCh38
rs138014445 Chr.7: 120741693-120741693 on Build GRCh38
rs773251474 Chr.7: 120741518-120741518 on Build GRCh38
rs1033181792 Chr.7: 120741845-120741845 on Build GRCh38
rs372791161 Chr.7: 120741668-120741668 on Build GRCh38
rs879300251 Chr.7: 120741906-120741906 on Build GRCh38
rs949723667 Chr.7: 120741613-120741613 on Build GRCh38
rs747777420 Chr.7: 120741612-120741612 on Build GRCh38
rs76856186 Chr.7: 120741914-120741914 on Build GRCh38
rs780298667 Chr.7: 120741647-120741647 on Build GRCh38
rs561063026 Chr.7: 120741600-120741600 on Build GRCh38
rs762221695 Chr.7: 120741678-120741678 on Build GRCh38
rs903206159 Chr.7: 120741946-120741946 on Build GRCh38
rs538058281 Chr.7: 120741749-120741749 on Build GRCh38
rs998576544 Chr.7: 120741785-120741785 on Build GRCh38
rs764312490 Chr.7: 120741570-120741570 on Build GRCh38
rs771924385 Chr.7: 120741632-120741632 on Build GRCh38
rs768238949 Chr.7: 120741491-120741491 on Build GRCh38
rs957617040 Chr.7: 120741580-120741580 on Build GRCh38
rs752136303 Chr.7: 120741573-120741573 on Build GRCh38
rs982410023 Chr.7: 120741763-120741764 on Build GRCh38
rs183970237 Chr.7: 120741662-120741662 on Build GRCh38
rs546140687 Chr.7: 120741918-120741918 on Build GRCh38
rs117232099 Chr.7: 120741624-120741624 on Build GRCh38
rs112141394 Chr.7: 120741763-120741763 on Build GRCh38
rs1039337545 Chr.7: 120741755-120741755 on Build GRCh38
rs571773441 Chr.7: 120741905-120741906 on Build GRCh38
rs556387104 Chr.7: 120741942-120741942 on Build GRCh38
rs998908212 Chr.7: 120741844-120741844 on Build GRCh38
rs190134172 Chr.7: 120741949-120741949 on Build GRCh38
rs748941098 Chr.7: 120741490-120741490 on Build GRCh38
rs767552627 Chr.7: 120741492-120741492 on Build GRCh38
rs116565037 Chr.7: 120741513-120741513 on Build GRCh38
rs200177734 Chr.7: 120741575-120741575 on Build GRCh38
rs1029789077 Chr.7: 120741800-120741800 on Build GRCh38
rs769967630 Chr.7: 120741664-120741664 on Build GRCh38
rs991754029 Chr.7: 120741895-120741895 on Build GRCh38
rs977585712 Chr.7: 120741554-120741554 on Build GRCh38
rs903263688 Chr.7: 120741829-120741829 on Build GRCh38

View Full Product Details