Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120810385-120810894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN12
Entrez Gene ID
23554
Gene Name
tetraspanin 12
Gene Aliases
EVR5, NET-2, NET2, TM4SF12
UniGene
Hs.16529
Gene Chromosome Location
Chr.7: 120787320-120858369 on Build GRCh38
Transcripts
NM_012338.3 XM_005250239.2 XM_011515993.1 XM_011515994.1 XM_017011912.1 XM_017011913.1 XM_017011914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012872 c.434G>A p.W145* Chr.7: 120810497-120810497 on Build GRCh38
316235 c.361-3C>T p.? Chr.7: 120810573-120810573 on Build GRCh38
1084545 c.403A>G p.M135V Chr.7: 120810528-120810528 on Build GRCh38
4546587 c.402G>A p.R134R Chr.7: 120810529-120810529 on Build GRCh38
6489970 c.415G>T p.G139* Chr.7: 120810516-120810516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745764457 Chr.7: 120810610-120810623 on Build GRCh38
rs774526602 Chr.7: 120810576-120810576 on Build GRCh38
rs979503966 Chr.7: 120810680-120810680 on Build GRCh38
rs555438361 Chr.7: 120810773-120810773 on Build GRCh38
rs753234010 Chr.7: 120810654-120810655 on Build GRCh38
rs768183948 Chr.7: 120810746-120810746 on Build GRCh38
rs771260400 Chr.7: 120810502-120810502 on Build GRCh38
rs975810826 Chr.7: 120810839-120810839 on Build GRCh38
rs879013291 Chr.7: 120810625-120810626 on Build GRCh38
rs587777283 Chr.7: 120810518-120810518 on Build GRCh38
rs748228842 Chr.7: 120810579-120810585 on Build GRCh38
rs1055359487 Chr.7: 120810450-120810450 on Build GRCh38
rs772411433 Chr.7: 120810588-120810588 on Build GRCh38
rs568207634 Chr.7: 120810609-120810609 on Build GRCh38
rs1017823493 Chr.7: 120810838-120810838 on Build GRCh38
rs1011039624 Chr.7: 120810663-120810663 on Build GRCh38
rs780145161 Chr.7: 120810565-120810565 on Build GRCh38
rs539463295 Chr.7: 120810688-120810688 on Build GRCh38
rs745746437 Chr.7: 120810560-120810560 on Build GRCh38
rs931657560 Chr.7: 120810844-120810844 on Build GRCh38
rs67615509 Chr.7: 120810656-120810657 on Build GRCh38
rs878935904 Chr.7: 120810624-120810624 on Build GRCh38
rs4621724 Chr.7: 120810724-120810724 on Build GRCh38
rs975773688 Chr.7: 120810641-120810641 on Build GRCh38
rs556151444 Chr.7: 120810696-120810696 on Build GRCh38
rs879086057 Chr.7: 120810625-120810628 on Build GRCh38
rs762144127 Chr.7: 120810463-120810463 on Build GRCh38
rs111673755 Chr.7: 120810784-120810784 on Build GRCh38
rs78990627 Chr.7: 120810715-120810715 on Build GRCh38
rs750419987 Chr.7: 120810621-120810621 on Build GRCh38
rs1008199620 Chr.7: 120810837-120810837 on Build GRCh38
rs369534829 Chr.7: 120810426-120810426 on Build GRCh38
rs776909218 Chr.7: 120810652-120810653 on Build GRCh38
rs763815303 Chr.7: 120810506-120810506 on Build GRCh38
rs886061958 Chr.7: 120810474-120810474 on Build GRCh38
rs768935300 Chr.7: 120810432-120810432 on Build GRCh38
rs768973880 Chr.7: 120810574-120810574 on Build GRCh38
rs761718057 Chr.7: 120810611-120810611 on Build GRCh38
rs966198318 Chr.7: 120810620-120810620 on Build GRCh38
rs3735467 Chr.7: 120810682-120810682 on Build GRCh38
rs1049517322 Chr.7: 120810815-120810815 on Build GRCh38
rs757210351 Chr.7: 120810421-120810421 on Build GRCh38
rs796939885 Chr.7: 120810655-120810655 on Build GRCh38
rs374977768 Chr.7: 120810500-120810500 on Build GRCh38
rs757607505 Chr.7: 120810499-120810499 on Build GRCh38
rs889984937 Chr.7: 120810623-120810623 on Build GRCh38
rs879041524 Chr.7: 120810628-120810629 on Build GRCh38
rs373314103 Chr.7: 120810601-120810601 on Build GRCh38
rs1045868025 Chr.7: 120810505-120810505 on Build GRCh38
rs759104377 Chr.7: 120810491-120810491 on Build GRCh38
rs201849300 Chr.7: 120810418-120810418 on Build GRCh38
rs758303870 Chr.7: 120810847-120810847 on Build GRCh38
rs984956698 Chr.7: 120810440-120810440 on Build GRCh38
rs148990243 Chr.7: 120810632-120810633 on Build GRCh38
rs773703997 Chr.7: 120810484-120810484 on Build GRCh38
rs993915239 Chr.7: 120810522-120810522 on Build GRCh38
rs762345144 Chr.7: 120810605-120810605 on Build GRCh38
rs191813478 Chr.7: 120810430-120810430 on Build GRCh38
rs897917338 Chr.7: 120810777-120810777 on Build GRCh38
rs761501943 Chr.7: 120810811-120810811 on Build GRCh38
rs71170214 Chr.7: 120810655-120810656 on Build GRCh38
rs794726655 Chr.7: 120810571-120810575 on Build GRCh38
rs907636038 Chr.7: 120810778-120810778 on Build GRCh38
rs145676655 Chr.7: 120810564-120810564 on Build GRCh38
rs758026917 Chr.7: 120810501-120810501 on Build GRCh38
rs773722409 Chr.7: 120810590-120810590 on Build GRCh38
rs375399891 Chr.7: 120810417-120810417 on Build GRCh38
rs112555207 Chr.7: 120810624-120810625 on Build GRCh38
rs149800297 Chr.7: 120810510-120810510 on Build GRCh38
rs7805733 Chr.7: 120810604-120810604 on Build GRCh38
rs879161768 Chr.7: 120810626-120810627 on Build GRCh38
rs761175439 Chr.7: 120810596-120810596 on Build GRCh38
rs769907084 Chr.7: 120810427-120810427 on Build GRCh38
rs781585728 Chr.7: 120810552-120810552 on Build GRCh38
rs774651541 Chr.7: 120810462-120810462 on Build GRCh38
rs533965852 Chr.7: 120810665-120810665 on Build GRCh38
rs570663073 Chr.7: 120810770-120810770 on Build GRCh38
rs267601247 Chr.7: 120810471-120810471 on Build GRCh38
rs776682011 Chr.7: 120810604-120810607 on Build GRCh38
rs768053082 Chr.7: 120810467-120810467 on Build GRCh38
rs763875873 Chr.7: 120810607-120810607 on Build GRCh38
rs535238166 Chr.7: 120810720-120810720 on Build GRCh38
rs951972632 Chr.7: 120810772-120810772 on Build GRCh38
rs10260155 Chr.7: 120810672-120810672 on Build GRCh38
rs781248624 Chr.7: 120810422-120810422 on Build GRCh38
rs922970652 Chr.7: 120810841-120810841 on Build GRCh38
rs267607153 Chr.7: 120810512-120810512 on Build GRCh38
rs772331600 Chr.7: 120810579-120810579 on Build GRCh38
rs1027868883 Chr.7: 120810529-120810529 on Build GRCh38
rs887806785 Chr.7: 120810831-120810831 on Build GRCh38
rs907583988 Chr.7: 120810460-120810460 on Build GRCh38
rs751444669 Chr.7: 120810507-120810507 on Build GRCh38
rs746971622 Chr.7: 120810419-120810419 on Build GRCh38
rs945049144 Chr.7: 120810414-120810414 on Build GRCh38
rs369377013 Chr.7: 120810587-120810587 on Build GRCh38
rs767358119 Chr.7: 120810615-120810615 on Build GRCh38
rs749496415 Chr.7: 120810568-120810568 on Build GRCh38
rs769871654 Chr.7: 120810621-120810624 on Build GRCh38

View Full Product Details