Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 120838689-120839198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN12
Entrez Gene ID
23554
Gene Name
tetraspanin 12
Gene Aliases
EVR5, NET-2, NET2, TM4SF12
UniGene
Hs.16529
Gene Chromosome Location
Chr.7: 120787320-120858369 on Build GRCh38
Transcripts
NM_012338.3 XM_005250239.2 XM_011515993.1 XM_011515994.1 XM_017011912.1 XM_017011913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6848764 c.158A>G p.E53G Chr.7: 120838904-120838904 on Build GRCh38
4774586 c.195G>A p.P65P Chr.7: 120838867-120838867 on Build GRCh38
6810594 c.194C>T p.P65L Chr.7: 120838868-120838868 on Build GRCh38
1699263 c.182C>T p.P61L Chr.7: 120838880-120838880 on Build GRCh38
6548675 c.182C>A p.P61H Chr.7: 120838880-120838880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181179087 Chr.7: 120839103-120839103 on Build GRCh38
rs145449060 Chr.7: 120838881-120838881 on Build GRCh38
rs140434334 Chr.7: 120838818-120838818 on Build GRCh38
rs925637676 Chr.7: 120839087-120839087 on Build GRCh38
rs188220108 Chr.7: 120839057-120839057 on Build GRCh38
rs767761287 Chr.7: 120838964-120838964 on Build GRCh38
rs765195097 Chr.7: 120838915-120838915 on Build GRCh38
rs751842331 Chr.7: 120838958-120838958 on Build GRCh38
rs371019535 Chr.7: 120838753-120838753 on Build GRCh38
rs1026189323 Chr.7: 120839154-120839154 on Build GRCh38
rs746413987 Chr.7: 120838862-120838862 on Build GRCh38
rs794726654 Chr.7: 120838843-120838844 on Build GRCh38
rs760578470 Chr.7: 120838729-120838729 on Build GRCh38
rs1055700555 Chr.7: 120839090-120839090 on Build GRCh38
rs550483569 Chr.7: 120839060-120839060 on Build GRCh38
rs751714380 Chr.7: 120838816-120838816 on Build GRCh38
rs904785814 Chr.7: 120838978-120838978 on Build GRCh38
rs945805041 Chr.7: 120839013-120839013 on Build GRCh38
rs961246174 Chr.7: 120838748-120838748 on Build GRCh38
rs933772121 Chr.7: 120838813-120838813 on Build GRCh38
rs772738184 Chr.7: 120838727-120838727 on Build GRCh38
rs745856503 Chr.7: 120838986-120838986 on Build GRCh38
rs780677498 Chr.7: 120838870-120838870 on Build GRCh38
rs747727475 Chr.7: 120838874-120838874 on Build GRCh38
rs571121272 Chr.7: 120839042-120839042 on Build GRCh38
rs552963027 Chr.7: 120839149-120839149 on Build GRCh38
rs994254198 Chr.7: 120839152-120839152 on Build GRCh38
rs1034717750 Chr.7: 120838719-120838719 on Build GRCh38
rs770789679 Chr.7: 120838909-120838909 on Build GRCh38
rs374709391 Chr.7: 120838945-120838945 on Build GRCh38
rs377299760 Chr.7: 120838867-120838867 on Build GRCh38
rs868618204 Chr.7: 120838885-120838885 on Build GRCh38
rs758437173 Chr.7: 120838799-120838799 on Build GRCh38
rs1045593231 Chr.7: 120839138-120839138 on Build GRCh38
rs776633275 Chr.7: 120838759-120838759 on Build GRCh38
rs111640790 Chr.7: 120838988-120838988 on Build GRCh38
rs775398063 Chr.7: 120838922-120838922 on Build GRCh38
rs1014072635 Chr.7: 120839123-120839123 on Build GRCh38
rs935687988 Chr.7: 120839089-120839089 on Build GRCh38
rs752819603 Chr.7: 120838783-120838783 on Build GRCh38
rs866451311 Chr.7: 120838842-120838843 on Build GRCh38
rs17852934 Chr.7: 120838892-120838892 on Build GRCh38
rs781374190 Chr.7: 120838824-120838824 on Build GRCh38
rs952855588 Chr.7: 120838890-120838890 on Build GRCh38
rs1041744533 Chr.7: 120839022-120839022 on Build GRCh38
rs2301782 Chr.7: 120839111-120839111 on Build GRCh38
rs587777285 Chr.7: 120838776-120838776 on Build GRCh38
rs764440915 Chr.7: 120838808-120838808 on Build GRCh38
rs774845824 Chr.7: 120838992-120838993 on Build GRCh38
rs576848610 Chr.7: 120838792-120838792 on Build GRCh38
rs763075900 Chr.7: 120838934-120838934 on Build GRCh38
rs964167833 Chr.7: 120838970-120838970 on Build GRCh38
rs759429271 Chr.7: 120838771-120838771 on Build GRCh38
rs755995749 Chr.7: 120838950-120838950 on Build GRCh38
rs1014123292 Chr.7: 120838749-120838749 on Build GRCh38
rs540584193 Chr.7: 120838712-120838712 on Build GRCh38
rs757374290 Chr.7: 120838960-120838960 on Build GRCh38
rs973105067 Chr.7: 120838827-120838827 on Build GRCh38
rs972570278 Chr.7: 120838807-120838807 on Build GRCh38
rs746574627 Chr.7: 120838897-120838897 on Build GRCh38
rs776239832 Chr.7: 120838911-120838911 on Build GRCh38
rs183489732 Chr.7: 120838887-120838887 on Build GRCh38
rs759302851 Chr.7: 120838912-120838912 on Build GRCh38
rs370601405 Chr.7: 120838931-120838931 on Build GRCh38
rs765321929 Chr.7: 120838781-120838781 on Build GRCh38
rs1025032511 Chr.7: 120838754-120838754 on Build GRCh38

View Full Product Details