Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 166989621-166990113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
UniGene
Hs.651245
Gene Chromosome Location
Chr.1: 166921121-167022214 on Build GRCh38
Transcripts
NM_001286377.1 NM_001286378.1 NM_032858.2 XM_006711583.1 XM_011510067.1 XM_011510068.1 XM_017002602.1 XM_017002603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3477700 c.187G>A p.A63T Chr.1: 166989791-166989791 on Build GRCh38
3477699 c.148G>A p.E50K Chr.1: 166989752-166989752 on Build GRCh38
5735717 c.153G>T p.K51N Chr.1: 166989757-166989757 on Build GRCh38
4817775 c.197G>A p.G66E Chr.1: 166989801-166989801 on Build GRCh38
4768895 c.158A>G p.K53R Chr.1: 166989762-166989762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025988323 Chr.1: 166990030-166990030 on Build GRCh38
rs115895291 Chr.1: 166989953-166989953 on Build GRCh38
rs1018858236 Chr.1: 166989970-166989970 on Build GRCh38
rs907917395 Chr.1: 166989673-166989673 on Build GRCh38
rs531769613 Chr.1: 166989955-166989955 on Build GRCh38
rs772379510 Chr.1: 166989809-166989809 on Build GRCh38
rs903559448 Chr.1: 166989856-166989856 on Build GRCh38
rs761727927 Chr.1: 166989817-166989817 on Build GRCh38
rs144606344 Chr.1: 166989750-166989750 on Build GRCh38
rs747881372 Chr.1: 166989675-166989675 on Build GRCh38
rs983827639 Chr.1: 166990064-166990064 on Build GRCh38
rs370260646 Chr.1: 166989699-166989699 on Build GRCh38
rs188285014 Chr.1: 166989690-166989690 on Build GRCh38
rs572443467 Chr.1: 166989881-166989881 on Build GRCh38
rs749056013 Chr.1: 166989838-166989838 on Build GRCh38
rs751317997 Chr.1: 166989867-166989867 on Build GRCh38
rs776095009 Chr.1: 166989695-166989695 on Build GRCh38
rs771486840 Chr.1: 166989784-166989784 on Build GRCh38
rs573752766 Chr.1: 166989702-166989702 on Build GRCh38
rs781309706 Chr.1: 166990057-166990057 on Build GRCh38
rs201306124 Chr.1: 166989803-166989803 on Build GRCh38
rs960802699 Chr.1: 166990090-166990090 on Build GRCh38
rs543621682 Chr.1: 166990043-166990043 on Build GRCh38
rs531171441 Chr.1: 166989918-166989918 on Build GRCh38
rs754648577 Chr.1: 166989869-166989869 on Build GRCh38
rs919001065 Chr.1: 166989710-166989710 on Build GRCh38
rs779808013 Chr.1: 166989766-166989766 on Build GRCh38
rs371694841 Chr.1: 166989840-166989840 on Build GRCh38
rs758190186 Chr.1: 166989761-166989761 on Build GRCh38
rs753036637 Chr.1: 166989718-166989718 on Build GRCh38
rs764321438 Chr.1: 166989724-166989724 on Build GRCh38
rs750232432 Chr.1: 166989760-166989760 on Build GRCh38
rs945371429 Chr.1: 166989819-166989819 on Build GRCh38
rs757239771 Chr.1: 166989758-166989758 on Build GRCh38
rs999559701 Chr.1: 166989857-166989857 on Build GRCh38
rs776860620 Chr.1: 166989815-166989815 on Build GRCh38
rs983558185 Chr.1: 166989648-166989648 on Build GRCh38
rs774654550 Chr.1: 166989796-166989796 on Build GRCh38
rs965681067 Chr.1: 166989687-166989687 on Build GRCh38
rs1009038953 Chr.1: 166989896-166989896 on Build GRCh38
rs965983851 Chr.1: 166989987-166989987 on Build GRCh38
rs560486398 Chr.1: 166989811-166989811 on Build GRCh38
rs952138770 Chr.1: 166989643-166989643 on Build GRCh38
rs775977856 Chr.1: 166989810-166989810 on Build GRCh38
rs971831837 Chr.1: 166989831-166989831 on Build GRCh38
rs760497592 Chr.1: 166989697-166989698 on Build GRCh38
rs565207441 Chr.1: 166990060-166990060 on Build GRCh38
rs199731527 Chr.1: 166989782-166989782 on Build GRCh38
rs551736834 Chr.1: 166989967-166989967 on Build GRCh38
rs187409599 Chr.1: 166989757-166989757 on Build GRCh38
rs532647800 Chr.1: 166990087-166990087 on Build GRCh38
rs746710942 Chr.1: 166989768-166989768 on Build GRCh38
rs369122088 Chr.1: 166990001-166990002 on Build GRCh38
rs550879943 Chr.1: 166989938-166989938 on Build GRCh38
rs918202757 Chr.1: 166989815-166989815 on Build GRCh38
rs890850165 Chr.1: 166989889-166989889 on Build GRCh38
rs951751157 Chr.1: 166990078-166990078 on Build GRCh38
rs758333507 Chr.1: 166989862-166989862 on Build GRCh38
rs770793006 Chr.1: 166989910-166989910 on Build GRCh38
rs41269702 Chr.1: 166989700-166989700 on Build GRCh38
rs868234878 Chr.1: 166989780-166989780 on Build GRCh38
rs1028281638 Chr.1: 166989990-166989990 on Build GRCh38
rs952085922 Chr.1: 166990020-166990020 on Build GRCh38
rs1000925267 Chr.1: 166989899-166989899 on Build GRCh38
rs1044026406 Chr.1: 166989793-166989793 on Build GRCh38
rs767785618 Chr.1: 166989705-166989705 on Build GRCh38
rs778657307 Chr.1: 166989759-166989759 on Build GRCh38
rs750384293 Chr.1: 166989852-166989852 on Build GRCh38
rs768269478 Chr.1: 166989779-166989779 on Build GRCh38
rs180806108 Chr.1: 166989721-166989721 on Build GRCh38
rs984569395 Chr.1: 166990086-166990087 on Build GRCh38

View Full Product Details