Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81480299-81480840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP4
Entrez Gene ID
2167
Gene Name
fatty acid binding protein 4
Gene Aliases
A-FABP, AFABP, ALBP, HEL-S-104, aP2
UniGene
Hs.391561
Gene Chromosome Location
Chr.8: 81478497-81483238 on Build GRCh38
Transcripts
NM_001442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746888 c.215A>G p.D72G Chr.8: 81480457-81480457 on Build GRCh38
272654 c.109G>A p.A37T Chr.8: 81480563-81480563 on Build GRCh38
5643287 c.132T>C p.S44S Chr.8: 81480540-81480540 on Build GRCh38
232280 c.79G>A p.G27S Chr.8: 81480593-81480593 on Build GRCh38
5016806 c.217G>A p.E73K Chr.8: 81480455-81480455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17848127 Chr.8: 81480640-81480640 on Build GRCh38
rs149072307 Chr.8: 81480477-81480477 on Build GRCh38
rs749161147 Chr.8: 81480545-81480545 on Build GRCh38
rs770448002 Chr.8: 81480472-81480472 on Build GRCh38
rs140925685 Chr.8: 81480592-81480592 on Build GRCh38
rs190755437 Chr.8: 81480793-81480793 on Build GRCh38
rs567997557 Chr.8: 81480778-81480778 on Build GRCh38
rs771035469 Chr.8: 81480549-81480549 on Build GRCh38
rs755892938 Chr.8: 81480528-81480528 on Build GRCh38
rs540850051 Chr.8: 81480365-81480365 on Build GRCh38
rs776736466 Chr.8: 81480609-81480610 on Build GRCh38
rs201912500 Chr.8: 81480551-81480551 on Build GRCh38
rs146187614 Chr.8: 81480447-81480447 on Build GRCh38
rs760702784 Chr.8: 81480499-81480507 on Build GRCh38
rs370351580 Chr.8: 81480790-81480794 on Build GRCh38
rs765731900 Chr.8: 81480468-81480468 on Build GRCh38
rs771796240 Chr.8: 81480573-81480573 on Build GRCh38
rs143128046 Chr.8: 81480387-81480387 on Build GRCh38
rs878895901 Chr.8: 81480585-81480585 on Build GRCh38
rs141947001 Chr.8: 81480695-81480695 on Build GRCh38
rs573313680 Chr.8: 81480361-81480361 on Build GRCh38
rs201051120 Chr.8: 81480772-81480772 on Build GRCh38
rs201471850 Chr.8: 81480552-81480552 on Build GRCh38
rs537703086 Chr.8: 81480740-81480740 on Build GRCh38
rs752469750 Chr.8: 81480524-81480524 on Build GRCh38
rs557452114 Chr.8: 81480769-81480769 on Build GRCh38
rs777044900 Chr.8: 81480455-81480455 on Build GRCh38
rs8192688 Chr.8: 81480614-81480614 on Build GRCh38
rs888266958 Chr.8: 81480814-81480814 on Build GRCh38
rs756397934 Chr.8: 81480645-81480645 on Build GRCh38
rs759740216 Chr.8: 81480614-81480614 on Build GRCh38
rs759508751 Chr.8: 81480354-81480354 on Build GRCh38
rs140243546 Chr.8: 81480644-81480644 on Build GRCh38
rs77097739 Chr.8: 81480601-81480601 on Build GRCh38
rs775032674 Chr.8: 81480582-81480582 on Build GRCh38
rs947293028 Chr.8: 81480721-81480721 on Build GRCh38
rs770962815 Chr.8: 81480599-81480601 on Build GRCh38
rs774055674 Chr.8: 81480443-81480443 on Build GRCh38
rs766412819 Chr.8: 81480486-81480486 on Build GRCh38
rs935352323 Chr.8: 81480704-81480704 on Build GRCh38
rs200054324 Chr.8: 81480583-81480583 on Build GRCh38
rs780959484 Chr.8: 81480414-81480414 on Build GRCh38
rs777576923 Chr.8: 81480435-81480435 on Build GRCh38
rs745432212 Chr.8: 81480567-81480567 on Build GRCh38
rs770030293 Chr.8: 81480590-81480590 on Build GRCh38
rs73283824 Chr.8: 81480817-81480817 on Build GRCh38
rs751756237 Chr.8: 81480396-81480396 on Build GRCh38
rs747950362 Chr.8: 81480420-81480420 on Build GRCh38
rs74275141 Chr.8: 81480783-81480784 on Build GRCh38
rs930238502 Chr.8: 81480755-81480755 on Build GRCh38
rs1044674364 Chr.8: 81480725-81480725 on Build GRCh38
rs60855359 Chr.8: 81480683-81480683 on Build GRCh38
rs987593428 Chr.8: 81480686-81480686 on Build GRCh38
rs777718554 Chr.8: 81480539-81480539 on Build GRCh38
rs746726865 Chr.8: 81480426-81480426 on Build GRCh38
rs113183638 Chr.8: 81480424-81480424 on Build GRCh38
rs780197541 Chr.8: 81480392-81480392 on Build GRCh38
rs528205058 Chr.8: 81480687-81480687 on Build GRCh38
rs751572335 Chr.8: 81480629-81480629 on Build GRCh38
rs781283836 Chr.8: 81480516-81480516 on Build GRCh38
rs758500548 Chr.8: 81480390-81480390 on Build GRCh38
rs755178833 Chr.8: 81480508-81480508 on Build GRCh38
rs755231928 Chr.8: 81480406-81480406 on Build GRCh38
rs571876036 Chr.8: 81480727-81480727 on Build GRCh38
rs751703096 Chr.8: 81480490-81480490 on Build GRCh38
rs924641369 Chr.8: 81480693-81480693 on Build GRCh38
rs748793227 Chr.8: 81480436-81480436 on Build GRCh38
rs113250386 Chr.8: 81480425-81480425 on Build GRCh38
rs1029171180 Chr.8: 81480657-81480657 on Build GRCh38
rs938494205 Chr.8: 81480763-81480763 on Build GRCh38
rs915205902 Chr.8: 81480720-81480720 on Build GRCh38
rs112293693 Chr.8: 81480785-81480785 on Build GRCh38
rs763497424 Chr.8: 81480479-81480479 on Build GRCh38
rs377237239 Chr.8: 81480456-81480456 on Build GRCh38
rs770588259 Chr.8: 81480440-81480440 on Build GRCh38
rs1057074314 Chr.8: 81480707-81480707 on Build GRCh38
rs894360917 Chr.8: 81480714-81480714 on Build GRCh38
rs773886233 Chr.8: 81480768-81480769 on Build GRCh38
rs766928281 Chr.8: 81480616-81480616 on Build GRCh38
rs947483575 Chr.8: 81480684-81480684 on Build GRCh38

View Full Product Details