Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 91018312-91018814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM55A
Entrez Gene ID
55529
Gene Name
transmembrane protein 55A
Gene Aliases
-
UniGene
Hs.202517
Gene Chromosome Location
Chr.8: 90994271-91040975 on Build GRCh38
Transcripts
NM_018710.2 XM_011517152.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3952031 c.377A>T p.N126I Chr.8: 91018499-91018499 on Build GRCh38
4734557 c.368G>A p.R123Q Chr.8: 91018508-91018508 on Build GRCh38
5938218 c.436C>T p.Q146* Chr.8: 91018440-91018440 on Build GRCh38
403027 c.424G>T p.A142S Chr.8: 91018452-91018452 on Build GRCh38
4807629 c.363-5C>T p.? Chr.8: 91018518-91018518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186670086 Chr.8: 91018343-91018343 on Build GRCh38
rs940183481 Chr.8: 91018345-91018345 on Build GRCh38
rs549749203 Chr.8: 91018725-91018725 on Build GRCh38
rs754196713 Chr.8: 91018372-91018372 on Build GRCh38
rs1041642284 Chr.8: 91018474-91018474 on Build GRCh38
rs773824790 Chr.8: 91018509-91018509 on Build GRCh38
rs750548617 Chr.8: 91018418-91018418 on Build GRCh38
rs974236706 Chr.8: 91018788-91018788 on Build GRCh38
rs557826022 Chr.8: 91018775-91018775 on Build GRCh38
rs373308646 Chr.8: 91018534-91018534 on Build GRCh38
rs765829435 Chr.8: 91018390-91018390 on Build GRCh38
rs780455042 Chr.8: 91018420-91018420 on Build GRCh38
rs747961318 Chr.8: 91018454-91018454 on Build GRCh38
rs548326101 Chr.8: 91018377-91018377 on Build GRCh38
rs111971792 Chr.8: 91018534-91018535 on Build GRCh38
rs373311274 Chr.8: 91018450-91018450 on Build GRCh38
rs908602978 Chr.8: 91018771-91018771 on Build GRCh38
rs754066298 Chr.8: 91018689-91018689 on Build GRCh38
rs760709596 Chr.8: 91018520-91018520 on Build GRCh38
rs751782382 Chr.8: 91018430-91018430 on Build GRCh38
rs748826712 Chr.8: 91018506-91018506 on Build GRCh38
rs112268572 Chr.8: 91018439-91018439 on Build GRCh38
rs758488670 Chr.8: 91018419-91018419 on Build GRCh38
rs764412701 Chr.8: 91018369-91018369 on Build GRCh38
rs1032680837 Chr.8: 91018660-91018660 on Build GRCh38
rs762297921 Chr.8: 91018378-91018378 on Build GRCh38
rs111702817 Chr.8: 91018368-91018368 on Build GRCh38
rs902369765 Chr.8: 91018579-91018579 on Build GRCh38
rs558480471 Chr.8: 91018647-91018647 on Build GRCh38
rs770375890 Chr.8: 91018508-91018508 on Build GRCh38
rs537051471 Chr.8: 91018591-91018591 on Build GRCh38
rs531706940 Chr.8: 91018676-91018676 on Build GRCh38
rs754856097 Chr.8: 91018532-91018533 on Build GRCh38
rs773949418 Chr.8: 91018350-91018350 on Build GRCh38
rs769630877 Chr.8: 91018455-91018455 on Build GRCh38
rs955343411 Chr.8: 91018758-91018758 on Build GRCh38
rs776921382 Chr.8: 91018551-91018551 on Build GRCh38
rs141135624 Chr.8: 91018533-91018534 on Build GRCh38
rs369556779 Chr.8: 91018515-91018515 on Build GRCh38
rs755207033 Chr.8: 91018438-91018438 on Build GRCh38
rs1036959868 Chr.8: 91018571-91018571 on Build GRCh38
rs149252123 Chr.8: 91018760-91018760 on Build GRCh38
rs567396529 Chr.8: 91018554-91018554 on Build GRCh38
rs759148544 Chr.8: 91018511-91018511 on Build GRCh38
rs1028359166 Chr.8: 91018726-91018726 on Build GRCh38
rs945571927 Chr.8: 91018464-91018464 on Build GRCh38
rs761120079 Chr.8: 91018351-91018351 on Build GRCh38
rs765848102 Chr.8: 91018564-91018564 on Build GRCh38
rs139160288 Chr.8: 91018490-91018490 on Build GRCh38
rs897098688 Chr.8: 91018581-91018581 on Build GRCh38

View Full Product Details