Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102205194-102205704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2B
Entrez Gene ID
50484
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
MTDPS8A, MTDPS8B, P53R2
UniGene
Hs.512592
Gene Chromosome Location
Chr.8: 102204501-102239118 on Build GRCh38
Transcripts
NM_001172477.1 NM_001172478.1 NM_015713.4

SNP Detail

SNP ID Chromosome Location
rs910971441 Chr.8: 102205479-102205479 on Build GRCh38
rs550154965 Chr.8: 102205679-102205679 on Build GRCh38
rs543127762 Chr.8: 102205411-102205411 on Build GRCh38
rs574147772 Chr.8: 102205387-102205387 on Build GRCh38
rs537438237 Chr.8: 102205316-102205316 on Build GRCh38
rs750126555 Chr.8: 102205636-102205636 on Build GRCh38
rs905327365 Chr.8: 102205354-102205354 on Build GRCh38
rs3204695 Chr.8: 102205254-102205254 on Build GRCh38
rs180944985 Chr.8: 102205393-102205393 on Build GRCh38
rs546403982 Chr.8: 102205603-102205603 on Build GRCh38
rs29000291 Chr.8: 102205248-102205248 on Build GRCh38
rs886062564 Chr.8: 102205632-102205636 on Build GRCh38
rs901081663 Chr.8: 102205233-102205233 on Build GRCh38
rs976598781 Chr.8: 102205486-102205486 on Build GRCh38
rs3735720 Chr.8: 102205458-102205458 on Build GRCh38
rs578154705 Chr.8: 102205678-102205682 on Build GRCh38
rs749539340 Chr.8: 102205284-102205314 on Build GRCh38
rs534028715 Chr.8: 102205217-102205217 on Build GRCh38
rs942460084 Chr.8: 102205480-102205480 on Build GRCh38
rs185809837 Chr.8: 102205503-102205503 on Build GRCh38
rs1052793273 Chr.8: 102205516-102205516 on Build GRCh38
rs29000290 Chr.8: 102205353-102205353 on Build GRCh38
rs144491662 Chr.8: 102205670-102205670 on Build GRCh38
rs189278573 Chr.8: 102205311-102205311 on Build GRCh38
rs886062563 Chr.8: 102205520-102205520 on Build GRCh38
rs886062562 Chr.8: 102205324-102205324 on Build GRCh38
rs905274893 Chr.8: 102205649-102205649 on Build GRCh38
rs962303933 Chr.8: 102205421-102205421 on Build GRCh38
rs3735721 Chr.8: 102205467-102205467 on Build GRCh38
rs759487106 Chr.8: 102205492-102205492 on Build GRCh38
rs972744943 Chr.8: 102205438-102205438 on Build GRCh38
rs553373185 Chr.8: 102205494-102205494 on Build GRCh38
rs1003709364 Chr.8: 102205363-102205363 on Build GRCh38
rs564619226 Chr.8: 102205667-102205667 on Build GRCh38
rs866323859 Chr.8: 102205620-102205620 on Build GRCh38
rs1052425936 Chr.8: 102205302-102205302 on Build GRCh38
rs893772818 Chr.8: 102205558-102205558 on Build GRCh38
rs1035627482 Chr.8: 102205378-102205378 on Build GRCh38
rs535246212 Chr.8: 102205377-102205377 on Build GRCh38
rs935237492 Chr.8: 102205499-102205499 on Build GRCh38

View Full Product Details