Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102218399-102218908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRM2B
Entrez Gene ID
50484
Gene Name
ribonucleotide reductase regulatory TP53 inducible subunit M2B
Gene Aliases
MTDPS8A, MTDPS8B, P53R2
UniGene
Hs.512592
Gene Chromosome Location
Chr.8: 102204501-102239118 on Build GRCh38
Transcripts
NM_001172477.1 NM_001172478.1 NM_015713.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6896655 c.591C>A p.F197L Chr.8: 102218907-102218907 on Build GRCh38
748068 c.610G>T p.A204S Chr.8: 102218888-102218888 on Build GRCh38
1094385 c.669C>A p.L223L Chr.8: 102218829-102218829 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144088421 Chr.8: 102218885-102218885 on Build GRCh38
rs949215273 Chr.8: 102218599-102218599 on Build GRCh38
rs538949495 Chr.8: 102218681-102218681 on Build GRCh38
rs959892227 Chr.8: 102218563-102218563 on Build GRCh38
rs905114768 Chr.8: 102218661-102218661 on Build GRCh38
rs751831616 Chr.8: 102218846-102218846 on Build GRCh38
rs534769738 Chr.8: 102218502-102218502 on Build GRCh38
rs775139335 Chr.8: 102218793-102218793 on Build GRCh38
rs965903359 Chr.8: 102218427-102218427 on Build GRCh38
rs894092642 Chr.8: 102218453-102218453 on Build GRCh38
rs183750253 Chr.8: 102218434-102218434 on Build GRCh38
rs863224193 Chr.8: 102218836-102218836 on Build GRCh38
rs515726195 Chr.8: 102218866-102218866 on Build GRCh38
rs515726196 Chr.8: 102218827-102218827 on Build GRCh38
rs771767658 Chr.8: 102218786-102218786 on Build GRCh38
rs770634759 Chr.8: 102218773-102218773 on Build GRCh38
rs77541058 Chr.8: 102218568-102218568 on Build GRCh38
rs557256275 Chr.8: 102218775-102218775 on Build GRCh38
rs924642454 Chr.8: 102218504-102218504 on Build GRCh38
rs573218615 Chr.8: 102218726-102218726 on Build GRCh38
rs200933469 Chr.8: 102218763-102218763 on Build GRCh38
rs905583122 Chr.8: 102218521-102218521 on Build GRCh38
rs765605411 Chr.8: 102218800-102218800 on Build GRCh38
rs74984203 Chr.8: 102218727-102218727 on Build GRCh38
rs761703030 Chr.8: 102218491-102218491 on Build GRCh38
rs763191678 Chr.8: 102218823-102218823 on Build GRCh38
rs911227481 Chr.8: 102218665-102218665 on Build GRCh38
rs1035886987 Chr.8: 102218544-102218544 on Build GRCh38
rs976862309 Chr.8: 102218743-102218743 on Build GRCh38
rs77691560 Chr.8: 102218725-102218725 on Build GRCh38
rs952646724 Chr.8: 102218638-102218638 on Build GRCh38
rs984056148 Chr.8: 102218651-102218651 on Build GRCh38
rs1042764302 Chr.8: 102218511-102218511 on Build GRCh38
rs863224914 Chr.8: 102218862-102218863 on Build GRCh38
rs202184126 Chr.8: 102218788-102218788 on Build GRCh38
rs28927370 Chr.8: 102218631-102218631 on Build GRCh38
rs536681204 Chr.8: 102218437-102218437 on Build GRCh38
rs774104496 Chr.8: 102218774-102218774 on Build GRCh38
rs755176920 Chr.8: 102218849-102218849 on Build GRCh38
rs28928595 Chr.8: 102218449-102218449 on Build GRCh38
rs1028978866 Chr.8: 102218451-102218451 on Build GRCh38
rs765866546 Chr.8: 102218787-102218787 on Build GRCh38
rs964056638 Chr.8: 102218698-102218698 on Build GRCh38
rs377299104 Chr.8: 102218807-102218807 on Build GRCh38
rs3018505 Chr.8: 102218533-102218533 on Build GRCh38
rs748974152 Chr.8: 102218766-102218766 on Build GRCh38
rs764984989 Chr.8: 102218506-102218506 on Build GRCh38
rs773652531 Chr.8: 102218802-102218802 on Build GRCh38
rs867576138 Chr.8: 102218839-102218839 on Build GRCh38
rs575109470 Chr.8: 102218852-102218852 on Build GRCh38
rs28928594 Chr.8: 102218478-102218478 on Build GRCh38
rs978756791 Chr.8: 102218484-102218484 on Build GRCh38
rs917359526 Chr.8: 102218580-102218580 on Build GRCh38
rs1003850483 Chr.8: 102218796-102218796 on Build GRCh38
rs762387155 Chr.8: 102218799-102218799 on Build GRCh38
rs752648938 Chr.8: 102218858-102218858 on Build GRCh38

View Full Product Details