Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 108469695-108470225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC2
Entrez Gene ID
9694
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
KIAA0103, TTC35
UniGene
Hs.654351
Gene Chromosome Location
Chr.8: 108443624-108486907 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1094800 c.391C>T p.R131* Chr.8: 108469853-108469853 on Build GRCh38
6269513 c.367G>T p.A123S Chr.8: 108469829-108469829 on Build GRCh38
1230786 c.380A>C p.K127T Chr.8: 108469842-108469842 on Build GRCh38
39527 c.412G>A p.V138M Chr.8: 108469874-108469874 on Build GRCh38
3833855 c.458G>C p.G153A Chr.8: 108470070-108470070 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752102266 Chr.8: 108470168-108470168 on Build GRCh38
rs368499508 Chr.8: 108470171-108470171 on Build GRCh38
rs758854858 Chr.8: 108470158-108470158 on Build GRCh38
rs964569172 Chr.8: 108469841-108469841 on Build GRCh38
rs751566215 Chr.8: 108469812-108469812 on Build GRCh38
rs536795785 Chr.8: 108470018-108470018 on Build GRCh38
rs755668477 Chr.8: 108469942-108469943 on Build GRCh38
rs17851827 Chr.8: 108469896-108469896 on Build GRCh38
rs574047734 Chr.8: 108470007-108470007 on Build GRCh38
rs369356984 Chr.8: 108469816-108469816 on Build GRCh38
rs753400222 Chr.8: 108469946-108469947 on Build GRCh38
rs754135623 Chr.8: 108469947-108469947 on Build GRCh38
rs577795355 Chr.8: 108470195-108470195 on Build GRCh38
rs756422174 Chr.8: 108469831-108469831 on Build GRCh38
rs921275109 Chr.8: 108470101-108470101 on Build GRCh38
rs566438057 Chr.8: 108470050-108470050 on Build GRCh38
rs762908717 Chr.8: 108469791-108469791 on Build GRCh38
rs747947041 Chr.8: 108470047-108470048 on Build GRCh38
rs760936568 Chr.8: 108469945-108469945 on Build GRCh38
rs761779629 Chr.8: 108470062-108470062 on Build GRCh38
rs769468091 Chr.8: 108469871-108469871 on Build GRCh38
rs938210482 Chr.8: 108470041-108470041 on Build GRCh38
rs763960443 Chr.8: 108469810-108469810 on Build GRCh38
rs75596185 Chr.8: 108469763-108469763 on Build GRCh38
rs772170315 Chr.8: 108470068-108470068 on Build GRCh38
rs780736494 Chr.8: 108469868-108469868 on Build GRCh38
rs557149789 Chr.8: 108469887-108469887 on Build GRCh38
rs1006189589 Chr.8: 108470145-108470145 on Build GRCh38
rs773373450 Chr.8: 108470075-108470075 on Build GRCh38
rs775306720 Chr.8: 108469881-108469881 on Build GRCh38
rs976685091 Chr.8: 108470066-108470066 on Build GRCh38
rs753064769 Chr.8: 108470151-108470151 on Build GRCh38
rs372330288 Chr.8: 108469826-108469826 on Build GRCh38
rs769542128 Chr.8: 108470089-108470089 on Build GRCh38
rs767775109 Chr.8: 108469824-108469824 on Build GRCh38
rs749642246 Chr.8: 108469839-108469839 on Build GRCh38
rs1043009987 Chr.8: 108470191-108470191 on Build GRCh38
rs932576075 Chr.8: 108469718-108469718 on Build GRCh38
rs887800354 Chr.8: 108470120-108470120 on Build GRCh38
rs1017458296 Chr.8: 108469814-108469814 on Build GRCh38
rs769343266 Chr.8: 108470034-108470034 on Build GRCh38
rs1006581775 Chr.8: 108469783-108469783 on Build GRCh38
rs745463952 Chr.8: 108469870-108469870 on Build GRCh38
rs774211750 Chr.8: 108470057-108470057 on Build GRCh38
rs932545019 Chr.8: 108470182-108470182 on Build GRCh38
rs765002918 Chr.8: 108469780-108469780 on Build GRCh38
rs779262950 Chr.8: 108469850-108469850 on Build GRCh38
rs376058448 Chr.8: 108469787-108469787 on Build GRCh38
rs997310242 Chr.8: 108469845-108469845 on Build GRCh38
rs1038975762 Chr.8: 108470147-108470147 on Build GRCh38
rs778231453 Chr.8: 108470166-108470166 on Build GRCh38
rs377098175 Chr.8: 108469837-108469837 on Build GRCh38
rs764180984 Chr.8: 108469994-108469994 on Build GRCh38
rs758609319 Chr.8: 108469758-108469758 on Build GRCh38
rs761902165 Chr.8: 108469939-108469939 on Build GRCh38
rs755263265 Chr.8: 108469949-108469949 on Build GRCh38
rs765460299 Chr.8: 108470117-108470117 on Build GRCh38
rs758918584 Chr.8: 108470012-108470012 on Build GRCh38
rs1056648853 Chr.8: 108470053-108470053 on Build GRCh38
rs369735522 Chr.8: 108470155-108470155 on Build GRCh38
rs748726090 Chr.8: 108469860-108469860 on Build GRCh38
rs766518418 Chr.8: 108470108-108470108 on Build GRCh38
rs532395513 Chr.8: 108470173-108470173 on Build GRCh38
rs779649104 Chr.8: 108469943-108469944 on Build GRCh38
rs376985145 Chr.8: 108469977-108469977 on Build GRCh38
rs536838549 Chr.8: 108469886-108469886 on Build GRCh38
rs1052909573 Chr.8: 108469801-108469801 on Build GRCh38
rs759859139 Chr.8: 108470116-108470116 on Build GRCh38
rs753999031 Chr.8: 108470112-108470112 on Build GRCh38
rs1030080319 Chr.8: 108469854-108469854 on Build GRCh38
rs998999930 Chr.8: 108469793-108469793 on Build GRCh38
rs1002478435 Chr.8: 108469924-108469924 on Build GRCh38
rs774154743 Chr.8: 108469923-108469923 on Build GRCh38
rs754634918 Chr.8: 108470027-108470028 on Build GRCh38
rs760811980 Chr.8: 108470081-108470081 on Build GRCh38
rs767652138 Chr.8: 108469941-108469941 on Build GRCh38
rs750957598 Chr.8: 108469937-108469938 on Build GRCh38
rs770422828 Chr.8: 108469862-108469862 on Build GRCh38
rs778481680 Chr.8: 108470038-108470038 on Build GRCh38
rs969600652 Chr.8: 108469853-108469853 on Build GRCh38
rs988569933 Chr.8: 108469872-108469872 on Build GRCh38
rs779543482 Chr.8: 108470038-108470038 on Build GRCh38
rs918092078 Chr.8: 108470056-108470056 on Build GRCh38
rs768326325 Chr.8: 108470054-108470054 on Build GRCh38
rs755460236 Chr.8: 108469844-108469844 on Build GRCh38
rs952937655 Chr.8: 108469991-108469991 on Build GRCh38
rs779398919 Chr.8: 108469951-108469951 on Build GRCh38
rs908664535 Chr.8: 108470047-108470047 on Build GRCh38
rs1013891522 Chr.8: 108469821-108469821 on Build GRCh38
rs997383759 Chr.8: 108470187-108470187 on Build GRCh38
rs926876584 Chr.8: 108470037-108470038 on Build GRCh38
rs753129204 Chr.8: 108469962-108469962 on Build GRCh38
rs1048234186 Chr.8: 108469741-108469741 on Build GRCh38

View Full Product Details