Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116942684-116943181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARD
Entrez Gene ID
441376
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
C8orf85
UniGene
Hs.437551
Gene Chromosome Location
Chr.8: 116937960-116944000 on Build GRCh38
Transcripts
NM_001025357.2 XM_011517043.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5805615 c.455C>A p.A152E Chr.8: 116942688-116942688 on Build GRCh38
3432064 c.456G>A p.A152A Chr.8: 116942689-116942689 on Build GRCh38
1246998 c.462G>A p.P154P Chr.8: 116942695-116942695 on Build GRCh38
1454452 c.455C>T p.A152V Chr.8: 116942688-116942688 on Build GRCh38
3164903 c.461C>T p.P154L Chr.8: 116942694-116942694 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs896357701 Chr.8: 116942856-116942856 on Build GRCh38
rs942325538 Chr.8: 116943061-116943061 on Build GRCh38
rs1030906281 Chr.8: 116943016-116943016 on Build GRCh38
rs1047458449 Chr.8: 116942721-116942721 on Build GRCh38
rs4629901 Chr.8: 116942766-116942766 on Build GRCh38
rs933530899 Chr.8: 116942779-116942779 on Build GRCh38
rs10105065 Chr.8: 116943027-116943027 on Build GRCh38
rs79054877 Chr.8: 116943022-116943022 on Build GRCh38
rs898360381 Chr.8: 116942709-116942709 on Build GRCh38
rs200936571 Chr.8: 116942728-116942728 on Build GRCh38
rs559415401 Chr.8: 116943029-116943029 on Build GRCh38
rs545762376 Chr.8: 116942948-116942948 on Build GRCh38
rs55952851 Chr.8: 116943021-116943021 on Build GRCh38
rs929840254 Chr.8: 116942824-116942824 on Build GRCh38
rs767022559 Chr.8: 116943155-116943155 on Build GRCh38
rs1037856836 Chr.8: 116942937-116942938 on Build GRCh38
rs564508367 Chr.8: 116942963-116942963 on Build GRCh38
rs957799875 Chr.8: 116942767-116942767 on Build GRCh38
rs75807544 Chr.8: 116942939-116942939 on Build GRCh38
rs537012355 Chr.8: 116942733-116942733 on Build GRCh38
rs949342235 Chr.8: 116942860-116942860 on Build GRCh38
rs987038603 Chr.8: 116943053-116943053 on Build GRCh38
rs542043329 Chr.8: 116942801-116942801 on Build GRCh38
rs1018156652 Chr.8: 116943030-116943030 on Build GRCh38
rs149590322 Chr.8: 116942713-116942713 on Build GRCh38
rs767475876 Chr.8: 116942726-116942726 on Build GRCh38
rs954747302 Chr.8: 116943052-116943053 on Build GRCh38
rs761085779 Chr.8: 116943050-116943050 on Build GRCh38
rs989069015 Chr.8: 116942771-116942771 on Build GRCh38
rs1045737486 Chr.8: 116942936-116942936 on Build GRCh38
rs529885246 Chr.8: 116943054-116943054 on Build GRCh38
rs964756940 Chr.8: 116943044-116943045 on Build GRCh38
rs1010744215 Chr.8: 116943028-116943028 on Build GRCh38
rs751404334 Chr.8: 116942717-116942717 on Build GRCh38
rs148356037 Chr.8: 116942772-116942772 on Build GRCh38
rs973825591 Chr.8: 116942803-116942803 on Build GRCh38
rs557362478 Chr.8: 116942999-116943000 on Build GRCh38
rs1046450922 Chr.8: 116942973-116942973 on Build GRCh38
rs919763677 Chr.8: 116942804-116942804 on Build GRCh38
rs919629010 Chr.8: 116943157-116943157 on Build GRCh38
rs755288009 Chr.8: 116942738-116942738 on Build GRCh38
rs575348899 Chr.8: 116942947-116942947 on Build GRCh38
rs574972421 Chr.8: 116942786-116942786 on Build GRCh38
rs7821541 Chr.8: 116943113-116943113 on Build GRCh38
rs756999480 Chr.8: 116942720-116942720 on Build GRCh38
rs916753148 Chr.8: 116943072-116943072 on Build GRCh38
rs780241540 Chr.8: 116942729-116942729 on Build GRCh38
rs4339677 Chr.8: 116943012-116943012 on Build GRCh38
rs955658908 Chr.8: 116942780-116942780 on Build GRCh38
rs182681943 Chr.8: 116942785-116942785 on Build GRCh38
rs973531414 Chr.8: 116943065-116943065 on Build GRCh38
rs536106214 Chr.8: 116942714-116942714 on Build GRCh38
rs558610080 Chr.8: 116942746-116942746 on Build GRCh38
rs763465588 Chr.8: 116943152-116943152 on Build GRCh38
rs1020464598 Chr.8: 116943039-116943039 on Build GRCh38
rs1030724370 Chr.8: 116943051-116943051 on Build GRCh38
rs964831888 Chr.8: 116942757-116942757 on Build GRCh38
rs906199696 Chr.8: 116942990-116942990 on Build GRCh38
rs371456182 Chr.8: 116942707-116942707 on Build GRCh38
rs1046163769 Chr.8: 116943140-116943140 on Build GRCh38
rs1046894339 Chr.8: 116942855-116942855 on Build GRCh38
rs117182846 Chr.8: 116942727-116942727 on Build GRCh38
rs973792857 Chr.8: 116943115-116943115 on Build GRCh38

View Full Product Details